Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1242866-1243099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM132A
Entrez Gene ID
388581
Gene Name
family with sequence similarity 132 member A
Gene Aliases
C1QDC2, C1QTNF12, CTRP12
UniGene
Hs.197613
Gene Chromosome Location
Chr.1: 1242446-1247218 on Build GRCh38
Transcripts
NM_001014980.2 XM_011541435.2 XM_011541436.2 XM_011541437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
894517 c.735C>T p.C245C Chr.1: 1242910-1242910 on Build GRCh38
1294791 c.757G>C p.E253Q Chr.1: 1242888-1242888 on Build GRCh38
4567515 c.743_744CC>TT p.A248V Chr.1: 1242901-1242902 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757308731 Chr.1: 1242961-1242961 on Build GRCh38
rs199863277 Chr.1: 1243023-1243023 on Build GRCh38
rs767386845 Chr.1: 1242962-1242962 on Build GRCh38
rs779604214 Chr.1: 1242892-1242892 on Build GRCh38
rs748628092 Chr.1: 1243034-1243034 on Build GRCh38
rs766564916 Chr.1: 1242945-1242945 on Build GRCh38
rs114921901 Chr.1: 1243073-1243073 on Build GRCh38
rs773609972 Chr.1: 1242929-1242929 on Build GRCh38
rs772407404 Chr.1: 1242910-1242910 on Build GRCh38
rs564944837 Chr.1: 1243024-1243024 on Build GRCh38
rs1012007794 Chr.1: 1242995-1242995 on Build GRCh38
rs778446449 Chr.1: 1243033-1243033 on Build GRCh38
rs750309414 Chr.1: 1243012-1243012 on Build GRCh38
rs77651834 Chr.1: 1242886-1242886 on Build GRCh38
rs751563244 Chr.1: 1242953-1242953 on Build GRCh38
rs368696075 Chr.1: 1243043-1243043 on Build GRCh38
rs201644366 Chr.1: 1242900-1242900 on Build GRCh38
rs748763798 Chr.1: 1242896-1242896 on Build GRCh38
rs755734976 Chr.1: 1243013-1243013 on Build GRCh38
rs764049219 Chr.1: 1242951-1242951 on Build GRCh38
rs1053283149 Chr.1: 1242983-1242983 on Build GRCh38
rs924169061 Chr.1: 1242988-1242988 on Build GRCh38
rs775751000 Chr.1: 1243068-1243068 on Build GRCh38
rs569856478 Chr.1: 1243005-1243005 on Build GRCh38
rs369515180 Chr.1: 1242901-1242901 on Build GRCh38
rs1011247924 Chr.1: 1243050-1243050 on Build GRCh38
rs891290043 Chr.1: 1242926-1242926 on Build GRCh38
rs149401953 Chr.1: 1243062-1243062 on Build GRCh38
rs374211737 Chr.1: 1243028-1243028 on Build GRCh38
rs776745340 Chr.1: 1243055-1243055 on Build GRCh38
rs772635809 Chr.1: 1243039-1243039 on Build GRCh38
rs759535031 Chr.1: 1242950-1242950 on Build GRCh38
rs550146828 Chr.1: 1242974-1242974 on Build GRCh38
rs533260195 Chr.1: 1242946-1242946 on Build GRCh38
rs143023194 Chr.1: 1243061-1243061 on Build GRCh38
rs773548968 Chr.1: 1243042-1243042 on Build GRCh38
rs201127831 Chr.1: 1243016-1243016 on Build GRCh38
rs760886471 Chr.1: 1242934-1242934 on Build GRCh38
rs778406520 Chr.1: 1242957-1242957 on Build GRCh38
rs1051968430 Chr.1: 1242980-1242980 on Build GRCh38
rs762119912 Chr.1: 1242909-1242909 on Build GRCh38
rs754469242 Chr.1: 1243024-1243024 on Build GRCh38

View Full Product Details