Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30655496-30655721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX16
Entrez Gene ID
8449
Gene Name
DEAH-box helicase 16
Gene Aliases
DBP2, DDX16, PRO2014, PRP8, PRPF2, Prp2
UniGene
Hs.485060
Gene Chromosome Location
Chr.6: 30653119-30673053 on Build GRCh38
Transcripts
NM_001164239.1 NM_003587.4 XM_011514938.2 XM_011514939.2 XM_011514940.2 XM_011514941.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6454773 c.2559C>G p.I853M Chr.6: 30655537-30655537 on Build GRCh38
4353901 c.1123C>T p.R375* Chr.6: 30655530-30655530 on Build GRCh38
6454775 c.1116C>G p.I372M Chr.6: 30655537-30655537 on Build GRCh38
6471055 c.2527G>A p.V843M Chr.6: 30655569-30655569 on Build GRCh38
3251596 c.2566C>T p.R856* Chr.6: 30655530-30655530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756617549 Chr.6: 30655614-30655614 on Build GRCh38
rs767645494 Chr.6: 30655652-30655652 on Build GRCh38
rs547096325 Chr.6: 30655653-30655653 on Build GRCh38
rs566028389 Chr.6: 30655630-30655630 on Build GRCh38
rs757650892 Chr.6: 30655589-30655589 on Build GRCh38
rs144182917 Chr.6: 30655655-30655655 on Build GRCh38
rs763910556 Chr.6: 30655578-30655578 on Build GRCh38
rs74945943 Chr.6: 30655562-30655562 on Build GRCh38
rs535048173 Chr.6: 30655631-30655631 on Build GRCh38
rs749334371 Chr.6: 30655544-30655544 on Build GRCh38
rs369004846 Chr.6: 30655640-30655640 on Build GRCh38
rs555409643 Chr.6: 30655537-30655537 on Build GRCh38
rs774542988 Chr.6: 30655546-30655546 on Build GRCh38
rs770792084 Chr.6: 30655643-30655643 on Build GRCh38
rs867597399 Chr.6: 30655559-30655562 on Build GRCh38
rs952062693 Chr.6: 30655694-30655695 on Build GRCh38
rs891765304 Chr.6: 30655530-30655530 on Build GRCh38
rs557196116 Chr.6: 30655689-30655689 on Build GRCh38
rs577909976 Chr.6: 30655648-30655648 on Build GRCh38
rs376877884 Chr.6: 30655601-30655601 on Build GRCh38
rs962206661 Chr.6: 30655659-30655659 on Build GRCh38
rs972139090 Chr.6: 30655661-30655661 on Build GRCh38
rs964917195 Chr.6: 30655676-30655676 on Build GRCh38
rs770574744 Chr.6: 30655559-30655559 on Build GRCh38
rs953731909 Chr.6: 30655621-30655621 on Build GRCh38
rs759353228 Chr.6: 30655560-30655560 on Build GRCh38
rs762543794 Chr.6: 30655572-30655572 on Build GRCh38
rs767722954 Chr.6: 30655599-30655599 on Build GRCh38
rs1029707149 Chr.6: 30655604-30655604 on Build GRCh38
rs557865026 Chr.6: 30655642-30655642 on Build GRCh38
rs746813060 Chr.6: 30655557-30655557 on Build GRCh38
rs779954614 Chr.6: 30655538-30655538 on Build GRCh38
rs775192444 Chr.6: 30655570-30655570 on Build GRCh38
rs973093160 Chr.6: 30655677-30655677 on Build GRCh38
rs780515536 Chr.6: 30655616-30655616 on Build GRCh38

View Full Product Details