Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27198047-27198300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA13
Entrez Gene ID
3209
Gene Name
homeobox A13
Gene Aliases
HOX1, HOX1J
UniGene
Hs.592172
Gene Chromosome Location
Chr.7: 27196880-27200106 on Build GRCh38
Transcripts
NM_000522.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4943635 c.1104C>G p.I368M Chr.7: 27198261-27198261 on Build GRCh38
5872549 c.1073C>T p.T358M Chr.7: 27198292-27198292 on Build GRCh38
5734919 c.1136_1137delAA p.K379fs*9 Chr.7: 27198228-27198229 on Build GRCh38
391499 c.1116delC p.N372fs*13 Chr.7: 27198249-27198249 on Build GRCh38
4690960 c.1142T>G p.I381S Chr.7: 27198223-27198223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749173832 Chr.7: 27198206-27198206 on Build GRCh38
rs992295437 Chr.7: 27198143-27198143 on Build GRCh38
rs138250182 Chr.7: 27198147-27198147 on Build GRCh38
rs745642211 Chr.7: 27198136-27198136 on Build GRCh38
rs753166426 Chr.7: 27198153-27198153 on Build GRCh38
rs774224020 Chr.7: 27198236-27198236 on Build GRCh38
rs775132909 Chr.7: 27198205-27198205 on Build GRCh38
rs1031128034 Chr.7: 27198123-27198123 on Build GRCh38
rs774811041 Chr.7: 27198176-27198176 on Build GRCh38
rs772586223 Chr.7: 27198243-27198243 on Build GRCh38
rs766983635 Chr.7: 27198264-27198264 on Build GRCh38
rs368423493 Chr.7: 27198270-27198270 on Build GRCh38
rs770101262 Chr.7: 27198201-27198201 on Build GRCh38
rs375825948 Chr.7: 27198174-27198174 on Build GRCh38
rs41303372 Chr.7: 27198085-27198085 on Build GRCh38
rs536675514 Chr.7: 27198171-27198171 on Build GRCh38
rs527841320 Chr.7: 27198115-27198115 on Build GRCh38
rs986802238 Chr.7: 27198090-27198090 on Build GRCh38
rs368079328 Chr.7: 27198149-27198149 on Build GRCh38
rs918920181 Chr.7: 27198128-27198128 on Build GRCh38
rs766125491 Chr.7: 27198273-27198273 on Build GRCh38
rs772829383 Chr.7: 27198268-27198268 on Build GRCh38
rs556198527 Chr.7: 27198120-27198120 on Build GRCh38
rs758828890 Chr.7: 27198159-27198159 on Build GRCh38
rs971738459 Chr.7: 27198127-27198127 on Build GRCh38
rs746043394 Chr.7: 27198177-27198177 on Build GRCh38
rs104894019 Chr.7: 27198258-27198258 on Build GRCh38
rs909428055 Chr.7: 27198119-27198119 on Build GRCh38
rs756935287 Chr.7: 27198075-27198075 on Build GRCh38
rs768443549 Chr.7: 27198217-27198217 on Build GRCh38
rs1046132538 Chr.7: 27198244-27198244 on Build GRCh38
rs948181722 Chr.7: 27198181-27198181 on Build GRCh38
rs121912542 Chr.7: 27198251-27198251 on Build GRCh38
rs763736448 Chr.7: 27198156-27198156 on Build GRCh38
rs757359297 Chr.7: 27198173-27198173 on Build GRCh38
rs868322285 Chr.7: 27198224-27198224 on Build GRCh38
rs199530619 Chr.7: 27198148-27198148 on Build GRCh38

View Full Product Details