Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240895568-240895841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf54
Entrez Gene ID
79919
Gene Name
chromosome 2 open reading frame 54
Gene Aliases
-
UniGene
Hs.193745
Gene Chromosome Location
Chr.2: 240886048-240896889 on Build GRCh38
Transcripts
NM_001085437.2 XM_011511877.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6615876 c.174C>T p.I58I Chr.2: 240895824-240895824 on Build GRCh38
2909984 c.204C>A p.F68L Chr.2: 240895794-240895794 on Build GRCh38
6861515 c.240C>T p.D80D Chr.2: 240895758-240895758 on Build GRCh38
5686646 c.409G>C p.A137P Chr.2: 240895589-240895589 on Build GRCh38
3580007 c.187C>T p.R63C Chr.2: 240895811-240895811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746979237 Chr.2: 240895731-240895731 on Build GRCh38
rs764873006 Chr.2: 240895655-240895655 on Build GRCh38
rs903726671 Chr.2: 240895774-240895774 on Build GRCh38
rs754262913 Chr.2: 240895698-240895698 on Build GRCh38
rs761199177 Chr.2: 240895768-240895768 on Build GRCh38
rs779165641 Chr.2: 240895783-240895783 on Build GRCh38
rs118111655 Chr.2: 240895824-240895824 on Build GRCh38
rs756591935 Chr.2: 240895594-240895594 on Build GRCh38
rs372135948 Chr.2: 240895753-240895753 on Build GRCh38
rs761434448 Chr.2: 240895815-240895815 on Build GRCh38
rs1023889045 Chr.2: 240895664-240895664 on Build GRCh38
rs375462480 Chr.2: 240895777-240895777 on Build GRCh38
rs755447519 Chr.2: 240895781-240895781 on Build GRCh38
rs781129095 Chr.2: 240895796-240895796 on Build GRCh38
rs867189724 Chr.2: 240895823-240895823 on Build GRCh38
rs776339450 Chr.2: 240895740-240895740 on Build GRCh38
rs200160556 Chr.2: 240895622-240895622 on Build GRCh38
rs556360296 Chr.2: 240895808-240895808 on Build GRCh38
rs574116147 Chr.2: 240895763-240895763 on Build GRCh38
rs770813596 Chr.2: 240895793-240895793 on Build GRCh38
rs1056511303 Chr.2: 240895700-240895700 on Build GRCh38
rs556004457 Chr.2: 240895737-240895737 on Build GRCh38
rs909393298 Chr.2: 240895626-240895626 on Build GRCh38
rs940859372 Chr.2: 240895657-240895657 on Build GRCh38
rs370356906 Chr.2: 240895648-240895648 on Build GRCh38
rs781580726 Chr.2: 240895589-240895589 on Build GRCh38
rs532509457 Chr.2: 240895799-240895799 on Build GRCh38
rs757869028 Chr.2: 240895665-240895665 on Build GRCh38
rs368019894 Chr.2: 240895671-240895671 on Build GRCh38
rs779029413 Chr.2: 240895715-240895715 on Build GRCh38
rs773697084 Chr.2: 240895812-240895812 on Build GRCh38
rs201848788 Chr.2: 240895600-240895600 on Build GRCh38
rs775049269 Chr.2: 240895757-240895757 on Build GRCh38
rs200698547 Chr.2: 240895730-240895730 on Build GRCh38
rs750925971 Chr.2: 240895765-240895765 on Build GRCh38
rs371994263 Chr.2: 240895692-240895692 on Build GRCh38
rs777658838 Chr.2: 240895788-240895788 on Build GRCh38
rs146162100 Chr.2: 240895693-240895693 on Build GRCh38
rs373507498 Chr.2: 240895633-240895633 on Build GRCh38
rs766973976 Chr.2: 240895820-240895820 on Build GRCh38
rs748237368 Chr.2: 240895721-240895721 on Build GRCh38
rs375087355 Chr.2: 240895668-240895668 on Build GRCh38
rs746191324 Chr.2: 240895590-240895590 on Build GRCh38
rs772085214 Chr.2: 240895727-240895727 on Build GRCh38
rs372524645 Chr.2: 240895790-240895790 on Build GRCh38
rs377398179 Chr.2: 240895810-240895810 on Build GRCh38
rs762489462 Chr.2: 240895762-240895762 on Build GRCh38
rs759355996 Chr.2: 240895751-240895751 on Build GRCh38
rs748211742 Chr.2: 240895619-240895619 on Build GRCh38
rs780389400 Chr.2: 240895595-240895595 on Build GRCh38
rs769015681 Chr.2: 240895614-240895614 on Build GRCh38
rs768362452 Chr.2: 240895811-240895811 on Build GRCh38
rs368820404 Chr.2: 240895780-240895780 on Build GRCh38
rs760488533 Chr.2: 240895628-240895628 on Build GRCh38
rs755214672 Chr.2: 240895699-240895699 on Build GRCh38
rs773072905 Chr.2: 240895623-240895623 on Build GRCh38
rs374762608 Chr.2: 240895615-240895615 on Build GRCh38
rs371171789 Chr.2: 240895787-240895787 on Build GRCh38

View Full Product Details