Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65640776-65641045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIPA1
Entrez Gene ID
6494
Gene Name
signal-induced proliferation-associated 1
Gene Aliases
SPA1
UniGene
Hs.530477
Gene Chromosome Location
Chr.11: 65638107-65650930 on Build GRCh38
Transcripts
NM_006747.3 NM_153253.29 XM_005274189.2 XM_011545214.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6396579 c.4C>T p.P2S Chr.11: 65640925-65640925 on Build GRCh38
6224565 c.77G>A p.R26H Chr.11: 65640998-65640998 on Build GRCh38
4647413 c.9G>A p.M3I Chr.11: 65640930-65640930 on Build GRCh38
358560 c.54G>T p.A18A Chr.11: 65640975-65640975 on Build GRCh38
4970584 c.37C>T p.R13W Chr.11: 65640958-65640958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961504325 Chr.11: 65640959-65640959 on Build GRCh38
rs777351144 Chr.11: 65640889-65640889 on Build GRCh38
rs1005728212 Chr.11: 65640955-65640955 on Build GRCh38
rs777470367 Chr.11: 65640967-65640967 on Build GRCh38
rs893615554 Chr.11: 65640865-65640865 on Build GRCh38
rs749443708 Chr.11: 65641025-65641025 on Build GRCh38
rs760601344 Chr.11: 65640936-65640936 on Build GRCh38
rs149267827 Chr.11: 65640916-65640916 on Build GRCh38
rs1018096731 Chr.11: 65640922-65640922 on Build GRCh38
rs899668267 Chr.11: 65640871-65640871 on Build GRCh38
rs765175393 Chr.11: 65640949-65640949 on Build GRCh38
rs747620788 Chr.11: 65640881-65640881 on Build GRCh38
rs758072381 Chr.11: 65640888-65640888 on Build GRCh38
rs759426839 Chr.11: 65640940-65640940 on Build GRCh38
rs770661437 Chr.11: 65640880-65640880 on Build GRCh38
rs763066867 Chr.11: 65640913-65640913 on Build GRCh38
rs868419583 Chr.11: 65640997-65640997 on Build GRCh38
rs768841771 Chr.11: 65641026-65641026 on Build GRCh38
rs199844787 Chr.11: 65641007-65641007 on Build GRCh38
rs747380737 Chr.11: 65640891-65640891 on Build GRCh38
rs578008506 Chr.11: 65640938-65640938 on Build GRCh38
rs781752485 Chr.11: 65640896-65640896 on Build GRCh38
rs556138902 Chr.11: 65640858-65640858 on Build GRCh38
rs371737660 Chr.11: 65640958-65640958 on Build GRCh38
rs1030480195 Chr.11: 65640904-65640904 on Build GRCh38
rs569233460 Chr.11: 65640900-65640900 on Build GRCh38
rs746195319 Chr.11: 65641005-65641005 on Build GRCh38
rs368104019 Chr.11: 65640956-65640956 on Build GRCh38
rs867327883 Chr.11: 65640962-65640962 on Build GRCh38
rs3183269 Chr.11: 65641011-65641011 on Build GRCh38
rs115642826 Chr.11: 65640901-65640901 on Build GRCh38
rs780639128 Chr.11: 65641023-65641023 on Build GRCh38
rs536141873 Chr.11: 65640844-65640844 on Build GRCh38
rs956804137 Chr.11: 65641021-65641021 on Build GRCh38
rs910479749 Chr.11: 65640832-65640832 on Build GRCh38
rs757837758 Chr.11: 65640963-65640963 on Build GRCh38
rs553528144 Chr.11: 65640975-65640975 on Build GRCh38
rs774708167 Chr.11: 65640918-65640918 on Build GRCh38
rs11602995 Chr.11: 65640807-65640807 on Build GRCh38
rs373617916 Chr.11: 65640974-65640974 on Build GRCh38
rs374266207 Chr.11: 65640939-65640939 on Build GRCh38
rs943229554 Chr.11: 65640851-65640851 on Build GRCh38
rs781439867 Chr.11: 65640988-65640988 on Build GRCh38
rs771283293 Chr.11: 65640895-65640895 on Build GRCh38
rs866532073 Chr.11: 65640998-65640998 on Build GRCh38
rs1032267810 Chr.11: 65640999-65640999 on Build GRCh38

View Full Product Details