Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81956558-81956827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOTUM
Entrez Gene ID
147111
Gene Name
NOTUM, palmitoleoyl-protein carboxylesterase
Gene Aliases
hNOTUM
UniGene
Hs.106137
Gene Chromosome Location
Chr.17: 81952507-81961181 on Build GRCh38
Transcripts
NM_178493.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1387376 c.715C>T p.R239C Chr.17: 81956725-81956725 on Build GRCh38
5013261 c.908C>A p.P303Q Chr.17: 81956730-81956730 on Build GRCh38
5104967 c.923G>A p.R308H Chr.17: 81956715-81956715 on Build GRCh38
4513222 c.732C>T p.F244F Chr.17: 81956708-81956708 on Build GRCh38
5104968 c.725G>A p.R242H Chr.17: 81956715-81956715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760375361 Chr.17: 81956645-81956645 on Build GRCh38
rs781721967 Chr.17: 81956708-81956708 on Build GRCh38
rs373215256 Chr.17: 81956780-81956780 on Build GRCh38
rs778089716 Chr.17: 81956616-81956616 on Build GRCh38
rs919128944 Chr.17: 81956619-81956619 on Build GRCh38
rs761416779 Chr.17: 81956637-81956637 on Build GRCh38
rs766230841 Chr.17: 81956651-81956651 on Build GRCh38
rs773949708 Chr.17: 81956633-81956633 on Build GRCh38
rs201842705 Chr.17: 81956775-81956775 on Build GRCh38
rs34047000 Chr.17: 81956729-81956730 on Build GRCh38
rs776143547 Chr.17: 81956716-81956716 on Build GRCh38
rs368557190 Chr.17: 81956624-81956624 on Build GRCh38
rs771382459 Chr.17: 81956623-81956623 on Build GRCh38
rs547986063 Chr.17: 81956754-81956754 on Build GRCh38
rs753745399 Chr.17: 81956653-81956653 on Build GRCh38
rs562220587 Chr.17: 81956615-81956615 on Build GRCh38
rs747320241 Chr.17: 81956699-81956699 on Build GRCh38
rs376813903 Chr.17: 81956724-81956724 on Build GRCh38
rs374364146 Chr.17: 81956711-81956711 on Build GRCh38
rs770383878 Chr.17: 81956712-81956712 on Build GRCh38
rs760415290 Chr.17: 81956737-81956737 on Build GRCh38
rs903099208 Chr.17: 81956577-81956577 on Build GRCh38
rs1035731464 Chr.17: 81956583-81956583 on Build GRCh38
rs35872546 Chr.17: 81956785-81956785 on Build GRCh38
rs772930335 Chr.17: 81956640-81956640 on Build GRCh38
rs958415921 Chr.17: 81956588-81956588 on Build GRCh38
rs1032662187 Chr.17: 81956718-81956718 on Build GRCh38
rs746413570 Chr.17: 81956631-81956631 on Build GRCh38
rs372347873 Chr.17: 81956639-81956639 on Build GRCh38
rs757527675 Chr.17: 81956706-81956706 on Build GRCh38
rs1039118536 Chr.17: 81956801-81956801 on Build GRCh38
rs369233567 Chr.17: 81956789-81956789 on Build GRCh38
rs950617786 Chr.17: 81956782-81956782 on Build GRCh38
rs757313108 Chr.17: 81956693-81956693 on Build GRCh38
rs772738091 Chr.17: 81956729-81956729 on Build GRCh38
rs771671091 Chr.17: 81956728-81956728 on Build GRCh38
rs767603340 Chr.17: 81956687-81956687 on Build GRCh38
rs776364617 Chr.17: 81956745-81956745 on Build GRCh38
rs998051453 Chr.17: 81956661-81956661 on Build GRCh38
rs781595587 Chr.17: 81956788-81956788 on Build GRCh38
rs749874686 Chr.17: 81956796-81956796 on Build GRCh38
rs770691002 Chr.17: 81956741-81956741 on Build GRCh38
rs756559107 Chr.17: 81956791-81956791 on Build GRCh38
rs1002796110 Chr.17: 81956578-81956578 on Build GRCh38
rs1049028540 Chr.17: 81956627-81956627 on Build GRCh38
rs774385518 Chr.17: 81956632-81956632 on Build GRCh38
rs910521226 Chr.17: 81956681-81956681 on Build GRCh38
rs777840883 Chr.17: 81956660-81956660 on Build GRCh38
rs909207656 Chr.17: 81956790-81956790 on Build GRCh38
rs149148127 Chr.17: 81956684-81956684 on Build GRCh38
rs192867542 Chr.17: 81956715-81956715 on Build GRCh38
rs151285733 Chr.17: 81956747-81956747 on Build GRCh38
rs748352231 Chr.17: 81956610-81956610 on Build GRCh38
rs572351960 Chr.17: 81956580-81956580 on Build GRCh38
rs545750044 Chr.17: 81956598-81956598 on Build GRCh38
rs780410607 Chr.17: 81956795-81956795 on Build GRCh38
rs898877133 Chr.17: 81956806-81956806 on Build GRCh38
rs188513827 Chr.17: 81956618-81956618 on Build GRCh38
rs752681632 Chr.17: 81956657-81956657 on Build GRCh38
rs762935584 Chr.17: 81956777-81956777 on Build GRCh38
rs369826654 Chr.17: 81956658-81956658 on Build GRCh38
rs199589875 Chr.17: 81956740-81956740 on Build GRCh38
rs764271317 Chr.17: 81956779-81956779 on Build GRCh38
rs758505308 Chr.17: 81956611-81956611 on Build GRCh38

View Full Product Details