Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 70326173-70326444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPFIA1
Entrez Gene ID
8500
Gene Name
PTPRF interacting protein alpha 1
Gene Aliases
LIP.1, LIP1, LIPRIN
UniGene
Hs.530749
Gene Chromosome Location
Chr.11: 70270687-70384501 on Build GRCh38
Transcripts
NM_003626.3 NM_177423.2 NR_045286.1 XM_006718716.3 XM_011545306.2 XM_011545307.2 XM_011545308.2 XM_011545309.2 XM_011545310.2 XM_011545311.2 XM_011545312.2 XM_011545313.2 XM_011545314.2 XM_011545315.2 XM_011545316.2 XM_011545317.2 XM_011545319.1 XM_017018448.1 XM_017018449.1 XM_017018450.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
284166 c.612G>T p.M204I Chr.11: 70326267-70326267 on Build GRCh38
5970352 c.699G>A p.T233T Chr.11: 70326354-70326354 on Build GRCh38
5724931 c.664G>A p.D222N Chr.11: 70326319-70326319 on Build GRCh38
7195818 c.683A>C p.E228A Chr.11: 70326338-70326338 on Build GRCh38
2043801 c.622G>T p.E208* Chr.11: 70326277-70326277 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751381400 Chr.11: 70326332-70326332 on Build GRCh38
rs369965878 Chr.11: 70326308-70326308 on Build GRCh38
rs777634542 Chr.11: 70326224-70326224 on Build GRCh38
rs771426596 Chr.11: 70326240-70326241 on Build GRCh38
rs770129091 Chr.11: 70326394-70326394 on Build GRCh38
rs371058178 Chr.11: 70326397-70326397 on Build GRCh38
rs141458298 Chr.11: 70326351-70326351 on Build GRCh38
rs757214242 Chr.11: 70326227-70326227 on Build GRCh38
rs773764044 Chr.11: 70326299-70326299 on Build GRCh38
rs759183650 Chr.11: 70326315-70326315 on Build GRCh38
rs373950779 Chr.11: 70326321-70326321 on Build GRCh38
rs762668966 Chr.11: 70326329-70326329 on Build GRCh38
rs376891820 Chr.11: 70326354-70326354 on Build GRCh38
rs748111587 Chr.11: 70326346-70326346 on Build GRCh38
rs750360108 Chr.11: 70326343-70326343 on Build GRCh38
rs771416110 Chr.11: 70326367-70326367 on Build GRCh38
rs758181754 Chr.11: 70326220-70326220 on Build GRCh38
rs781061387 Chr.11: 70326236-70326236 on Build GRCh38
rs571405866 Chr.11: 70326314-70326314 on Build GRCh38
rs200085975 Chr.11: 70326226-70326226 on Build GRCh38
rs151186142 Chr.11: 70326344-70326344 on Build GRCh38
rs372386598 Chr.11: 70326241-70326241 on Build GRCh38
rs201134555 Chr.11: 70326311-70326311 on Build GRCh38
rs757026360 Chr.11: 70326333-70326333 on Build GRCh38
rs970205859 Chr.11: 70326211-70326211 on Build GRCh38
rs764786699 Chr.11: 70326317-70326317 on Build GRCh38
rs752389792 Chr.11: 70326212-70326212 on Build GRCh38
rs140522822 Chr.11: 70326390-70326390 on Build GRCh38
rs768695396 Chr.11: 70326239-70326239 on Build GRCh38
rs763612171 Chr.11: 70326331-70326331 on Build GRCh38
rs201181680 Chr.11: 70326238-70326238 on Build GRCh38
rs778116748 Chr.11: 70326353-70326353 on Build GRCh38
rs532296062 Chr.11: 70326336-70326336 on Build GRCh38
rs1017730061 Chr.11: 70326357-70326357 on Build GRCh38
rs201970155 Chr.11: 70326296-70326296 on Build GRCh38
rs142170424 Chr.11: 70326292-70326292 on Build GRCh38
rs748390540 Chr.11: 70326277-70326277 on Build GRCh38
rs770254855 Chr.11: 70326290-70326290 on Build GRCh38

View Full Product Details