Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 14295685-14295932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRAS2
Entrez Gene ID
22800
Gene Name
related RAS viral (r-ras) oncogene homolog 2
Gene Aliases
TC21
UniGene
Hs.502004
Gene Chromosome Location
Chr.11: 14277920-14364506 on Build GRCh38
Transcripts
NM_001102669.2 NM_001177314.1 NM_001177315.1 NM_012250.5 XM_017017363.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6991821 c.113A>G p.Y38C Chr.11: 14295851-14295851 on Build GRCh38
3383359 c.109-3C>T p.? Chr.11: 14295858-14295858 on Build GRCh38
4722823 c.158A>T p.K53M Chr.11: 14295806-14295806 on Build GRCh38
6281883 c.145G>A p.D49N Chr.11: 14295819-14295819 on Build GRCh38
4722822 c.188G>A p.R63Q Chr.11: 14295776-14295776 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782067450 Chr.11: 14295727-14295727 on Build GRCh38
rs183165083 Chr.11: 14295902-14295902 on Build GRCh38
rs782132298 Chr.11: 14295722-14295722 on Build GRCh38
rs782748277 Chr.11: 14295723-14295723 on Build GRCh38
rs782300279 Chr.11: 14295867-14295868 on Build GRCh38
rs781909362 Chr.11: 14295734-14295734 on Build GRCh38
rs782481088 Chr.11: 14295773-14295773 on Build GRCh38
rs913808099 Chr.11: 14295830-14295830 on Build GRCh38
rs782535692 Chr.11: 14295744-14295744 on Build GRCh38
rs782670869 Chr.11: 14295842-14295842 on Build GRCh38
rs527938617 Chr.11: 14295775-14295775 on Build GRCh38
rs782813661 Chr.11: 14295891-14295891 on Build GRCh38
rs782189872 Chr.11: 14295777-14295777 on Build GRCh38
rs782245195 Chr.11: 14295826-14295826 on Build GRCh38
rs781926268 Chr.11: 14295754-14295754 on Build GRCh38
rs782166856 Chr.11: 14295879-14295879 on Build GRCh38
rs560418967 Chr.11: 14295716-14295716 on Build GRCh38
rs564675458 Chr.11: 14295862-14295862 on Build GRCh38
rs782656684 Chr.11: 14295748-14295748 on Build GRCh38
rs782273898 Chr.11: 14295846-14295846 on Build GRCh38
rs552513136 Chr.11: 14295785-14295785 on Build GRCh38
rs368177668 Chr.11: 14295759-14295759 on Build GRCh38
rs782119209 Chr.11: 14295904-14295904 on Build GRCh38
rs782328747 Chr.11: 14295722-14295723 on Build GRCh38
rs542721390 Chr.11: 14295724-14295724 on Build GRCh38
rs782691594 Chr.11: 14295732-14295732 on Build GRCh38
rs781975658 Chr.11: 14295850-14295850 on Build GRCh38
rs550633773 Chr.11: 14295906-14295906 on Build GRCh38
rs782329465 Chr.11: 14295868-14295868 on Build GRCh38
rs782381350 Chr.11: 14295847-14295847 on Build GRCh38

View Full Product Details