Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46729483-46729756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYLK3
Entrez Gene ID
91807
Gene Name
myosin light chain kinase 3
Gene Aliases
MLCK, MLCK2, caMLCK
UniGene
Hs.130465
Gene Chromosome Location
Chr.16: 46702282-46763246 on Build GRCh38
Transcripts
NM_001308301.1 NM_182493.2 XM_006721334.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
245877 c.1650C>T p.S550S Chr.16: 46729606-46729606 on Build GRCh38
5725662 c.1581C>T p.G527G Chr.16: 46729675-46729675 on Build GRCh38
6719727 c.1622T>A p.L541Q Chr.16: 46729634-46729634 on Build GRCh38
5513195 c.1649G>A p.S550N Chr.16: 46729607-46729607 on Build GRCh38
3509680 c.1656G>A p.K552K Chr.16: 46729600-46729600 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752274045 Chr.16: 46729652-46729652 on Build GRCh38
rs369846131 Chr.16: 46729702-46729702 on Build GRCh38
rs200617596 Chr.16: 46729698-46729698 on Build GRCh38
rs370418930 Chr.16: 46729558-46729558 on Build GRCh38
rs775706296 Chr.16: 46729682-46729682 on Build GRCh38
rs770787846 Chr.16: 46729669-46729669 on Build GRCh38
rs752422884 Chr.16: 46729554-46729554 on Build GRCh38
rs781224041 Chr.16: 46729672-46729672 on Build GRCh38
rs752129895 Chr.16: 46729717-46729717 on Build GRCh38
rs142285931 Chr.16: 46729699-46729699 on Build GRCh38
rs374127974 Chr.16: 46729646-46729646 on Build GRCh38
rs764444623 Chr.16: 46729618-46729618 on Build GRCh38
rs531593186 Chr.16: 46729553-46729553 on Build GRCh38
rs769926748 Chr.16: 46729681-46729681 on Build GRCh38
rs376737122 Chr.16: 46729708-46729708 on Build GRCh38
rs564472600 Chr.16: 46729508-46729508 on Build GRCh38
rs547611755 Chr.16: 46729697-46729697 on Build GRCh38
rs762278874 Chr.16: 46729627-46729627 on Build GRCh38
rs370177490 Chr.16: 46729666-46729666 on Build GRCh38
rs908316569 Chr.16: 46729523-46729523 on Build GRCh38
rs767465661 Chr.16: 46729530-46729530 on Build GRCh38
rs140150530 Chr.16: 46729638-46729638 on Build GRCh38
rs770014661 Chr.16: 46729589-46729589 on Build GRCh38
rs761168895 Chr.16: 46729707-46729707 on Build GRCh38
rs755898293 Chr.16: 46729690-46729690 on Build GRCh38
rs754429348 Chr.16: 46729647-46729647 on Build GRCh38
rs774666158 Chr.16: 46729686-46729686 on Build GRCh38
rs896950928 Chr.16: 46729536-46729536 on Build GRCh38
rs143794902 Chr.16: 46729683-46729683 on Build GRCh38
rs777526469 Chr.16: 46729659-46729659 on Build GRCh38
rs943755836 Chr.16: 46729529-46729529 on Build GRCh38
rs759646582 Chr.16: 46729569-46729569 on Build GRCh38
rs199585118 Chr.16: 46729556-46729556 on Build GRCh38
rs775796132 Chr.16: 46729596-46729596 on Build GRCh38
rs1042599022 Chr.16: 46729692-46729692 on Build GRCh38
rs765781403 Chr.16: 46729708-46729724 on Build GRCh38
rs375577871 Chr.16: 46729660-46729660 on Build GRCh38
rs778403033 Chr.16: 46729648-46729648 on Build GRCh38
rs757977738 Chr.16: 46729657-46729657 on Build GRCh38
rs185999875 Chr.16: 46729623-46729623 on Build GRCh38
rs201506640 Chr.16: 46729597-46729597 on Build GRCh38
rs889683915 Chr.16: 46729609-46729609 on Build GRCh38
rs757961855 Chr.16: 46729725-46729725 on Build GRCh38
rs9923813 Chr.16: 46729650-46729650 on Build GRCh38
rs747279793 Chr.16: 46729512-46729512 on Build GRCh38

View Full Product Details