Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40815662-40815935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R14D
Entrez Gene ID
54866
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14D
Gene Aliases
CPI17-like, GBPI-1, GBPI1
UniGene
Hs.192927
Gene Chromosome Location
Chr.15: 40815445-40828793 on Build GRCh38
Transcripts
NM_001130143.1 NM_017726.7 XM_017022372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6306660 c.388C>A p.L130M Chr.15: 40815746-40815746 on Build GRCh38
6416640 c.489G>C p.R163S Chr.15: 40815761-40815761 on Build GRCh38
1221738 c.578C>T p.A193V Chr.15: 40815672-40815672 on Build GRCh38
6416641 c.373G>C p.A125P Chr.15: 40815761-40815761 on Build GRCh38
7148025 c.509C>T p.S170L Chr.15: 40815741-40815741 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186411966 Chr.15: 40815878-40815878 on Build GRCh38
rs754860833 Chr.15: 40815792-40815793 on Build GRCh38
rs77318894 Chr.15: 40815812-40815812 on Build GRCh38
rs933376478 Chr.15: 40815789-40815789 on Build GRCh38
rs147588923 Chr.15: 40815709-40815709 on Build GRCh38
rs760046191 Chr.15: 40815711-40815711 on Build GRCh38
rs898447705 Chr.15: 40815852-40815852 on Build GRCh38
rs918516874 Chr.15: 40815728-40815728 on Build GRCh38
rs906881759 Chr.15: 40815887-40815887 on Build GRCh38
rs369502844 Chr.15: 40815796-40815796 on Build GRCh38
rs1002602267 Chr.15: 40815905-40815905 on Build GRCh38
rs868858907 Chr.15: 40815872-40815872 on Build GRCh38
rs770191865 Chr.15: 40815779-40815779 on Build GRCh38
rs140774733 Chr.15: 40815741-40815741 on Build GRCh38
rs929914171 Chr.15: 40815858-40815858 on Build GRCh38
rs199678247 Chr.15: 40815730-40815730 on Build GRCh38
rs781756533 Chr.15: 40815752-40815752 on Build GRCh38
rs923302126 Chr.15: 40815753-40815753 on Build GRCh38
rs771725184 Chr.15: 40815790-40815790 on Build GRCh38
rs200229830 Chr.15: 40815707-40815707 on Build GRCh38
rs750712180 Chr.15: 40815723-40815723 on Build GRCh38
rs747121904 Chr.15: 40815744-40815744 on Build GRCh38
rs55987500 Chr.15: 40815716-40815716 on Build GRCh38
rs200833224 Chr.15: 40815802-40815802 on Build GRCh38
rs762283847 Chr.15: 40815874-40815874 on Build GRCh38
rs748588107 Chr.15: 40815755-40815755 on Build GRCh38
rs778143776 Chr.15: 40815781-40815781 on Build GRCh38
rs776682553 Chr.15: 40815811-40815811 on Build GRCh38
rs769223399 Chr.15: 40815682-40815682 on Build GRCh38
rs1038281496 Chr.15: 40815816-40815816 on Build GRCh38
rs200817876 Chr.15: 40815706-40815706 on Build GRCh38
rs971986511 Chr.15: 40815875-40815875 on Build GRCh38
rs377153944 Chr.15: 40815721-40815721 on Build GRCh38
rs201309340 Chr.15: 40815800-40815800 on Build GRCh38
rs942434524 Chr.15: 40815786-40815786 on Build GRCh38
rs753719241 Chr.15: 40815718-40815718 on Build GRCh38
rs111997023 Chr.15: 40815715-40815715 on Build GRCh38
rs776945699 Chr.15: 40815686-40815686 on Build GRCh38
rs775089368 Chr.15: 40815710-40815710 on Build GRCh38
rs768251308 Chr.15: 40815901-40815901 on Build GRCh38
rs372494011 Chr.15: 40815915-40815915 on Build GRCh38
rs765104613 Chr.15: 40815722-40815722 on Build GRCh38
rs750262382 Chr.15: 40815913-40815913 on Build GRCh38
rs145045388 Chr.15: 40815695-40815695 on Build GRCh38
rs754946380 Chr.15: 40815751-40815751 on Build GRCh38
rs764102243 Chr.15: 40815712-40815712 on Build GRCh38
rs903477580 Chr.15: 40815797-40815797 on Build GRCh38

View Full Product Details