Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63741391-63741636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC2A4RG
Entrez Gene ID
56731
Gene Name
SLC2A4 regulator
Gene Aliases
GEF, HDBP-1, HDBP1, Si-1-2, Si-1-2-19
UniGene
Hs.435126
Gene Chromosome Location
Chr.20: 63739858-63744050 on Build GRCh38
Transcripts
NM_020062.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4134813 c.372G>A p.P124P Chr.20: 63741460-63741460 on Build GRCh38
6160712 c.331C>G p.L111V Chr.20: 63741419-63741419 on Build GRCh38
6781369 c.380C>T p.A127V Chr.20: 63741468-63741468 on Build GRCh38
1226242 c.358C>A p.L120I Chr.20: 63741446-63741446 on Build GRCh38
6363766 c.362T>C p.L121P Chr.20: 63741450-63741450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2427536 Chr.20: 63741460-63741460 on Build GRCh38
rs756752505 Chr.20: 63741440-63741440 on Build GRCh38
rs774338818 Chr.20: 63741459-63741459 on Build GRCh38
rs181885589 Chr.20: 63741484-63741484 on Build GRCh38
rs776100778 Chr.20: 63741510-63741510 on Build GRCh38
rs780902928 Chr.20: 63741503-63741503 on Build GRCh38
rs779906227 Chr.20: 63741485-63741485 on Build GRCh38
rs754724224 Chr.20: 63741496-63741496 on Build GRCh38
rs776900762 Chr.20: 63741516-63741516 on Build GRCh38
rs779818384 Chr.20: 63741449-63741449 on Build GRCh38
rs745887196 Chr.20: 63741508-63741508 on Build GRCh38
rs150827036 Chr.20: 63741471-63741471 on Build GRCh38
rs981267525 Chr.20: 63741479-63741479 on Build GRCh38
rs767965534 Chr.20: 63741413-63741413 on Build GRCh38
rs761520872 Chr.20: 63741521-63741521 on Build GRCh38
rs199996953 Chr.20: 63741454-63741454 on Build GRCh38
rs139717672 Chr.20: 63741472-63741472 on Build GRCh38
rs375432430 Chr.20: 63741456-63741456 on Build GRCh38
rs773890422 Chr.20: 63741572-63741572 on Build GRCh38
rs753182069 Chr.20: 63741418-63741418 on Build GRCh38
rs773413233 Chr.20: 63741455-63741455 on Build GRCh38
rs764771010 Chr.20: 63741478-63741478 on Build GRCh38
rs761421194 Chr.20: 63741474-63741474 on Build GRCh38
rs772284704 Chr.20: 63741509-63741509 on Build GRCh38
rs760015271 Chr.20: 63741412-63741412 on Build GRCh38
rs565810341 Chr.20: 63741507-63741507 on Build GRCh38
rs116980580 Chr.20: 63741445-63741445 on Build GRCh38
rs1040973973 Chr.20: 63741608-63741608 on Build GRCh38
rs751498268 Chr.20: 63741490-63741490 on Build GRCh38
rs926135392 Chr.20: 63741578-63741578 on Build GRCh38
rs2253823 Chr.20: 63741603-63741603 on Build GRCh38
rs200105433 Chr.20: 63741525-63741525 on Build GRCh38
rs778307771 Chr.20: 63741441-63741441 on Build GRCh38
rs978914796 Chr.20: 63741575-63741575 on Build GRCh38
rs913134248 Chr.20: 63741518-63741518 on Build GRCh38
rs758370846 Chr.20: 63741446-63741446 on Build GRCh38
rs763783604 Chr.20: 63741434-63741434 on Build GRCh38
rs536517009 Chr.20: 63741536-63741536 on Build GRCh38
rs774876751 Chr.20: 63741411-63741411 on Build GRCh38
rs943722292 Chr.20: 63741605-63741605 on Build GRCh38
rs776838158 Chr.20: 63741618-63741618 on Build GRCh38
rs761054013 Chr.20: 63741469-63741469 on Build GRCh38
rs747489045 Chr.20: 63741589-63741589 on Build GRCh38
rs761287069 Chr.20: 63741511-63741511 on Build GRCh38
rs1042864660 Chr.20: 63741557-63741557 on Build GRCh38
rs201632478 Chr.20: 63741461-63741461 on Build GRCh38
rs201917388 Chr.20: 63741439-63741439 on Build GRCh38
rs749499955 Chr.20: 63741506-63741506 on Build GRCh38
rs746862083 Chr.20: 63741452-63741452 on Build GRCh38
rs904370811 Chr.20: 63741598-63741598 on Build GRCh38
rs755815764 Chr.20: 63741429-63741429 on Build GRCh38
rs982983446 Chr.20: 63741584-63741584 on Build GRCh38
rs967611533 Chr.20: 63741547-63741547 on Build GRCh38
rs764855468 Chr.20: 63741523-63741523 on Build GRCh38

View Full Product Details