Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 97687292-97687527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTDH
Entrez Gene ID
92140
Gene Name
metadherin
Gene Aliases
3D3, AEG-1, AEG1, LYRIC, LYRIC/3D3
UniGene
Hs.377155
Gene Chromosome Location
Chr.8: 97643972-97730260 on Build GRCh38
Transcripts
NM_178812.3 XM_005251099.3 XM_011517367.2 XM_011517368.2 XM_011517369.2 XM_011517370.2 XM_017013966.1 XM_017013967.1 XM_017013968.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4846660 c.641C>G p.S214C Chr.8: 97687501-97687501 on Build GRCh38
4703515 c.593A>G p.H198R Chr.8: 97687453-97687453 on Build GRCh38
3902301 c.605G>A p.R202Q Chr.8: 97687465-97687465 on Build GRCh38
3902302 c.619C>T p.R207C Chr.8: 97687479-97687479 on Build GRCh38
6516397 c.600G>A p.E200E Chr.8: 97687460-97687460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021377399 Chr.8: 97687346-97687346 on Build GRCh38
rs760090387 Chr.8: 97687423-97687423 on Build GRCh38
rs990775009 Chr.8: 97687328-97687328 on Build GRCh38
rs774959822 Chr.8: 97687422-97687422 on Build GRCh38
rs949226409 Chr.8: 97687391-97687391 on Build GRCh38
rs969376651 Chr.8: 97687492-97687492 on Build GRCh38
rs769453134 Chr.8: 97687419-97687419 on Build GRCh38
rs747607129 Chr.8: 97687413-97687413 on Build GRCh38
rs1022807416 Chr.8: 97687488-97687488 on Build GRCh38
rs888798739 Chr.8: 97687334-97687334 on Build GRCh38
rs139649151 Chr.8: 97687497-97687497 on Build GRCh38
rs544898775 Chr.8: 97687368-97687368 on Build GRCh38
rs902899556 Chr.8: 97687355-97687355 on Build GRCh38
rs761167308 Chr.8: 97687458-97687458 on Build GRCh38
rs959321671 Chr.8: 97687323-97687323 on Build GRCh38
rs180867681 Chr.8: 97687473-97687473 on Build GRCh38
rs368404042 Chr.8: 97687394-97687394 on Build GRCh38
rs759519703 Chr.8: 97687405-97687406 on Build GRCh38
rs11998518 Chr.8: 97687456-97687456 on Build GRCh38
rs768193329 Chr.8: 97687399-97687399 on Build GRCh38
rs776922654 Chr.8: 97687474-97687474 on Build GRCh38
rs761829194 Chr.8: 97687481-97687481 on Build GRCh38
rs575742664 Chr.8: 97687343-97687343 on Build GRCh38
rs780905504 Chr.8: 97687405-97687405 on Build GRCh38
rs564791221 Chr.8: 97687426-97687426 on Build GRCh38
rs775703791 Chr.8: 97687443-97687443 on Build GRCh38
rs923705458 Chr.8: 97687482-97687482 on Build GRCh38
rs549652426 Chr.8: 97687322-97687322 on Build GRCh38

View Full Product Details