Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140966993-140967236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHA1
Entrez Gene ID
56147
Gene Name
protocadherin alpha 1
Gene Aliases
PCDH-ALPHA1
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140786136-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5 NM_031883.2

Gene Symbol
PCDHA10
Entrez Gene ID
56139
Gene Name
protocadherin alpha 10
Gene Aliases
CNR8, CNRN8, CNRS8, CRNR8, PCDH-ALPHA10
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140855883-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5 NM_031883.2

Gene Symbol
PCDHA11
Entrez Gene ID
56138
Gene Name
protocadherin alpha 11
Gene Aliases
CNR7, CNRN7, CNRS7, CRNR7, PCDH-ALPHA11
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140867513-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5 NM_031883.2

Gene Symbol
PCDHA12
Entrez Gene ID
56137
Gene Name
protocadherin alpha 12
Gene Aliases
PCDH-ALPHA12
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140875302-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5 NM_031883.2

Gene Symbol
PCDHA13
Entrez Gene ID
56136
Gene Name
protocadherin alpha 13
Gene Aliases
CNR5, CNRN5, CNRS5, CRNR5, PCDH-ALPHA13
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140882269-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5 NM_031883.2

Gene Symbol
PCDHA2
Entrez Gene ID
56146
Gene Name
protocadherin alpha 2
Gene Aliases
PCDH-ALPHA2
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140794859-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5 NM_031883.2

Gene Symbol
PCDHA3
Entrez Gene ID
56145
Gene Name
protocadherin alpha 3
Gene Aliases
PCDH-ALPHA3
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140801198-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5 NM_031883.2

Gene Symbol
PCDHA4
Entrez Gene ID
56144
Gene Name
protocadherin alpha 4
Gene Aliases
CNR1, CNRN1, CRNR1, PCDH-ALPHA4
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140807074-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5 NM_031883.2

Gene Symbol
PCDHA5
Entrez Gene ID
56143
Gene Name
protocadherin alpha 5
Gene Aliases
CNR6, CNRN6, CNRS6, CRNR6, PCDH-ALPHA5
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140821776-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5 NM_031883.2

Gene Symbol
PCDHA6
Entrez Gene ID
56142
Gene Name
protocadherin alpha 6
Gene Aliases
CNR2, CNRN2, CNRS2, CRNR2, PCDH-ALPHA6
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140827922-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5 NM_031883.2

Gene Symbol
PCDHA7
Entrez Gene ID
56141
Gene Name
protocadherin alpha 7
Gene Aliases
CNR4, CNRN4, CNRS4, CRNR4, PCDH-ALPHA7
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140834384-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5 NM_031883.2

Gene Symbol
PCDHA8
Entrez Gene ID
56140
Gene Name
protocadherin alpha 8
Gene Aliases
PCDH-ALPHA8
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140841322-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5 NM_031883.2

Gene Symbol
PCDHA9
Entrez Gene ID
9752
Gene Name
protocadherin alpha 9
Gene Aliases
PCDH-ALPHA9
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140847772-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5 NM_031883.2

Gene Symbol
PCDHAC1
Entrez Gene ID
56135
Gene Name
protocadherin alpha subfamily C, 1
Gene Aliases
PCDH-ALPHA-C1
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140926717-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5 NM_031883.2

Gene Symbol
PCDHAC2
Entrez Gene ID
56134
Gene Name
protocadherin alpha subfamily C, 2
Gene Aliases
PCDH-ALPHA-C2
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140966162-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031860.2 NM_018902.4 NM_018903.3 NM_018904.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1 NM_018898.3 NM_018899.5 NM_031883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6043892 c.395G>T p.S132I Chr.5: 140967161-140967161 on Build GRCh38
6740206 c.384C>A p.P128P Chr.5: 140967150-140967150 on Build GRCh38
1261328 c.277G>A p.D93N Chr.5: 140967043-140967043 on Build GRCh38
6740207 c.368T>C p.V123A Chr.5: 140967134-140967134 on Build GRCh38
5619089 c.361T>C p.L121L Chr.5: 140967127-140967127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782433025 Chr.5: 140967181-140967181 on Build GRCh38
rs782261207 Chr.5: 140967091-140967091 on Build GRCh38
rs782199698 Chr.5: 140967107-140967107 on Build GRCh38
rs782556858 Chr.5: 140967087-140967087 on Build GRCh38
rs376266648 Chr.5: 140967207-140967207 on Build GRCh38
rs563863114 Chr.5: 140967166-140967166 on Build GRCh38
rs782147091 Chr.5: 140967020-140967020 on Build GRCh38
rs374744732 Chr.5: 140967104-140967104 on Build GRCh38
rs782407413 Chr.5: 140967114-140967114 on Build GRCh38
rs781848948 Chr.5: 140967042-140967042 on Build GRCh38
rs370329233 Chr.5: 140967117-140967117 on Build GRCh38
rs782524334 Chr.5: 140967162-140967162 on Build GRCh38
rs782593106 Chr.5: 140967084-140967084 on Build GRCh38
rs782715200 Chr.5: 140967066-140967066 on Build GRCh38
rs782815903 Chr.5: 140967147-140967147 on Build GRCh38
rs146322183 Chr.5: 140967210-140967210 on Build GRCh38
rs1042081336 Chr.5: 140967144-140967144 on Build GRCh38
rs781942171 Chr.5: 140967080-140967080 on Build GRCh38
rs181864889 Chr.5: 140967171-140967171 on Build GRCh38
rs782387198 Chr.5: 140967124-140967124 on Build GRCh38
rs762120187 Chr.5: 140967164-140967164 on Build GRCh38
rs781981464 Chr.5: 140967191-140967191 on Build GRCh38
rs782344374 Chr.5: 140967092-140967092 on Build GRCh38
rs782810130 Chr.5: 140967034-140967034 on Build GRCh38
rs374670692 Chr.5: 140967119-140967119 on Build GRCh38
rs781831975 Chr.5: 140967070-140967070 on Build GRCh38
rs139717123 Chr.5: 140967213-140967213 on Build GRCh38
rs562713934 Chr.5: 140967172-140967172 on Build GRCh38
rs547096956 Chr.5: 140967167-140967167 on Build GRCh38
rs782089524 Chr.5: 140967135-140967135 on Build GRCh38
rs887648506 Chr.5: 140967019-140967019 on Build GRCh38
rs549238768 Chr.5: 140967211-140967211 on Build GRCh38
rs155808 Chr.5: 140967127-140967127 on Build GRCh38
rs781864120 Chr.5: 140967149-140967149 on Build GRCh38
rs782646028 Chr.5: 140967215-140967215 on Build GRCh38
rs781814682 Chr.5: 140967140-140967140 on Build GRCh38
rs782698899 Chr.5: 140967136-140967136 on Build GRCh38
rs155809 Chr.5: 140967085-140967085 on Build GRCh38
rs782123539 Chr.5: 140967049-140967049 on Build GRCh38
rs782158011 Chr.5: 140967146-140967146 on Build GRCh38
rs782811758 Chr.5: 140967101-140967102 on Build GRCh38

View Full Product Details