Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52286935-52287194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT81
Entrez Gene ID
3887
Gene Name
keratin 81
Gene Aliases
HB1, Hb-1, KRTHB1, MLN137, ghHkb1, hHAKB2-1
UniGene
Hs.658118
Gene Chromosome Location
Chr.12: 52285913-52291515 on Build GRCh38
Transcripts
NM_002281.3 NM_001320198.1 XM_005268866.4 XM_017019296.1

Gene Symbol
KRT86
Entrez Gene ID
3892
Gene Name
keratin 86
Gene Aliases
HB6, Hb1, K86, KRTHB1, KRTHB6, MNX
UniGene
Hs.278658
Gene Chromosome Location
Chr.12: 52270507-52309163 on Build GRCh38
Transcripts
NM_002281.3 NM_001320198.1 XM_005268866.4 XM_017019296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
162065 c.1237G>A p.E413K Chr.12: 52287112-52287112 on Build GRCh38
2010275 c.1203C>T p.I401I Chr.12: 52287146-52287146 on Build GRCh38
4461183 c.1195C>T p.L399L Chr.12: 52287154-52287154 on Build GRCh38
400158 c.1161G>T p.R387S Chr.12: 52287188-52287188 on Build GRCh38
694236 c.1176G>C p.V392V Chr.12: 52287173-52287173 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751051005 Chr.12: 52287157-52287157 on Build GRCh38
rs892582153 Chr.12: 52286999-52286999 on Build GRCh38
rs113097323 Chr.12: 52286998-52286999 on Build GRCh38
rs111382015 Chr.12: 52287122-52287122 on Build GRCh38
rs939356400 Chr.12: 52286989-52286991 on Build GRCh38
rs750223770 Chr.12: 52287073-52287073 on Build GRCh38
rs934290936 Chr.12: 52287091-52287091 on Build GRCh38
rs59448276 Chr.12: 52286982-52286982 on Build GRCh38
rs761893737 Chr.12: 52287161-52287161 on Build GRCh38
rs757215183 Chr.12: 52287066-52287066 on Build GRCh38
rs374751440 Chr.12: 52287088-52287088 on Build GRCh38
rs751509447 Chr.12: 52287055-52287055 on Build GRCh38
rs57419521 Chr.12: 52287112-52287112 on Build GRCh38
rs1016878787 Chr.12: 52286971-52286971 on Build GRCh38
rs771393943 Chr.12: 52287127-52287127 on Build GRCh38
rs141521550 Chr.12: 52287140-52287140 on Build GRCh38
rs961632486 Chr.12: 52286973-52286973 on Build GRCh38
rs551405099 Chr.12: 52286981-52286981 on Build GRCh38
rs759844211 Chr.12: 52287139-52287139 on Build GRCh38
rs777027305 Chr.12: 52287129-52287129 on Build GRCh38
rs74095619 Chr.12: 52287005-52287005 on Build GRCh38
rs534459175 Chr.12: 52287097-52287097 on Build GRCh38
rs200188584 Chr.12: 52287069-52287069 on Build GRCh38
rs771983295 Chr.12: 52287113-52287113 on Build GRCh38
rs991354798 Chr.12: 52286996-52286996 on Build GRCh38
rs1044307771 Chr.12: 52287008-52287008 on Build GRCh38
rs1005543455 Chr.12: 52286969-52286969 on Build GRCh38
rs750276983 Chr.12: 52287171-52287171 on Build GRCh38
rs368237680 Chr.12: 52287101-52287101 on Build GRCh38
rs763579563 Chr.12: 52287155-52287155 on Build GRCh38
rs767515484 Chr.12: 52287168-52287168 on Build GRCh38
rs762654620 Chr.12: 52287149-52287149 on Build GRCh38
rs755276088 Chr.12: 52287085-52287085 on Build GRCh38
rs201447608 Chr.12: 52287126-52287126 on Build GRCh38
rs570237300 Chr.12: 52287010-52287010 on Build GRCh38
rs978452210 Chr.12: 52287032-52287032 on Build GRCh38
rs56821304 Chr.12: 52287145-52287145 on Build GRCh38
rs540172456 Chr.12: 52286994-52286994 on Build GRCh38

View Full Product Details