Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 227524701-227524972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGFG1
Entrez Gene ID
3267
Gene Name
ArfGAP with FG repeats 1
Gene Aliases
HRB, RAB, RIP
UniGene
Hs.352962
Gene Chromosome Location
Chr.2: 227472169-227561222 on Build GRCh38
Transcripts
NM_001135187.1 NM_001135188.1 NM_001135189.1 NM_004504.4 XM_005246516.3 XM_006712478.3 XM_006712479.3 XM_006712480.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3578525 c.616G>A p.D206N Chr.2: 227524837-227524837 on Build GRCh38
1172854 c.614C>T p.S205L Chr.2: 227524835-227524835 on Build GRCh38
3391549 c.541-8G>T p.? Chr.2: 227524754-227524754 on Build GRCh38
1129293 c.569A>C p.Q190P Chr.2: 227524790-227524790 on Build GRCh38
5620221 c.646A>G p.T216A Chr.2: 227524867-227524867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747198025 Chr.2: 227524780-227524780 on Build GRCh38
rs553384337 Chr.2: 227524935-227524935 on Build GRCh38
rs747446259 Chr.2: 227524733-227524738 on Build GRCh38
rs781458811 Chr.2: 227524912-227524912 on Build GRCh38
rs755915101 Chr.2: 227524852-227524852 on Build GRCh38
rs756036502 Chr.2: 227524758-227524758 on Build GRCh38
rs770142841 Chr.2: 227524918-227524918 on Build GRCh38
rs750224811 Chr.2: 227524848-227524848 on Build GRCh38
rs780196903 Chr.2: 227524854-227524854 on Build GRCh38
rs771085675 Chr.2: 227524910-227524910 on Build GRCh38
rs751014599 Chr.2: 227524842-227524842 on Build GRCh38
rs769179839 Chr.2: 227524796-227524798 on Build GRCh38
rs761355360 Chr.2: 227524937-227524937 on Build GRCh38
rs775767947 Chr.2: 227524919-227524919 on Build GRCh38
rs780107649 Chr.2: 227524766-227524766 on Build GRCh38
rs181559703 Chr.2: 227524777-227524777 on Build GRCh38
rs745627392 Chr.2: 227524754-227524754 on Build GRCh38
rs35543167 Chr.2: 227524903-227524904 on Build GRCh38
rs775966354 Chr.2: 227524830-227524830 on Build GRCh38
rs776878345 Chr.2: 227524804-227524804 on Build GRCh38
rs995298199 Chr.2: 227524888-227524888 on Build GRCh38
rs756869199 Chr.2: 227524736-227524736 on Build GRCh38
rs767124071 Chr.2: 227524940-227524940 on Build GRCh38
rs763601084 Chr.2: 227524836-227524836 on Build GRCh38
rs1027816631 Chr.2: 227524906-227524906 on Build GRCh38
rs142029380 Chr.2: 227524778-227524778 on Build GRCh38
rs750076788 Chr.2: 227524944-227524944 on Build GRCh38
rs747058494 Chr.2: 227524904-227524904 on Build GRCh38
rs754894480 Chr.2: 227524890-227524890 on Build GRCh38
rs996806941 Chr.2: 227524772-227524772 on Build GRCh38
rs774831247 Chr.2: 227524924-227524924 on Build GRCh38
rs146333019 Chr.2: 227524855-227524855 on Build GRCh38
rs759918358 Chr.2: 227524809-227524809 on Build GRCh38
rs776068296 Chr.2: 227524799-227524801 on Build GRCh38
rs765786150 Chr.2: 227524822-227524822 on Build GRCh38
rs763466601 Chr.2: 227524933-227524933 on Build GRCh38
rs375289804 Chr.2: 227524897-227524897 on Build GRCh38
rs748286113 Chr.2: 227524775-227524775 on Build GRCh38
rs958372725 Chr.2: 227524894-227524894 on Build GRCh38
rs370510772 Chr.2: 227524752-227524752 on Build GRCh38
rs568024413 Chr.2: 227524873-227524873 on Build GRCh38
rs749588476 Chr.2: 227524800-227524800 on Build GRCh38
rs773590954 Chr.2: 227524739-227524739 on Build GRCh38
rs367965065 Chr.2: 227524735-227524735 on Build GRCh38
rs963156327 Chr.2: 227524795-227524795 on Build GRCh38
rs899629523 Chr.2: 227524743-227524743 on Build GRCh38
rs771481805 Chr.2: 227524833-227524833 on Build GRCh38
rs761356716 Chr.2: 227524928-227524929 on Build GRCh38
rs961223235 Chr.2: 227524946-227524946 on Build GRCh38
rs142310218 Chr.2: 227524805-227524805 on Build GRCh38
rs201688253 Chr.2: 227524936-227524936 on Build GRCh38
rs767158295 Chr.2: 227524847-227524847 on Build GRCh38
rs746280113 Chr.2: 227524913-227524913 on Build GRCh38
rs1051000722 Chr.2: 227524798-227524798 on Build GRCh38

View Full Product Details