Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11194578-11194847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL7
Entrez Gene ID
10218
Gene Name
angiopoietin like 7
Gene Aliases
AngX, CDT6, dJ647M16.1
UniGene
Hs.146559
Gene Chromosome Location
Chr.1: 11189289-11195981 on Build GRCh38
Transcripts
NM_021146.3 NM_004958.3 XM_005263438.2 XM_011541166.2 XM_017000900.1 XM_017000901.1 XM_017000902.1

Gene Symbol
MTOR
Entrez Gene ID
2475
Gene Name
mechanistic target of rapamycin
Gene Aliases
FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS
UniGene
Hs.338207
Gene Chromosome Location
Chr.1: 11106531-11262551 on Build GRCh38
Transcripts
NM_021146.3 NM_004958.3 XM_005263438.2 XM_011541166.2 XM_017000900.1 XM_017000901.1 XM_017000902.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6506906 c.855T>A p.C285* Chr.1: 11194643-11194643 on Build GRCh38
4142150 c.840C>G p.N280K Chr.1: 11194628-11194628 on Build GRCh38
893901 c.865C>T p.R289C Chr.1: 11194653-11194653 on Build GRCh38
4791104 c.828G>A p.K276K Chr.1: 11194616-11194616 on Build GRCh38
1332703 c.792G>C p.Q264H Chr.1: 11194580-11194580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373848807 Chr.1: 11194702-11194702 on Build GRCh38
rs1022839881 Chr.1: 11194759-11194759 on Build GRCh38
rs991369985 Chr.1: 11194815-11194815 on Build GRCh38
rs760316438 Chr.1: 11194695-11194695 on Build GRCh38
rs182647808 Chr.1: 11194641-11194641 on Build GRCh38
rs930577660 Chr.1: 11194607-11194607 on Build GRCh38
rs766005976 Chr.1: 11194698-11194698 on Build GRCh38
rs758931322 Chr.1: 11194643-11194643 on Build GRCh38
rs199596424 Chr.1: 11194665-11194665 on Build GRCh38
rs773850987 Chr.1: 11194622-11194622 on Build GRCh38
rs774185284 Chr.1: 11194678-11194678 on Build GRCh38
rs749047731 Chr.1: 11194671-11194671 on Build GRCh38
rs767116119 Chr.1: 11194632-11194632 on Build GRCh38
rs577199443 Chr.1: 11194668-11194669 on Build GRCh38
rs989872411 Chr.1: 11194802-11194802 on Build GRCh38
rs910797684 Chr.1: 11194617-11194617 on Build GRCh38
rs1048350672 Chr.1: 11194627-11194627 on Build GRCh38
rs772748275 Chr.1: 11194692-11194692 on Build GRCh38
rs112845751 Chr.1: 11194677-11194677 on Build GRCh38
rs1013349536 Chr.1: 11194724-11194724 on Build GRCh38
rs750098318 Chr.1: 11194640-11194640 on Build GRCh38
rs759220907 Chr.1: 11194789-11194789 on Build GRCh38
rs376581686 Chr.1: 11194808-11194808 on Build GRCh38
rs12143172 Chr.1: 11194786-11194786 on Build GRCh38
rs777514998 Chr.1: 11194711-11194711 on Build GRCh38
rs766991443 Chr.1: 11194686-11194686 on Build GRCh38
rs753071795 Chr.1: 11194642-11194642 on Build GRCh38
rs775339107 Chr.1: 11194670-11194670 on Build GRCh38
rs747046261 Chr.1: 11194651-11194651 on Build GRCh38
rs763361214 Chr.1: 11194731-11194731 on Build GRCh38
rs567905477 Chr.1: 11194791-11194791 on Build GRCh38
rs368746195 Chr.1: 11194625-11194625 on Build GRCh38
rs778158402 Chr.1: 11194650-11194650 on Build GRCh38
rs187022848 Chr.1: 11194780-11194780 on Build GRCh38
rs951928385 Chr.1: 11194801-11194801 on Build GRCh38
rs1046916466 Chr.1: 11194657-11194657 on Build GRCh38
rs200767191 Chr.1: 11194683-11194683 on Build GRCh38
rs1008601354 Chr.1: 11194765-11194765 on Build GRCh38
rs916591081 Chr.1: 11194647-11194647 on Build GRCh38
rs887750725 Chr.1: 11194715-11194715 on Build GRCh38
rs781519687 Chr.1: 11194654-11194654 on Build GRCh38
rs897910471 Chr.1: 11194669-11194669 on Build GRCh38
rs953021737 Chr.1: 11194756-11194756 on Build GRCh38
rs752137567 Chr.1: 11194818-11194818 on Build GRCh38
rs746069379 Chr.1: 11194660-11194660 on Build GRCh38
rs147373987 Chr.1: 11194736-11194736 on Build GRCh38
rs753049976 Chr.1: 11194700-11194700 on Build GRCh38
rs772486834 Chr.1: 11194812-11194813 on Build GRCh38
rs757447081 Chr.1: 11194653-11194653 on Build GRCh38

View Full Product Details