Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 129751945-129752202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XPNPEP2
Entrez Gene ID
7512
Gene Name
X-prolyl aminopeptidase 2
Gene Aliases
AEACEI, APP2
UniGene
Hs.170499
Gene Chromosome Location
Chr.X: 129738970-129769549 on Build GRCh38
Transcripts
NM_003399.5

SNP Detail

SNP ID Chromosome Location
rs779207668 Chr.X: 129751998-129751998 on Build GRCh38
rs370630291 Chr.X: 129752131-129752131 on Build GRCh38
rs981242401 Chr.X: 129752012-129752012 on Build GRCh38
rs745990596 Chr.X: 129752077-129752077 on Build GRCh38
rs1771602 Chr.X: 129752009-129752009 on Build GRCh38
rs377568487 Chr.X: 129752170-129752170 on Build GRCh38
rs375045624 Chr.X: 129752166-129752166 on Build GRCh38
rs903692737 Chr.X: 129751992-129751993 on Build GRCh38
rs140648206 Chr.X: 129752174-129752174 on Build GRCh38
rs776450426 Chr.X: 129752125-129752125 on Build GRCh38
rs1038916838 Chr.X: 129752008-129752008 on Build GRCh38
rs759257272 Chr.X: 129752132-129752132 on Build GRCh38
rs775059212 Chr.X: 129752169-129752169 on Build GRCh38
rs767940039 Chr.X: 129752179-129752179 on Build GRCh38
rs138907933 Chr.X: 129751974-129751974 on Build GRCh38
rs747000784 Chr.X: 129752105-129752105 on Build GRCh38
rs752088523 Chr.X: 129752171-129752171 on Build GRCh38
rs772309488 Chr.X: 129752049-129752049 on Build GRCh38
rs750777576 Chr.X: 129752044-129752044 on Build GRCh38
rs377457991 Chr.X: 129752114-129752114 on Build GRCh38

View Full Product Details