Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155208737-155208980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTX1
Entrez Gene ID
4580
Gene Name
metaxin 1
Gene Aliases
MTX, MTXN
UniGene
Hs.490874
Gene Chromosome Location
Chr.1: 155208699-155213839 on Build GRCh38
Transcripts
NM_002455.4 NM_198883.3 XM_011509931.1 XM_011509933.1 XM_011509935.1

Gene Symbol
THBS3
Entrez Gene ID
7059
Gene Name
thrombospondin 3
Gene Aliases
TSP3
UniGene
Hs.169875
Gene Chromosome Location
Chr.1: 155195588-155209180 on Build GRCh38
Transcripts
NM_002455.4 NM_198883.3 XM_011509931.1 XM_011509933.1 XM_011509935.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6321710 c.23G>T p.R8L Chr.1: 155208827-155208827 on Build GRCh38
6201732 c.140C>T p.A47V Chr.1: 155208944-155208944 on Build GRCh38
1600915 c.85G>C p.G29R Chr.1: 155208889-155208889 on Build GRCh38
6037206 c.16C>A p.P6T Chr.1: 155208820-155208820 on Build GRCh38
1741230 c.35C>T p.S12L Chr.1: 155208839-155208839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934439730 Chr.1: 155208765-155208765 on Build GRCh38
rs757046514 Chr.1: 155208869-155208869 on Build GRCh38
rs34921532 Chr.1: 155208955-155208955 on Build GRCh38
rs999290958 Chr.1: 155208819-155208819 on Build GRCh38
rs1052880669 Chr.1: 155208768-155208768 on Build GRCh38
rs752637621 Chr.1: 155208818-155208819 on Build GRCh38
rs771102265 Chr.1: 155208852-155208857 on Build GRCh38
rs748319329 Chr.1: 155208916-155208916 on Build GRCh38
rs775010955 Chr.1: 155208825-155208825 on Build GRCh38
rs974516355 Chr.1: 155208796-155208796 on Build GRCh38
rs752313659 Chr.1: 155208781-155208781 on Build GRCh38
rs371540197 Chr.1: 155208814-155208814 on Build GRCh38
rs774957655 Chr.1: 155208947-155208947 on Build GRCh38
rs747037609 Chr.1: 155208807-155208807 on Build GRCh38
rs766966387 Chr.1: 155208853-155208853 on Build GRCh38
rs768645002 Chr.1: 155208813-155208813 on Build GRCh38
rs755845788 Chr.1: 155208820-155208820 on Build GRCh38
rs751111095 Chr.1: 155208772-155208772 on Build GRCh38
rs753679940 Chr.1: 155208787-155208787 on Build GRCh38
rs773753911 Chr.1: 155208763-155208763 on Build GRCh38
rs539980904 Chr.1: 155208852-155208852 on Build GRCh38
rs750361868 Chr.1: 155208882-155208882 on Build GRCh38
rs1016386132 Chr.1: 155208790-155208791 on Build GRCh38
rs770138171 Chr.1: 155208932-155208932 on Build GRCh38
rs544260151 Chr.1: 155208917-155208917 on Build GRCh38
rs138982566 Chr.1: 155208915-155208915 on Build GRCh38
rs770105309 Chr.1: 155208821-155208821 on Build GRCh38
rs201584620 Chr.1: 155208954-155208954 on Build GRCh38
rs753621959 Chr.1: 155208867-155208867 on Build GRCh38
rs746005856 Chr.1: 155208858-155208871 on Build GRCh38
rs576992799 Chr.1: 155208859-155208859 on Build GRCh38
rs772698213 Chr.1: 155208940-155208940 on Build GRCh38
rs779029977 Chr.1: 155208852-155208853 on Build GRCh38
rs368634289 Chr.1: 155208864-155208864 on Build GRCh38
rs763532919 Chr.1: 155208764-155208764 on Build GRCh38
rs375103223 Chr.1: 155208769-155208769 on Build GRCh38
rs781174422 Chr.1: 155208810-155208835 on Build GRCh38
rs748289669 Chr.1: 155208820-155208820 on Build GRCh38
rs749607249 Chr.1: 155208941-155208941 on Build GRCh38
rs765554307 Chr.1: 155208760-155208760 on Build GRCh38
rs758403843 Chr.1: 155208901-155208901 on Build GRCh38
rs754450710 Chr.1: 155208776-155208776 on Build GRCh38
rs377309496 Chr.1: 155208823-155208823 on Build GRCh38
rs762192691 Chr.1: 155208758-155208758 on Build GRCh38
rs778777390 Chr.1: 155208880-155208880 on Build GRCh38
rs774846663 Chr.1: 155208956-155208956 on Build GRCh38
rs759927568 Chr.1: 155208950-155208950 on Build GRCh38
rs367798895 Chr.1: 155208786-155208786 on Build GRCh38
rs767139762 Chr.1: 155208780-155208780 on Build GRCh38
rs963070351 Chr.1: 155208792-155208792 on Build GRCh38
rs925310076 Chr.1: 155208834-155208834 on Build GRCh38
rs1055156452 Chr.1: 155208927-155208927 on Build GRCh38
rs191739539 Chr.1: 155208793-155208793 on Build GRCh38
rs760296290 Chr.1: 155208962-155208962 on Build GRCh38
rs757169344 Chr.1: 155208790-155208790 on Build GRCh38
rs778147019 Chr.1: 155208937-155208937 on Build GRCh38
rs759037263 Chr.1: 155208849-155208849 on Build GRCh38
rs951001345 Chr.1: 155208770-155208770 on Build GRCh38
rs777452259 Chr.1: 155208819-155208820 on Build GRCh38
rs558723935 Chr.1: 155208855-155208855 on Build GRCh38

View Full Product Details