Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54014161-54014434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDLRAD1
Entrez Gene ID
388633
Gene Name
low density lipoprotein receptor class A domain containing 1
Gene Aliases
-
UniGene
Hs.439583
Gene Chromosome Location
Chr.1: 54007298-54018186 on Build GRCh38
Transcripts
NM_001010978.3 NM_001276392.1 NM_001276393.1 NM_001276394.1 NM_001276395.1 XM_011541446.1 XM_017001264.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
50312 c.160A>G p.I54V Chr.1: 54014278-54014278 on Build GRCh38
107920 c.181G>A p.G61R Chr.1: 54014257-54014257 on Build GRCh38
5547548 c.189_199del11 p.S64fs*18 Chr.1: 54014239-54014249 on Build GRCh38
106350 c.200C>T p.P67L Chr.1: 54014238-54014238 on Build GRCh38
5555146 c.79G>A p.G27S Chr.1: 54014359-54014359 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919342124 Chr.1: 54014379-54014379 on Build GRCh38
rs374253464 Chr.1: 54014182-54014182 on Build GRCh38
rs867095762 Chr.1: 54014286-54014286 on Build GRCh38
rs111862115 Chr.1: 54014228-54014228 on Build GRCh38
rs780500333 Chr.1: 54014371-54014371 on Build GRCh38
rs200640296 Chr.1: 54014337-54014337 on Build GRCh38
rs1047034814 Chr.1: 54014304-54014304 on Build GRCh38
rs769666186 Chr.1: 54014392-54014392 on Build GRCh38
rs1044870644 Chr.1: 54014255-54014255 on Build GRCh38
rs149114405 Chr.1: 54014338-54014338 on Build GRCh38
rs866087332 Chr.1: 54014236-54014236 on Build GRCh38
rs899543884 Chr.1: 54014354-54014354 on Build GRCh38
rs752067669 Chr.1: 54014204-54014204 on Build GRCh38
rs374942731 Chr.1: 54014371-54014371 on Build GRCh38
rs767339237 Chr.1: 54014226-54014226 on Build GRCh38
rs570661160 Chr.1: 54014200-54014200 on Build GRCh38
rs896069941 Chr.1: 54014231-54014231 on Build GRCh38
rs562281788 Chr.1: 54014393-54014393 on Build GRCh38
rs745672207 Chr.1: 54014296-54014296 on Build GRCh38
rs751067486 Chr.1: 54014370-54014370 on Build GRCh38
rs957871567 Chr.1: 54014381-54014381 on Build GRCh38
rs745810760 Chr.1: 54014373-54014373 on Build GRCh38
rs746563764 Chr.1: 54014248-54014248 on Build GRCh38
rs777453751 Chr.1: 54014362-54014362 on Build GRCh38
rs906398648 Chr.1: 54014266-54014266 on Build GRCh38
rs368717070 Chr.1: 54014245-54014245 on Build GRCh38
rs369288149 Chr.1: 54014413-54014413 on Build GRCh38
rs770593287 Chr.1: 54014192-54014192 on Build GRCh38
rs776456624 Chr.1: 54014193-54014193 on Build GRCh38
rs955580498 Chr.1: 54014361-54014361 on Build GRCh38
rs201278228 Chr.1: 54014332-54014332 on Build GRCh38
rs757810555 Chr.1: 54014219-54014219 on Build GRCh38
rs767947971 Chr.1: 54014225-54014225 on Build GRCh38
rs372096942 Chr.1: 54014340-54014340 on Build GRCh38
rs138083713 Chr.1: 54014314-54014314 on Build GRCh38
rs990126746 Chr.1: 54014388-54014388 on Build GRCh38
rs764227423 Chr.1: 54014331-54014331 on Build GRCh38
rs866943826 Chr.1: 54014359-54014359 on Build GRCh38
rs539528425 Chr.1: 54014227-54014227 on Build GRCh38
rs773651225 Chr.1: 54014414-54014414 on Build GRCh38
rs973531843 Chr.1: 54014205-54014205 on Build GRCh38
rs141441714 Chr.1: 54014354-54014356 on Build GRCh38
rs761844221 Chr.1: 54014333-54014333 on Build GRCh38
rs151154139 Chr.1: 54014257-54014257 on Build GRCh38
rs76122098 Chr.1: 54014313-54014313 on Build GRCh38
rs555799872 Chr.1: 54014276-54014276 on Build GRCh38
rs770149196 Chr.1: 54014412-54014412 on Build GRCh38
rs758017277 Chr.1: 54014357-54014357 on Build GRCh38
rs765781877 Chr.1: 54014308-54014315 on Build GRCh38
rs1029780200 Chr.1: 54014360-54014360 on Build GRCh38
rs143163846 Chr.1: 54014238-54014238 on Build GRCh38
rs764284928 Chr.1: 54014203-54014203 on Build GRCh38
rs752296313 Chr.1: 54014348-54014348 on Build GRCh38
rs765484370 Chr.1: 54014199-54014199 on Build GRCh38
rs759299808 Chr.1: 54014196-54014196 on Build GRCh38
rs775800500 Chr.1: 54014325-54014325 on Build GRCh38
rs763292654 Chr.1: 54014326-54014326 on Build GRCh38
rs892241900 Chr.1: 54014334-54014334 on Build GRCh38
rs985907607 Chr.1: 54014190-54014190 on Build GRCh38
rs370365819 Chr.1: 54014285-54014285 on Build GRCh38
rs754545690 Chr.1: 54014370-54014371 on Build GRCh38
rs200507421 Chr.1: 54014232-54014232 on Build GRCh38
rs1031915450 Chr.1: 54014372-54014372 on Build GRCh38
rs757582315 Chr.1: 54014264-54014264 on Build GRCh38
rs1005693396 Chr.1: 54014191-54014191 on Build GRCh38
rs112985435 Chr.1: 54014328-54014328 on Build GRCh38
rs955590397 Chr.1: 54014216-54014216 on Build GRCh38
rs770881675 Chr.1: 54014263-54014263 on Build GRCh38
rs763064501 Chr.1: 54014202-54014202 on Build GRCh38

View Full Product Details