Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 110875394-110875653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LPAR1
Entrez Gene ID
1902
Gene Name
lysophosphatidic acid receptor 1
Gene Aliases
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
UniGene
Hs.126667
Gene Chromosome Location
Chr.9: 110873252-111039259 on Build GRCh38
Transcripts
NM_001401.3 NM_057159.2 XM_005251781.3 XM_005251782.3 XM_017014383.1 XM_017014384.1 XM_017014385.1 XM_017014386.1 XM_017014387.1 XM_017014388.1 XM_017014389.1 XM_017014390.1 XM_017014391.1 XM_017014392.1 XM_017014393.1 XM_017014394.1 XM_017014395.1 XM_017014396.1 XM_017014397.1 XM_017014398.1 XM_017014399.1 XM_017014400.1 XM_017014401.1 XM_017014402.1 XM_017014403.1 XM_017014404.1 XM_017014405.1 XM_017014406.1 XM_017014407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4698430 c.975C>A p.I325I Chr.9: 110875541-110875541 on Build GRCh38
1459297 c.942C>T p.R314R Chr.9: 110875574-110875574 on Build GRCh38
1330802 c.944A>G p.D315G Chr.9: 110875572-110875572 on Build GRCh38
137973 c.976C>T p.L326F Chr.9: 110875540-110875540 on Build GRCh38
6182067 c.968G>C p.R323T Chr.9: 110875548-110875548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187429734 Chr.9: 110875528-110875528 on Build GRCh38
rs778764227 Chr.9: 110875527-110875527 on Build GRCh38
rs147367410 Chr.9: 110875461-110875461 on Build GRCh38
rs916946397 Chr.9: 110875607-110875607 on Build GRCh38
rs759235908 Chr.9: 110875493-110875493 on Build GRCh38
rs369807009 Chr.9: 110875591-110875591 on Build GRCh38
rs868046435 Chr.9: 110875598-110875598 on Build GRCh38
rs1035543568 Chr.9: 110875511-110875511 on Build GRCh38
rs756095056 Chr.9: 110875432-110875432 on Build GRCh38
rs61753623 Chr.9: 110875559-110875559 on Build GRCh38
rs868098537 Chr.9: 110875556-110875556 on Build GRCh38
rs201640956 Chr.9: 110875523-110875523 on Build GRCh38
rs753749260 Chr.9: 110875512-110875512 on Build GRCh38
rs200850518 Chr.9: 110875602-110875602 on Build GRCh38
rs761832922 Chr.9: 110875476-110875476 on Build GRCh38
rs377117526 Chr.9: 110875418-110875418 on Build GRCh38
rs147557148 Chr.9: 110875462-110875462 on Build GRCh38
rs111935355 Chr.9: 110875417-110875417 on Build GRCh38
rs890550046 Chr.9: 110875456-110875456 on Build GRCh38
rs527765818 Chr.9: 110875485-110875485 on Build GRCh38
rs768789545 Chr.9: 110875470-110875470 on Build GRCh38
rs747270836 Chr.9: 110875553-110875553 on Build GRCh38
rs770237313 Chr.9: 110875483-110875483 on Build GRCh38
rs142117056 Chr.9: 110875510-110875510 on Build GRCh38
rs910875116 Chr.9: 110875600-110875600 on Build GRCh38
rs767182008 Chr.9: 110875499-110875499 on Build GRCh38
rs546256005 Chr.9: 110875488-110875488 on Build GRCh38
rs942312773 Chr.9: 110875616-110875616 on Build GRCh38
rs3739709 Chr.9: 110875574-110875574 on Build GRCh38
rs773653971 Chr.9: 110875576-110875576 on Build GRCh38
rs760775038 Chr.9: 110875484-110875484 on Build GRCh38
rs1049844 Chr.9: 110875436-110875436 on Build GRCh38
rs769855870 Chr.9: 110875575-110875575 on Build GRCh38
rs777712568 Chr.9: 110875434-110875434 on Build GRCh38
rs139563572 Chr.9: 110875463-110875463 on Build GRCh38
rs751134225 Chr.9: 110875510-110875518 on Build GRCh38
rs1049843 Chr.9: 110875498-110875498 on Build GRCh38
rs959967577 Chr.9: 110875518-110875518 on Build GRCh38
rs752274065 Chr.9: 110875502-110875502 on Build GRCh38
rs752725919 Chr.9: 110875425-110875425 on Build GRCh38
rs145864224 Chr.9: 110875558-110875558 on Build GRCh38
rs979976981 Chr.9: 110875490-110875490 on Build GRCh38
rs1048242692 Chr.9: 110875440-110875440 on Build GRCh38

View Full Product Details