Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5412496-5412729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NET1
Entrez Gene ID
10276
Gene Name
neuroepithelial cell transforming 1
Gene Aliases
ARHGEF8, NET1A
UniGene
Hs.25155
Gene Chromosome Location
Chr.10: 5412551-5459056 on Build GRCh38
Transcripts
NM_001047160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6436875 c.17C>T p.A6V Chr.10: 5412709-5412709 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs926407723 Chr.10: 5412519-5412519 on Build GRCh38
rs1033402924 Chr.10: 5412632-5412632 on Build GRCh38
rs775840996 Chr.10: 5412698-5412698 on Build GRCh38
rs985797530 Chr.10: 5412584-5412584 on Build GRCh38
rs962083936 Chr.10: 5412575-5412575 on Build GRCh38
rs944633602 Chr.10: 5412613-5412613 on Build GRCh38
rs937736142 Chr.10: 5412535-5412535 on Build GRCh38
rs895951784 Chr.10: 5412572-5412572 on Build GRCh38
rs1054509205 Chr.10: 5412661-5412661 on Build GRCh38
rs1042011307 Chr.10: 5412598-5412598 on Build GRCh38
rs955231589 Chr.10: 5412677-5412677 on Build GRCh38
rs376475251 Chr.10: 5412580-5412580 on Build GRCh38
rs1000229985 Chr.10: 5412616-5412616 on Build GRCh38
rs894553250 Chr.10: 5412671-5412671 on Build GRCh38
rs898180617 Chr.10: 5412640-5412640 on Build GRCh38
rs576200765 Chr.10: 5412675-5412675 on Build GRCh38
rs973217596 Chr.10: 5412577-5412577 on Build GRCh38
rs899917805 Chr.10: 5412633-5412633 on Build GRCh38
rs1029744866 Chr.10: 5412641-5412641 on Build GRCh38
rs1055888925 Chr.10: 5412569-5412569 on Build GRCh38
rs996901459 Chr.10: 5412638-5412638 on Build GRCh38
rs903414772 Chr.10: 5412609-5412609 on Build GRCh38
rs745904750 Chr.10: 5412681-5412681 on Build GRCh38
rs770064622 Chr.10: 5412686-5412686 on Build GRCh38

View Full Product Details