Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4448193-4448454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBXN6
Entrez Gene ID
80700
Gene Name
UBX domain protein 6
Gene Aliases
UBXD1, UBXDC2
UniGene
Hs.435255
Gene Chromosome Location
Chr.19: 4445006-4458759 on Build GRCh38
Transcripts
NM_001171091.1 NM_025241.2 XM_017027325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7209469 c.463_464insCG p.A156fs*5 Chr.19: 4448393-4448394 on Build GRCh38
5031729 c.477G>A p.M159I Chr.19: 4448380-4448380 on Build GRCh38
1393459 c.475A>T p.M159L Chr.19: 4448382-4448382 on Build GRCh38
4495149 c.447C>T p.F149F Chr.19: 4448410-4448410 on Build GRCh38
4977083 c.509G>T p.R170L Chr.19: 4448348-4448348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781209925 Chr.19: 4448305-4448305 on Build GRCh38
rs932096718 Chr.19: 4448229-4448229 on Build GRCh38
rs369398559 Chr.19: 4448391-4448391 on Build GRCh38
rs139656461 Chr.19: 4448365-4448365 on Build GRCh38
rs756042421 Chr.19: 4448369-4448369 on Build GRCh38
rs149717969 Chr.19: 4448368-4448368 on Build GRCh38
rs73541319 Chr.19: 4448228-4448228 on Build GRCh38
rs538789592 Chr.19: 4448426-4448426 on Build GRCh38
rs953202807 Chr.19: 4448303-4448303 on Build GRCh38
rs904470368 Chr.19: 4448256-4448256 on Build GRCh38
rs942205117 Chr.19: 4448214-4448214 on Build GRCh38
rs764193289 Chr.19: 4448338-4448338 on Build GRCh38
rs769092781 Chr.19: 4448423-4448423 on Build GRCh38
rs777310844 Chr.19: 4448377-4448377 on Build GRCh38
rs755597683 Chr.19: 4448283-4448284 on Build GRCh38
rs766246007 Chr.19: 4448286-4448286 on Build GRCh38
rs745536160 Chr.19: 4448403-4448403 on Build GRCh38
rs199967454 Chr.19: 4448278-4448278 on Build GRCh38
rs987517747 Chr.19: 4448399-4448399 on Build GRCh38
rs1041641237 Chr.19: 4448255-4448255 on Build GRCh38
rs746295954 Chr.19: 4448315-4448315 on Build GRCh38
rs953439135 Chr.19: 4448392-4448392 on Build GRCh38
rs754760001 Chr.19: 4448352-4448352 on Build GRCh38
rs761106610 Chr.19: 4448337-4448337 on Build GRCh38
rs747520058 Chr.19: 4448406-4448406 on Build GRCh38
rs946058729 Chr.19: 4448430-4448430 on Build GRCh38
rs779457601 Chr.19: 4448306-4448330 on Build GRCh38
rs113855338 Chr.19: 4448335-4448335 on Build GRCh38
rs755604057 Chr.19: 4448311-4448311 on Build GRCh38
rs754963132 Chr.19: 4448300-4448300 on Build GRCh38
rs35081974 Chr.19: 4448255-4448256 on Build GRCh38
rs974019954 Chr.19: 4448220-4448220 on Build GRCh38
rs147213791 Chr.19: 4448358-4448358 on Build GRCh38
rs755247775 Chr.19: 4448215-4448215 on Build GRCh38
rs1000017959 Chr.19: 4448271-4448271 on Build GRCh38
rs997343025 Chr.19: 4448265-4448265 on Build GRCh38
rs945913861 Chr.19: 4448242-4448242 on Build GRCh38
rs770615062 Chr.19: 4448314-4448314 on Build GRCh38
rs1051685860 Chr.19: 4448232-4448232 on Build GRCh38
rs890474132 Chr.19: 4448241-4448241 on Build GRCh38
rs202166922 Chr.19: 4448282-4448282 on Build GRCh38
rs776614212 Chr.19: 4448339-4448339 on Build GRCh38
rs752069457 Chr.19: 4448267-4448268 on Build GRCh38
rs748810938 Chr.19: 4448381-4448381 on Build GRCh38
rs776766795 Chr.19: 4448425-4448425 on Build GRCh38
rs1004320073 Chr.19: 4448346-4448346 on Build GRCh38
rs762081150 Chr.19: 4448340-4448340 on Build GRCh38
rs757451645 Chr.19: 4448285-4448285 on Build GRCh38
rs973529485 Chr.19: 4448375-4448375 on Build GRCh38
rs143704241 Chr.19: 4448350-4448350 on Build GRCh38
rs748050606 Chr.19: 4448307-4448307 on Build GRCh38
rs772380278 Chr.19: 4448404-4448404 on Build GRCh38
rs778473018 Chr.19: 4448396-4448396 on Build GRCh38
rs894203541 Chr.19: 4448279-4448279 on Build GRCh38
rs781312851 Chr.19: 4448348-4448348 on Build GRCh38
rs748942813 Chr.19: 4448312-4448312 on Build GRCh38
rs753495583 Chr.19: 4448376-4448378 on Build GRCh38

View Full Product Details