Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2317365-2317590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGM3
Entrez Gene ID
7053
Gene Name
transglutaminase 3
Gene Aliases
TGE
UniGene
Hs.2022
Gene Chromosome Location
Chr.20: 2295967-2341079 on Build GRCh38
Transcripts
NM_003245.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3544681 c.949G>A p.G317R Chr.20: 2317451-2317451 on Build GRCh38
443538 c.884C>A p.T295N Chr.20: 2317386-2317386 on Build GRCh38
1713076 c.950G>A p.G317E Chr.20: 2317452-2317452 on Build GRCh38
1713077 c.953_954AC>TT p.N318I Chr.20: 2317455-2317456 on Build GRCh38
1713078 c.968G>A p.G323D Chr.20: 2317470-2317470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759645776 Chr.20: 2317422-2317422 on Build GRCh38
rs376202925 Chr.20: 2317441-2317441 on Build GRCh38
rs185040767 Chr.20: 2317526-2317526 on Build GRCh38
rs374601295 Chr.20: 2317440-2317440 on Build GRCh38
rs764317584 Chr.20: 2317456-2317456 on Build GRCh38
rs772526693 Chr.20: 2317395-2317395 on Build GRCh38
rs370570929 Chr.20: 2317434-2317434 on Build GRCh38
rs562802461 Chr.20: 2317416-2317416 on Build GRCh38
rs766252464 Chr.20: 2317502-2317502 on Build GRCh38
rs760405976 Chr.20: 2317500-2317500 on Build GRCh38
rs778285811 Chr.20: 2317465-2317465 on Build GRCh38
rs376340232 Chr.20: 2317515-2317515 on Build GRCh38
rs761712369 Chr.20: 2317430-2317430 on Build GRCh38
rs370121854 Chr.20: 2317481-2317481 on Build GRCh38
rs753583774 Chr.20: 2317511-2317511 on Build GRCh38
rs767356864 Chr.20: 2317483-2317483 on Build GRCh38
rs747341040 Chr.20: 2317468-2317468 on Build GRCh38
rs768237602 Chr.20: 2317389-2317389 on Build GRCh38
rs866351901 Chr.20: 2317539-2317539 on Build GRCh38
rs776863698 Chr.20: 2317477-2317477 on Build GRCh38
rs769905860 Chr.20: 2317480-2317480 on Build GRCh38
rs539342653 Chr.20: 2317461-2317461 on Build GRCh38
rs182697627 Chr.20: 2317458-2317458 on Build GRCh38
rs754582243 Chr.20: 2317449-2317449 on Build GRCh38
rs904635380 Chr.20: 2317558-2317558 on Build GRCh38
rs757617451 Chr.20: 2317510-2317518 on Build GRCh38
rs998889492 Chr.20: 2317569-2317569 on Build GRCh38
rs753489818 Chr.20: 2317442-2317442 on Build GRCh38
rs572591561 Chr.20: 2317545-2317545 on Build GRCh38
rs938583326 Chr.20: 2317488-2317488 on Build GRCh38
rs1055871365 Chr.20: 2317388-2317388 on Build GRCh38
rs375169739 Chr.20: 2317551-2317551 on Build GRCh38
rs747503416 Chr.20: 2317529-2317529 on Build GRCh38
rs757859321 Chr.20: 2317517-2317517 on Build GRCh38
rs748641552 Chr.20: 2317390-2317390 on Build GRCh38
rs377223591 Chr.20: 2317427-2317427 on Build GRCh38
rs767417627 Chr.20: 2317432-2317432 on Build GRCh38
rs746070685 Chr.20: 2317479-2317479 on Build GRCh38
rs754673741 Chr.20: 2317512-2317512 on Build GRCh38
rs751549420 Chr.20: 2317428-2317428 on Build GRCh38
rs778533516 Chr.20: 2317457-2317457 on Build GRCh38
rs773159254 Chr.20: 2317489-2317489 on Build GRCh38
rs1043264299 Chr.20: 2317540-2317540 on Build GRCh38
rs146355154 Chr.20: 2317415-2317415 on Build GRCh38
rs141606749 Chr.20: 2317484-2317484 on Build GRCh38
rs771288538 Chr.20: 2317469-2317469 on Build GRCh38
rs368213901 Chr.20: 2317552-2317552 on Build GRCh38
rs376191636 Chr.20: 2317508-2317508 on Build GRCh38
rs778176820 Chr.20: 2317519-2317519 on Build GRCh38
rs570209478 Chr.20: 2317514-2317514 on Build GRCh38
rs773443568 Chr.20: 2317404-2317404 on Build GRCh38

View Full Product Details