Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40262444-40262717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WIPF2
Entrez Gene ID
147179
Gene Name
WAS/WASL interacting protein family member 2
Gene Aliases
WICH, WIRE
UniGene
Hs.421622
Gene Chromosome Location
Chr.17: 40219304-40284136 on Build GRCh38
Transcripts
NM_133264.4 XM_005257083.2 XM_005257084.2 XM_011524412.1 XM_011524413.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3717333 c.210C>G p.S70R Chr.17: 40262538-40262538 on Build GRCh38
4741252 c.303G>T p.K101N Chr.17: 40262631-40262631 on Build GRCh38
3516991 c.307G>A p.G103S Chr.17: 40262635-40262635 on Build GRCh38
213398 c.238G>T p.A80S Chr.17: 40262566-40262566 on Build GRCh38
148264 c.283C>T p.L95F Chr.17: 40262611-40262611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141891164 Chr.17: 40262514-40262514 on Build GRCh38
rs139822756 Chr.17: 40262528-40262528 on Build GRCh38
rs774324531 Chr.17: 40262632-40262632 on Build GRCh38
rs751900012 Chr.17: 40262503-40262503 on Build GRCh38
rs765324585 Chr.17: 40262489-40262489 on Build GRCh38
rs763121929 Chr.17: 40262555-40262555 on Build GRCh38
rs1021167402 Chr.17: 40262679-40262679 on Build GRCh38
rs368856744 Chr.17: 40262546-40262546 on Build GRCh38
rs754415148 Chr.17: 40262589-40262589 on Build GRCh38
rs750892977 Chr.17: 40262505-40262505 on Build GRCh38
rs376837833 Chr.17: 40262594-40262594 on Build GRCh38
rs754281859 Chr.17: 40262666-40262666 on Build GRCh38
rs1004428907 Chr.17: 40262549-40262549 on Build GRCh38
rs371864156 Chr.17: 40262504-40262504 on Build GRCh38
rs760538904 Chr.17: 40262474-40262474 on Build GRCh38
rs778530020 Chr.17: 40262521-40262521 on Build GRCh38
rs774576183 Chr.17: 40262569-40262569 on Build GRCh38
rs763177173 Chr.17: 40262498-40262498 on Build GRCh38
rs775685539 Chr.17: 40262490-40262490 on Build GRCh38
rs767882438 Chr.17: 40262576-40262576 on Build GRCh38
rs958007741 Chr.17: 40262664-40262664 on Build GRCh38
rs770972333 Chr.17: 40262543-40262543 on Build GRCh38
rs765776207 Chr.17: 40262667-40262667 on Build GRCh38
rs766856730 Chr.17: 40262587-40262587 on Build GRCh38
rs778403469 Chr.17: 40262592-40262592 on Build GRCh38
rs746865748 Chr.17: 40262541-40262541 on Build GRCh38
rs776155027 Chr.17: 40262660-40262660 on Build GRCh38
rs565623956 Chr.17: 40262625-40262625 on Build GRCh38
rs780577025 Chr.17: 40262508-40262508 on Build GRCh38
rs771838533 Chr.17: 40262529-40262529 on Build GRCh38
rs989702503 Chr.17: 40262672-40262672 on Build GRCh38
rs763540033 Chr.17: 40262495-40262495 on Build GRCh38
rs369729357 Chr.17: 40262660-40262660 on Build GRCh38
rs752129880 Chr.17: 40262668-40262668 on Build GRCh38
rs757986483 Chr.17: 40262673-40262673 on Build GRCh38
rs776770272 Chr.17: 40262548-40262548 on Build GRCh38
rs761992936 Chr.17: 40262640-40262640 on Build GRCh38
rs772160728 Chr.17: 40262646-40262646 on Build GRCh38
rs768809051 Chr.17: 40262631-40262631 on Build GRCh38
rs756549259 Chr.17: 40262585-40262585 on Build GRCh38
rs750722091 Chr.17: 40262579-40262579 on Build GRCh38
rs772915533 Chr.17: 40262539-40262539 on Build GRCh38
rs890200456 Chr.17: 40262472-40262472 on Build GRCh38
rs756609377 Chr.17: 40262506-40262506 on Build GRCh38
rs764399419 Chr.17: 40262502-40262502 on Build GRCh38
rs773539967 Chr.17: 40262654-40262654 on Build GRCh38
rs777289348 Chr.17: 40262691-40262691 on Build GRCh38
rs150747102 Chr.17: 40262615-40262615 on Build GRCh38
rs367652270 Chr.17: 40262512-40262512 on Build GRCh38
rs761019731 Chr.17: 40262655-40262655 on Build GRCh38
rs757960999 Chr.17: 40262604-40262604 on Build GRCh38
rs770917313 Chr.17: 40262620-40262620 on Build GRCh38
rs141184179 Chr.17: 40262564-40262564 on Build GRCh38
rs1040835799 Chr.17: 40262678-40262689 on Build GRCh38
rs777534743 Chr.17: 40262606-40262606 on Build GRCh38

View Full Product Details