Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205663291-205663528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC45A3
Entrez Gene ID
85414
Gene Name
solute carrier family 45 member 3
Gene Aliases
IPCA-2, IPCA-6, IPCA-8, IPCA6, PCANAP2, PCANAP6, PCANAP8, PRST
UniGene
Hs.278695
Gene Chromosome Location
Chr.1: 205657851-205680502 on Build GRCh38
Transcripts
NM_033102.2 XM_005245556.3 XM_005245557.4 XM_005245560.2 XM_017002638.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6243750 c.356C>A p.P119H Chr.1: 205663435-205663435 on Build GRCh38
4548793 c.456G>A p.R152R Chr.1: 205663335-205663335 on Build GRCh38
2215443 c.354G>A p.R118R Chr.1: 205663437-205663437 on Build GRCh38
1473313 c.385G>T p.V129L Chr.1: 205663406-205663406 on Build GRCh38
4027471 c.473G>A p.R158H Chr.1: 205663318-205663318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748067402 Chr.1: 205663387-205663387 on Build GRCh38
rs935846440 Chr.1: 205663419-205663419 on Build GRCh38
rs764477376 Chr.1: 205663438-205663438 on Build GRCh38
rs755721150 Chr.1: 205663350-205663350 on Build GRCh38
rs749053831 Chr.1: 205663367-205663367 on Build GRCh38
rs752738443 Chr.1: 205663500-205663500 on Build GRCh38
rs555794455 Chr.1: 205663394-205663394 on Build GRCh38
rs868374522 Chr.1: 205663434-205663434 on Build GRCh38
rs758404940 Chr.1: 205663502-205663502 on Build GRCh38
rs866353899 Chr.1: 205663313-205663313 on Build GRCh38
rs376727859 Chr.1: 205663397-205663397 on Build GRCh38
rs571524594 Chr.1: 205663318-205663318 on Build GRCh38
rs535560877 Chr.1: 205663368-205663368 on Build GRCh38
rs775721751 Chr.1: 205663413-205663413 on Build GRCh38
rs779692620 Chr.1: 205663365-205663365 on Build GRCh38
rs765973160 Chr.1: 205663473-205663473 on Build GRCh38
rs763246487 Chr.1: 205663425-205663425 on Build GRCh38
rs747821251 Chr.1: 205663428-205663432 on Build GRCh38
rs367899036 Chr.1: 205663337-205663337 on Build GRCh38
rs760177726 Chr.1: 205663472-205663472 on Build GRCh38
rs768368689 Chr.1: 205663446-205663446 on Build GRCh38
rs749944670 Chr.1: 205663459-205663459 on Build GRCh38
rs141243660 Chr.1: 205663454-205663454 on Build GRCh38
rs779026388 Chr.1: 205663374-205663374 on Build GRCh38
rs759809054 Chr.1: 205663319-205663319 on Build GRCh38
rs770220291 Chr.1: 205663433-205663434 on Build GRCh38
rs766979336 Chr.1: 205663340-205663340 on Build GRCh38
rs199734030 Chr.1: 205663341-205663341 on Build GRCh38
rs145880670 Chr.1: 205663482-205663482 on Build GRCh38
rs778829876 Chr.1: 205663485-205663485 on Build GRCh38
rs150450020 Chr.1: 205663336-205663336 on Build GRCh38
rs151325374 Chr.1: 205663330-205663330 on Build GRCh38
rs200709431 Chr.1: 205663324-205663324 on Build GRCh38
rs769911329 Chr.1: 205663407-205663407 on Build GRCh38
rs574763250 Chr.1: 205663329-205663329 on Build GRCh38
rs912558398 Chr.1: 205663314-205663314 on Build GRCh38
rs776836737 Chr.1: 205663406-205663406 on Build GRCh38

View Full Product Details