Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 43046241-43046468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100506679
Entrez Gene ID
100506679
Gene Name
uncharacterized LOC100506679
Gene Aliases
-
UniGene
Hs.706553
Gene Chromosome Location
Chr.22: 43038585-43052366 on Build GRCh38
Transcripts
NR_125362.1 NM_012263.4 NR_027779.1 XM_006724225.3 XM_011530108.2 XM_011530110.2 XM_017028751.1 XM_017028752.1 XM_017028753.1

Gene Symbol
TTLL1
Entrez Gene ID
25809
Gene Name
tubulin tyrosine ligase like 1
Gene Aliases
C22orf7, HS323M22B
UniGene
Hs.660298
Gene Chromosome Location
Chr.22: 43039516-43089428 on Build GRCh38
Transcripts
NR_125362.1 NM_012263.4 NR_027779.1 XM_006724225.3 XM_011530108.2 XM_011530110.2 XM_017028751.1 XM_017028752.1 XM_017028753.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5760361 c.1126G>A p.G376S Chr.22: 43046426-43046426 on Build GRCh38
286074 c.1134C>T p.Y378Y Chr.22: 43046418-43046418 on Build GRCh38
6446821 c.1106C>T p.S369L Chr.22: 43046446-43046446 on Build GRCh38
4104597 c.1109C>T p.P370L Chr.22: 43046443-43046443 on Build GRCh38
3729781 c.1109C>A p.P370Q Chr.22: 43046443-43046443 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760792235 Chr.22: 43046367-43046367 on Build GRCh38
rs767195098 Chr.22: 43046388-43046388 on Build GRCh38
rs768653912 Chr.22: 43046395-43046396 on Build GRCh38
rs750763846 Chr.22: 43046384-43046384 on Build GRCh38
rs372957466 Chr.22: 43046361-43046361 on Build GRCh38
rs925986977 Chr.22: 43046371-43046371 on Build GRCh38
rs199511297 Chr.22: 43046387-43046387 on Build GRCh38
rs1026949997 Chr.22: 43046286-43046286 on Build GRCh38
rs777155754 Chr.22: 43046446-43046446 on Build GRCh38
rs762195263 Chr.22: 43046378-43046378 on Build GRCh38
rs542202384 Chr.22: 43046391-43046391 on Build GRCh38
rs576221011 Chr.22: 43046322-43046322 on Build GRCh38
rs753789984 Chr.22: 43046403-43046403 on Build GRCh38
rs772302425 Chr.22: 43046360-43046360 on Build GRCh38
rs376132818 Chr.22: 43046368-43046368 on Build GRCh38
rs777860075 Chr.22: 43046402-43046402 on Build GRCh38
rs527949683 Chr.22: 43046445-43046445 on Build GRCh38
rs920267263 Chr.22: 43046315-43046315 on Build GRCh38
rs377552490 Chr.22: 43046433-43046433 on Build GRCh38
rs780261863 Chr.22: 43046439-43046439 on Build GRCh38
rs747075337 Chr.22: 43046443-43046443 on Build GRCh38
rs560814513 Chr.22: 43046430-43046430 on Build GRCh38
rs879537309 Chr.22: 43046325-43046325 on Build GRCh38
rs373363791 Chr.22: 43046427-43046427 on Build GRCh38
rs755593979 Chr.22: 43046401-43046401 on Build GRCh38
rs561672917 Chr.22: 43046379-43046379 on Build GRCh38
rs757143367 Chr.22: 43046426-43046426 on Build GRCh38
rs917150654 Chr.22: 43046418-43046418 on Build GRCh38
rs993778292 Chr.22: 43046280-43046280 on Build GRCh38
rs749239183 Chr.22: 43046393-43046394 on Build GRCh38
rs150617360 Chr.22: 43046370-43046370 on Build GRCh38
rs974462017 Chr.22: 43046275-43046275 on Build GRCh38

View Full Product Details