Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114930010-114930263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFSF8
Entrez Gene ID
944
Gene Name
tumor necrosis factor superfamily member 8
Gene Aliases
CD153, CD30L, CD30LG, TNLG3A
UniGene
Hs.494901
Gene Chromosome Location
Chr.9: 114893343-114930595 on Build GRCh38
Transcripts
NM_001244.3 NM_001252290.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
455229 c.195G>C p.T65T Chr.9: 114930109-114930109 on Build GRCh38
6321389 c.51C>A p.D17E Chr.9: 114930253-114930253 on Build GRCh38
6319801 c.126C>A p.T42T Chr.9: 114930178-114930178 on Build GRCh38
4433150 c.172A>G p.M58V Chr.9: 114930132-114930132 on Build GRCh38
5947331 c.50A>T p.D17V Chr.9: 114930254-114930254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374677956 Chr.9: 114930046-114930046 on Build GRCh38
rs753232916 Chr.9: 114930194-114930194 on Build GRCh38
rs201431210 Chr.9: 114930073-114930073 on Build GRCh38
rs758158682 Chr.9: 114930224-114930224 on Build GRCh38
rs766642549 Chr.9: 114930075-114930075 on Build GRCh38
rs944685660 Chr.9: 114930243-114930243 on Build GRCh38
rs939120206 Chr.9: 114930077-114930077 on Build GRCh38
rs3181358 Chr.9: 114930062-114930062 on Build GRCh38
rs538902986 Chr.9: 114930140-114930140 on Build GRCh38
rs200403281 Chr.9: 114930099-114930099 on Build GRCh38
rs770923739 Chr.9: 114930137-114930137 on Build GRCh38
rs745799964 Chr.9: 114930058-114930058 on Build GRCh38
rs771949979 Chr.9: 114930142-114930142 on Build GRCh38
rs141145398 Chr.9: 114930121-114930121 on Build GRCh38
rs768175916 Chr.9: 114930191-114930191 on Build GRCh38
rs992349846 Chr.9: 114930093-114930093 on Build GRCh38
rs746569681 Chr.9: 114930164-114930164 on Build GRCh38
rs776716866 Chr.9: 114930131-114930131 on Build GRCh38
rs3181357 Chr.9: 114930064-114930064 on Build GRCh38
rs760865978 Chr.9: 114930197-114930197 on Build GRCh38
rs577547653 Chr.9: 114930180-114930180 on Build GRCh38
rs752998769 Chr.9: 114930051-114930051 on Build GRCh38
rs575875443 Chr.9: 114930232-114930232 on Build GRCh38
rs767725717 Chr.9: 114930110-114930110 on Build GRCh38
rs902645635 Chr.9: 114930207-114930207 on Build GRCh38
rs775013111 Chr.9: 114930151-114930151 on Build GRCh38
rs150757350 Chr.9: 114930183-114930183 on Build GRCh38
rs139509296 Chr.9: 114930200-114930200 on Build GRCh38
rs774925441 Chr.9: 114930063-114930063 on Build GRCh38
rs377482345 Chr.9: 114930127-114930127 on Build GRCh38
rs995404589 Chr.9: 114930042-114930042 on Build GRCh38
rs750682898 Chr.9: 114930068-114930070 on Build GRCh38
rs760147722 Chr.9: 114930185-114930185 on Build GRCh38
rs780020377 Chr.9: 114930225-114930225 on Build GRCh38
rs746911830 Chr.9: 114930229-114930229 on Build GRCh38
rs1053938694 Chr.9: 114930215-114930215 on Build GRCh38
rs79494183 Chr.9: 114930177-114930177 on Build GRCh38
rs988038877 Chr.9: 114930052-114930052 on Build GRCh38
rs754094339 Chr.9: 114930203-114930203 on Build GRCh38
rs926324660 Chr.9: 114930076-114930076 on Build GRCh38
rs774804337 Chr.9: 114930184-114930184 on Build GRCh38
rs756209858 Chr.9: 114930102-114930103 on Build GRCh38
rs139251479 Chr.9: 114930175-114930176 on Build GRCh38
rs768032202 Chr.9: 114930168-114930168 on Build GRCh38
rs1039710395 Chr.9: 114930202-114930202 on Build GRCh38
rs371300725 Chr.9: 114930210-114930210 on Build GRCh38
rs200566415 Chr.9: 114930101-114930101 on Build GRCh38
rs757365804 Chr.9: 114930208-114930208 on Build GRCh38
rs780504470 Chr.9: 114930236-114930236 on Build GRCh38
rs778611604 Chr.9: 114930138-114930138 on Build GRCh38
rs1001085385 Chr.9: 114930212-114930212 on Build GRCh38
rs368943241 Chr.9: 114930209-114930209 on Build GRCh38
rs573372085 Chr.9: 114930118-114930118 on Build GRCh38

View Full Product Details