Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 47744968-47745241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX21
Entrez Gene ID
30009
Gene Name
T-box 21
Gene Aliases
T-PET, T-bet, TBET, TBLYM
UniGene
Hs.272409
Gene Chromosome Location
Chr.17: 47733240-47746122 on Build GRCh38
Transcripts
NM_013351.1 XM_011524698.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3518597 c.1394G>A p.G465E Chr.17: 47745152-47745152 on Build GRCh38
6797941 c.1451G>T p.R484L Chr.17: 47745209-47745209 on Build GRCh38
3518595 c.1345C>T p.R449C Chr.17: 47745103-47745103 on Build GRCh38
129628 c.1253C>T p.S418F Chr.17: 47745011-47745011 on Build GRCh38
3518596 c.1364C>T p.P455L Chr.17: 47745122-47745122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200396841 Chr.17: 47745092-47745092 on Build GRCh38
rs752687196 Chr.17: 47745113-47745113 on Build GRCh38
rs1021917662 Chr.17: 47745110-47745110 on Build GRCh38
rs575626754 Chr.17: 47745009-47745009 on Build GRCh38
rs1006587099 Chr.17: 47745132-47745132 on Build GRCh38
rs751380458 Chr.17: 47745131-47745131 on Build GRCh38
rs187953749 Chr.17: 47745114-47745114 on Build GRCh38
rs779042535 Chr.17: 47745064-47745064 on Build GRCh38
rs552669547 Chr.17: 47745133-47745133 on Build GRCh38
rs756229202 Chr.17: 47745031-47745031 on Build GRCh38
rs746994815 Chr.17: 47745199-47745200 on Build GRCh38
rs965073773 Chr.17: 47745137-47745137 on Build GRCh38
rs201980865 Chr.17: 47745071-47745071 on Build GRCh38
rs755818303 Chr.17: 47745222-47745222 on Build GRCh38
rs962901761 Chr.17: 47745080-47745080 on Build GRCh38
rs777841794 Chr.17: 47745124-47745124 on Build GRCh38
rs759076098 Chr.17: 47744994-47744994 on Build GRCh38
rs772631559 Chr.17: 47745178-47745178 on Build GRCh38
rs750645061 Chr.17: 47745030-47745030 on Build GRCh38
rs779985885 Chr.17: 47745150-47745155 on Build GRCh38
rs773622233 Chr.17: 47745103-47745103 on Build GRCh38
rs950443527 Chr.17: 47745018-47745018 on Build GRCh38
rs763123615 Chr.17: 47745208-47745208 on Build GRCh38
rs748779791 Chr.17: 47745175-47745175 on Build GRCh38
rs12721470 Chr.17: 47745213-47745213 on Build GRCh38
rs771278196 Chr.17: 47745187-47745187 on Build GRCh38
rs765422654 Chr.17: 47745200-47745200 on Build GRCh38
rs373895100 Chr.17: 47745089-47745089 on Build GRCh38
rs764170358 Chr.17: 47745209-47745209 on Build GRCh38
rs142939473 Chr.17: 47745010-47745010 on Build GRCh38
rs777645635 Chr.17: 47745072-47745072 on Build GRCh38
rs764578360 Chr.17: 47744997-47744997 on Build GRCh38
rs200867606 Chr.17: 47745090-47745090 on Build GRCh38
rs952548643 Chr.17: 47745192-47745192 on Build GRCh38
rs767879056 Chr.17: 47745100-47745100 on Build GRCh38
rs776749369 Chr.17: 47745193-47745193 on Build GRCh38
rs371923765 Chr.17: 47745046-47745046 on Build GRCh38
rs758355151 Chr.17: 47745117-47745117 on Build GRCh38
rs371795833 Chr.17: 47745197-47745197 on Build GRCh38
rs373817582 Chr.17: 47745055-47745055 on Build GRCh38
rs889442970 Chr.17: 47745129-47745129 on Build GRCh38
rs201143567 Chr.17: 47745106-47745106 on Build GRCh38
rs760993880 Chr.17: 47745179-47745179 on Build GRCh38
rs769294204 Chr.17: 47745158-47745158 on Build GRCh38
rs745479696 Chr.17: 47745155-47745155 on Build GRCh38
rs746690078 Chr.17: 47745077-47745077 on Build GRCh38
rs751496463 Chr.17: 47745211-47745211 on Build GRCh38
rs752883663 Chr.17: 47745205-47745205 on Build GRCh38
rs200345177 Chr.17: 47745104-47745104 on Build GRCh38
rs774866145 Chr.17: 47745091-47745091 on Build GRCh38
rs758300158 Chr.17: 47745068-47745068 on Build GRCh38
rs144197741 Chr.17: 47745040-47745040 on Build GRCh38
rs752147705 Chr.17: 47745154-47745154 on Build GRCh38
rs759557452 Chr.17: 47745109-47745109 on Build GRCh38
rs555375520 Chr.17: 47745002-47745002 on Build GRCh38
rs779478307 Chr.17: 47745159-47745159 on Build GRCh38
rs748050753 Chr.17: 47745066-47745066 on Build GRCh38
rs770550215 Chr.17: 47745082-47745082 on Build GRCh38
rs540412579 Chr.17: 47745112-47745112 on Build GRCh38
rs757029551 Chr.17: 47745212-47745212 on Build GRCh38

View Full Product Details