Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 104079306-104079555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCFC2
Entrez Gene ID
29915
Gene Name
host cell factor C2
Gene Aliases
HCF-2, HCF2
UniGene
Hs.506558
Gene Chromosome Location
Chr.12: 104064458-104108954 on Build GRCh38
Transcripts
NM_013320.2 XM_017019241.1 XM_017019242.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1209349 c.571G>T p.E191* Chr.12: 104079542-104079542 on Build GRCh38
1706181 c.565C>T p.P189S Chr.12: 104079536-104079536 on Build GRCh38
4622412 c.582A>G p.T194T Chr.12: 104079553-104079553 on Build GRCh38
5630488 c.552G>A p.G184G Chr.12: 104079523-104079523 on Build GRCh38
5896166 c.474-5C>T p.? Chr.12: 104079440-104079440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767865783 Chr.12: 104079441-104079441 on Build GRCh38
rs749095117 Chr.12: 104079338-104079338 on Build GRCh38
rs911575177 Chr.12: 104079352-104079352 on Build GRCh38
rs895987886 Chr.12: 104079373-104079373 on Build GRCh38
rs755850811 Chr.12: 104079414-104079414 on Build GRCh38
rs146213048 Chr.12: 104079529-104079529 on Build GRCh38
rs189667875 Chr.12: 104079333-104079333 on Build GRCh38
rs753445157 Chr.12: 104079480-104079480 on Build GRCh38
rs756604761 Chr.12: 104079475-104079475 on Build GRCh38
rs34011353 Chr.12: 104079508-104079509 on Build GRCh38
rs778014115 Chr.12: 104079431-104079431 on Build GRCh38
rs944930735 Chr.12: 104079361-104079361 on Build GRCh38
rs202081777 Chr.12: 104079479-104079479 on Build GRCh38
rs766631676 Chr.12: 104079407-104079407 on Build GRCh38
rs1023626383 Chr.12: 104079385-104079385 on Build GRCh38
rs985782586 Chr.12: 104079349-104079349 on Build GRCh38
rs745498802 Chr.12: 104079525-104079525 on Build GRCh38
rs753149730 Chr.12: 104079451-104079451 on Build GRCh38
rs758281586 Chr.12: 104079534-104079534 on Build GRCh38
rs773299562 Chr.12: 104079400-104079400 on Build GRCh38
rs751778852 Chr.12: 104079421-104079421 on Build GRCh38
rs756794025 Chr.12: 104079489-104079489 on Build GRCh38
rs763182590 Chr.12: 104079403-104079403 on Build GRCh38
rs113829238 Chr.12: 104079506-104079506 on Build GRCh38
rs759956458 Chr.12: 104079433-104079433 on Build GRCh38
rs1014493961 Chr.12: 104079376-104079376 on Build GRCh38

View Full Product Details