Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56319037-56319286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAN2
Entrez Gene ID
9924
Gene Name
PAN2 poly(A) specific ribonuclease subunit
Gene Aliases
USP52
UniGene
Hs.273397
Gene Chromosome Location
Chr.12: 56316223-56334053 on Build GRCh38
Transcripts
NM_001127460.2 NM_001166279.1 NM_014871.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6462742 c.3357C>A p.Y1119* Chr.12: 56319095-56319095 on Build GRCh38
7249900 c.3277A>C p.K1093Q Chr.12: 56319175-56319175 on Build GRCh38
6737179 c.3320G>A p.R1107Q Chr.12: 56319132-56319132 on Build GRCh38
941631 c.3296C>T p.T1099I Chr.12: 56319156-56319156 on Build GRCh38
5753973 c.3341G>A p.R1114Q Chr.12: 56319111-56319111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985138362 Chr.12: 56319237-56319237 on Build GRCh38
rs746705011 Chr.12: 56319142-56319142 on Build GRCh38
rs764512601 Chr.12: 56319150-56319150 on Build GRCh38
rs368479834 Chr.12: 56319224-56319224 on Build GRCh38
rs756450889 Chr.12: 56319226-56319226 on Build GRCh38
rs753871107 Chr.12: 56319260-56319260 on Build GRCh38
rs757765689 Chr.12: 56319182-56319183 on Build GRCh38
rs767949324 Chr.12: 56319211-56319211 on Build GRCh38
rs780614559 Chr.12: 56319266-56319273 on Build GRCh38
rs925609685 Chr.12: 56319257-56319257 on Build GRCh38
rs754940108 Chr.12: 56319070-56319070 on Build GRCh38
rs779503477 Chr.12: 56319193-56319194 on Build GRCh38
rs771809067 Chr.12: 56319127-56319127 on Build GRCh38
rs749363067 Chr.12: 56319066-56319066 on Build GRCh38
rs375649878 Chr.12: 56319133-56319133 on Build GRCh38
rs777873996 Chr.12: 56319179-56319180 on Build GRCh38
rs903401894 Chr.12: 56319065-56319065 on Build GRCh38
rs747912929 Chr.12: 56319112-56319112 on Build GRCh38
rs766524798 Chr.12: 56319196-56319197 on Build GRCh38
rs774862616 Chr.12: 56319185-56319185 on Build GRCh38
rs772524256 Chr.12: 56319264-56319265 on Build GRCh38
rs149050168 Chr.12: 56319199-56319199 on Build GRCh38
rs371114875 Chr.12: 56319095-56319095 on Build GRCh38
rs759080302 Chr.12: 56319146-56319146 on Build GRCh38
rs777169068 Chr.12: 56319201-56319201 on Build GRCh38
rs377276536 Chr.12: 56319072-56319072 on Build GRCh38
rs766622495 Chr.12: 56319231-56319231 on Build GRCh38
rs753930384 Chr.12: 56319180-56319181 on Build GRCh38
rs776065284 Chr.12: 56319143-56319143 on Build GRCh38

View Full Product Details