Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102168515-102168788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1R1
Entrez Gene ID
3554
Gene Name
interleukin 1 receptor type 1
Gene Aliases
CD121A, D2S1473, IL-1R-alpha, IL1R, IL1RA, P80
UniGene
Hs.701982
Gene Chromosome Location
Chr.2: 102069638-102179874 on Build GRCh38
Transcripts
NM_000877.3 NM_001288706.1 NM_001320978.1 NM_001320980.1 NM_001320981.1 NM_001320982.1 NM_001320983.1 NM_001320984.1 NM_001320985.1 XM_005263930.4 XM_005263934.4 XM_011511115.2 XM_011511116.1 XM_011511117.1 XM_011511118.2 XM_017003989.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3564417 c.658G>A p.E220K Chr.2: 102168600-102168600 on Build GRCh38
340610 c.679G>T p.V227L Chr.2: 102168621-102168621 on Build GRCh38
3971462 c.680T>C p.V227A Chr.2: 102168622-102168622 on Build GRCh38
4804534 c.687G>T p.V229V Chr.2: 102168629-102168629 on Build GRCh38
4969499 c.701A>G p.E234G Chr.2: 102168643-102168643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150045652 Chr.2: 102168632-102168632 on Build GRCh38
rs376447148 Chr.2: 102168609-102168609 on Build GRCh38
rs371552451 Chr.2: 102168698-102168698 on Build GRCh38
rs375925077 Chr.2: 102168749-102168749 on Build GRCh38
rs546822534 Chr.2: 102168699-102168699 on Build GRCh38
rs397897631 Chr.2: 102168710-102168711 on Build GRCh38
rs1052625989 Chr.2: 102168716-102168716 on Build GRCh38
rs369654655 Chr.2: 102168697-102168697 on Build GRCh38
rs762247866 Chr.2: 102168559-102168559 on Build GRCh38
rs201939068 Chr.2: 102168555-102168555 on Build GRCh38
rs1011114427 Chr.2: 102168754-102168754 on Build GRCh38
rs35268097 Chr.2: 102168699-102168699 on Build GRCh38
rs780926737 Chr.2: 102168684-102168684 on Build GRCh38
rs199768199 Chr.2: 102168696-102168696 on Build GRCh38
rs375835023 Chr.2: 102168727-102168727 on Build GRCh38
rs201863057 Chr.2: 102168576-102168576 on Build GRCh38
rs199921919 Chr.2: 102168565-102168565 on Build GRCh38
rs3213736 Chr.2: 102168682-102168682 on Build GRCh38
rs756976874 Chr.2: 102168563-102168563 on Build GRCh38
rs147536577 Chr.2: 102168604-102168604 on Build GRCh38
rs201216211 Chr.2: 102168711-102168711 on Build GRCh38
rs764163507 Chr.2: 102168667-102168667 on Build GRCh38
rs865802708 Chr.2: 102168575-102168575 on Build GRCh38
rs565713120 Chr.2: 102168756-102168756 on Build GRCh38
rs55639750 Chr.2: 102168698-102168699 on Build GRCh38
rs201015375 Chr.2: 102168573-102168573 on Build GRCh38
rs970815613 Chr.2: 102168586-102168586 on Build GRCh38
rs777164994 Chr.2: 102168629-102168629 on Build GRCh38
rs956193502 Chr.2: 102168569-102168569 on Build GRCh38
rs796707076 Chr.2: 102168698-102168699 on Build GRCh38
rs758499965 Chr.2: 102168662-102168662 on Build GRCh38
rs748809764 Chr.2: 102168577-102168577 on Build GRCh38
rs202198085 Chr.2: 102168697-102168697 on Build GRCh38
rs201931236 Chr.2: 102168658-102168658 on Build GRCh38
rs397872908 Chr.2: 102168710-102168710 on Build GRCh38
rs201809361 Chr.2: 102168680-102168680 on Build GRCh38
rs751438954 Chr.2: 102168668-102168668 on Build GRCh38
rs752916518 Chr.2: 102168659-102168659 on Build GRCh38
rs200016174 Chr.2: 102168583-102168583 on Build GRCh38
rs3213735 Chr.2: 102168698-102168698 on Build GRCh38
rs752790270 Chr.2: 102168551-102168551 on Build GRCh38
rs201547297 Chr.2: 102168710-102168710 on Build GRCh38
rs764844643 Chr.2: 102168687-102168687 on Build GRCh38
rs867765901 Chr.2: 102168616-102168616 on Build GRCh38
rs768343748 Chr.2: 102168581-102168581 on Build GRCh38
rs548049991 Chr.2: 102168699-102168700 on Build GRCh38
rs753719795 Chr.2: 102168697-102168698 on Build GRCh38
rs373423043 Chr.2: 102168582-102168582 on Build GRCh38
rs796882918 Chr.2: 102168699-102168700 on Build GRCh38

View Full Product Details