Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36864906-36865157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCF4
Entrez Gene ID
4689
Gene Name
neutrophil cytosolic factor 4
Gene Aliases
CGD3, NCF, P40PHOX, SH3PXD4
UniGene
Hs.474781
Gene Chromosome Location
Chr.22: 36860988-36878017 on Build GRCh38
Transcripts
NM_000631.4 NM_013416.3 XM_017028808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4164933 c.179G>T p.R60L Chr.22: 36864980-36864980 on Build GRCh38
1665955 c.118-1G>T p.? Chr.22: 36864918-36864918 on Build GRCh38
4103851 c.150A>G p.G50G Chr.22: 36864951-36864951 on Build GRCh38
4164934 c.179G>T p.R60L Chr.22: 36864980-36864980 on Build GRCh38
6722894 c.179G>A p.R60H Chr.22: 36864980-36864980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767200492 Chr.22: 36864970-36864970 on Build GRCh38
rs751228394 Chr.22: 36865113-36865113 on Build GRCh38
rs750577331 Chr.22: 36865015-36865015 on Build GRCh38
rs144929792 Chr.22: 36864968-36864969 on Build GRCh38
rs370754874 Chr.22: 36865071-36865071 on Build GRCh38
rs780484949 Chr.22: 36865043-36865043 on Build GRCh38
rs776616848 Chr.22: 36865082-36865082 on Build GRCh38
rs373041130 Chr.22: 36865020-36865020 on Build GRCh38
rs748376366 Chr.22: 36865068-36865068 on Build GRCh38
rs759457602 Chr.22: 36865088-36865088 on Build GRCh38
rs112306225 Chr.22: 36865055-36865055 on Build GRCh38
rs201881905 Chr.22: 36865079-36865079 on Build GRCh38
rs148715332 Chr.22: 36864946-36864946 on Build GRCh38
rs183179978 Chr.22: 36864979-36864979 on Build GRCh38
rs869025585 Chr.22: 36864953-36864954 on Build GRCh38
rs200052796 Chr.22: 36865078-36865078 on Build GRCh38
rs750743658 Chr.22: 36865002-36865002 on Build GRCh38
rs774613957 Chr.22: 36865059-36865059 on Build GRCh38
rs35431748 Chr.22: 36865041-36865041 on Build GRCh38
rs767737419 Chr.22: 36865106-36865107 on Build GRCh38
rs753635134 Chr.22: 36864930-36864930 on Build GRCh38
rs763314327 Chr.22: 36864974-36864974 on Build GRCh38
rs932330052 Chr.22: 36864965-36864965 on Build GRCh38
rs746353194 Chr.22: 36864971-36864971 on Build GRCh38
rs188209920 Chr.22: 36865029-36865029 on Build GRCh38
rs377510654 Chr.22: 36865023-36865023 on Build GRCh38
rs767668921 Chr.22: 36864989-36864989 on Build GRCh38
rs773596460 Chr.22: 36865072-36865072 on Build GRCh38
rs749822456 Chr.22: 36865054-36865054 on Build GRCh38
rs749761418 Chr.22: 36865124-36865124 on Build GRCh38
rs766377093 Chr.22: 36865021-36865021 on Build GRCh38
rs143532979 Chr.22: 36864973-36864973 on Build GRCh38
rs777426455 Chr.22: 36865030-36865030 on Build GRCh38
rs780637356 Chr.22: 36865122-36865122 on Build GRCh38
rs771597330 Chr.22: 36864963-36864963 on Build GRCh38
rs747769633 Chr.22: 36864958-36864958 on Build GRCh38
rs756750092 Chr.22: 36865118-36865118 on Build GRCh38
rs371236295 Chr.22: 36865089-36865089 on Build GRCh38
rs876657377 Chr.22: 36864944-36864953 on Build GRCh38
rs754759226 Chr.22: 36864933-36864933 on Build GRCh38
rs908040747 Chr.22: 36865076-36865076 on Build GRCh38
rs373447489 Chr.22: 36865039-36865039 on Build GRCh38
rs770239095 Chr.22: 36864972-36864972 on Build GRCh38
rs192221353 Chr.22: 36865133-36865133 on Build GRCh38
rs369847561 Chr.22: 36864980-36864980 on Build GRCh38
rs374147489 Chr.22: 36865094-36865094 on Build GRCh38
rs778630595 Chr.22: 36865026-36865026 on Build GRCh38
rs752387068 Chr.22: 36865092-36865092 on Build GRCh38
rs763670860 Chr.22: 36865109-36865109 on Build GRCh38
rs900312893 Chr.22: 36864931-36864931 on Build GRCh38
rs762003847 Chr.22: 36864981-36864981 on Build GRCh38

View Full Product Details