Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30604438-30604675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF689
Entrez Gene ID
115509
Gene Name
zinc finger protein 689
Gene Aliases
TIPUH1
UniGene
Hs.454685
Gene Chromosome Location
Chr.16: 30602558-30610775 on Build GRCh38
Transcripts
NM_138447.2 NR_073480.1 NR_073481.1 NR_073482.1 NR_104166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
969885 c.1184A>G p.H395R Chr.16: 30604583-30604583 on Build GRCh38
969886 c.1105G>A p.G369R Chr.16: 30604662-30604662 on Build GRCh38
6274582 c.1176C>A p.R392R Chr.16: 30604591-30604591 on Build GRCh38
3112511 c.1104C>T p.T368T Chr.16: 30604663-30604663 on Build GRCh38
1377433 c.1175G>A p.R392H Chr.16: 30604592-30604592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868699076 Chr.16: 30604548-30604548 on Build GRCh38
rs374782420 Chr.16: 30604492-30604492 on Build GRCh38
rs1035174066 Chr.16: 30604473-30604473 on Build GRCh38
rs114724275 Chr.16: 30604470-30604470 on Build GRCh38
rs373446010 Chr.16: 30604530-30604530 on Build GRCh38
rs754483862 Chr.16: 30604578-30604578 on Build GRCh38
rs368841160 Chr.16: 30604503-30604503 on Build GRCh38
rs1010686788 Chr.16: 30604544-30604544 on Build GRCh38
rs765048971 Chr.16: 30604542-30604542 on Build GRCh38
rs753630174 Chr.16: 30604457-30604457 on Build GRCh38
rs774797385 Chr.16: 30604593-30604593 on Build GRCh38
rs142008271 Chr.16: 30604589-30604589 on Build GRCh38
rs745813021 Chr.16: 30604460-30604460 on Build GRCh38
rs760059619 Chr.16: 30604512-30604512 on Build GRCh38
rs778773599 Chr.16: 30604458-30604458 on Build GRCh38
rs59446493 Chr.16: 30604528-30604528 on Build GRCh38
rs779691783 Chr.16: 30604471-30604471 on Build GRCh38
rs138494235 Chr.16: 30604585-30604585 on Build GRCh38
rs772568230 Chr.16: 30604602-30604602 on Build GRCh38
rs758385353 Chr.16: 30604555-30604555 on Build GRCh38
rs145758365 Chr.16: 30604508-30604508 on Build GRCh38
rs746176569 Chr.16: 30604595-30604595 on Build GRCh38
rs775575780 Chr.16: 30604529-30604529 on Build GRCh38
rs746523884 Chr.16: 30604571-30604571 on Build GRCh38
rs750378070 Chr.16: 30604550-30604550 on Build GRCh38
rs931749221 Chr.16: 30604658-30604658 on Build GRCh38
rs749631841 Chr.16: 30604505-30604505 on Build GRCh38
rs775915074 Chr.16: 30604619-30604619 on Build GRCh38
rs754078044 Chr.16: 30604533-30604533 on Build GRCh38
rs746470782 Chr.16: 30604478-30604478 on Build GRCh38
rs753704392 Chr.16: 30604482-30604496 on Build GRCh38
rs907805125 Chr.16: 30604557-30604557 on Build GRCh38
rs757064638 Chr.16: 30604538-30604538 on Build GRCh38
rs757003261 Chr.16: 30604559-30604570 on Build GRCh38
rs115586197 Chr.16: 30604651-30604651 on Build GRCh38
rs772040607 Chr.16: 30604564-30604564 on Build GRCh38
rs780261613 Chr.16: 30604573-30604575 on Build GRCh38
rs764310676 Chr.16: 30604531-30604531 on Build GRCh38
rs35784211 Chr.16: 30604508-30604509 on Build GRCh38
rs771521007 Chr.16: 30604463-30604463 on Build GRCh38

View Full Product Details