Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 51698783-51699054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSTF2T
Entrez Gene ID
23283
Gene Name
cleavage stimulation factor subunit 2, tau variant
Gene Aliases
CstF-64T
UniGene
Hs.591358
Gene Chromosome Location
Chr.10: 51695486-51699595 on Build GRCh38
Transcripts
NM_015235.2 NM_001098512.2 NM_006258.3 XM_011539952.2 XM_017016412.1 XM_017016413.1

Gene Symbol
PRKG1
Entrez Gene ID
5592
Gene Name
protein kinase, cGMP-dependent, type I
Gene Aliases
AAT8, PKG, PRKG1B, PRKGR1B, cGK, cGK 1, cGK1, cGKI, cGKI-BETA, cGKI-alpha
UniGene
Hs.407535
Gene Chromosome Location
Chr.10: 50991151-52298350 on Build GRCh38
Transcripts
NM_015235.2 NM_001098512.2 NM_006258.3 XM_011539952.2 XM_017016412.1 XM_017016413.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6637050 c.513_514delAC p.L172fs*54 Chr.10: 51699036-51699037 on Build GRCh38
4428477 c.621T>C p.S207S Chr.10: 51698929-51698929 on Build GRCh38
7363115 c.682C>A p.L228M Chr.10: 51698868-51698868 on Build GRCh38
4602681 c.543C>G p.I181M Chr.10: 51699007-51699007 on Build GRCh38
4552238 c.553G>A p.E185K Chr.10: 51698997-51698997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146358831 Chr.10: 51698938-51698938 on Build GRCh38
rs1056166157 Chr.10: 51698966-51698966 on Build GRCh38
rs770253904 Chr.10: 51698875-51698875 on Build GRCh38
rs779091680 Chr.10: 51698984-51698984 on Build GRCh38
rs573426485 Chr.10: 51698829-51698829 on Build GRCh38
rs750365324 Chr.10: 51698909-51698909 on Build GRCh38
rs575311852 Chr.10: 51698890-51698891 on Build GRCh38
rs765848711 Chr.10: 51698933-51698933 on Build GRCh38
rs746610656 Chr.10: 51698847-51698847 on Build GRCh38
rs773738151 Chr.10: 51698877-51698877 on Build GRCh38
rs938994632 Chr.10: 51698957-51698957 on Build GRCh38
rs138223207 Chr.10: 51698840-51698840 on Build GRCh38
rs773032001 Chr.10: 51699025-51699025 on Build GRCh38
rs368413928 Chr.10: 51698890-51698890 on Build GRCh38
rs774625713 Chr.10: 51698819-51698819 on Build GRCh38
rs1053413546 Chr.10: 51698952-51698952 on Build GRCh38
rs762376482 Chr.10: 51698891-51698896 on Build GRCh38
rs369397658 Chr.10: 51698963-51698963 on Build GRCh38
rs769578681 Chr.10: 51699007-51699007 on Build GRCh38
rs749945139 Chr.10: 51698838-51698838 on Build GRCh38
rs781498470 Chr.10: 51698857-51698857 on Build GRCh38
rs1022157636 Chr.10: 51698813-51698813 on Build GRCh38
rs761673073 Chr.10: 51698901-51698901 on Build GRCh38
rs559685615 Chr.10: 51698814-51698814 on Build GRCh38
rs571708863 Chr.10: 51698828-51698830 on Build GRCh38
rs144891921 Chr.10: 51698904-51698904 on Build GRCh38
rs768186402 Chr.10: 51698897-51698898 on Build GRCh38
rs903663317 Chr.10: 51699034-51699034 on Build GRCh38
rs150439329 Chr.10: 51698835-51698835 on Build GRCh38
rs1043880061 Chr.10: 51699015-51699015 on Build GRCh38
rs141640089 Chr.10: 51698821-51698821 on Build GRCh38
rs547119353 Chr.10: 51698973-51698973 on Build GRCh38
rs112627642 Chr.10: 51698895-51698895 on Build GRCh38
rs142284264 Chr.10: 51698932-51698932 on Build GRCh38
rs372468137 Chr.10: 51698896-51698896 on Build GRCh38
rs779633285 Chr.10: 51698846-51698846 on Build GRCh38
rs202077988 Chr.10: 51698849-51698849 on Build GRCh38
rs772469448 Chr.10: 51698989-51698989 on Build GRCh38
rs754734534 Chr.10: 51698897-51698908 on Build GRCh38
rs780801967 Chr.10: 51698945-51698945 on Build GRCh38
rs770916281 Chr.10: 51698979-51698979 on Build GRCh38
rs775853473 Chr.10: 51698991-51698991 on Build GRCh38
rs200221098 Chr.10: 51699013-51699013 on Build GRCh38
rs761574288 Chr.10: 51698994-51698994 on Build GRCh38
rs760675594 Chr.10: 51698913-51698914 on Build GRCh38
rs749394479 Chr.10: 51698878-51698878 on Build GRCh38
rs866466836 Chr.10: 51698961-51698961 on Build GRCh38
rs764007054 Chr.10: 51698899-51698899 on Build GRCh38
rs763921013 Chr.10: 51698823-51698823 on Build GRCh38
rs947852929 Chr.10: 51698998-51698998 on Build GRCh38
rs143249904 Chr.10: 51698886-51698886 on Build GRCh38
rs753099523 Chr.10: 51698950-51698950 on Build GRCh38
rs575649480 Chr.10: 51698891-51698902 on Build GRCh38
rs746112702 Chr.10: 51698985-51698985 on Build GRCh38
rs561326555 Chr.10: 51698913-51698913 on Build GRCh38
rs749744522 Chr.10: 51698975-51698975 on Build GRCh38
rs149125442 Chr.10: 51698868-51698868 on Build GRCh38
rs926658890 Chr.10: 51698852-51698852 on Build GRCh38
rs750527976 Chr.10: 51698904-51698921 on Build GRCh38
rs1030885581 Chr.10: 51698848-51698848 on Build GRCh38

View Full Product Details