Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 184631702-184631941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP3
Entrez Gene ID
836
Gene Name
caspase 3
Gene Aliases
CPP32, CPP32B, SCA-1
UniGene
Hs.141125
Gene Chromosome Location
Chr.4: 184627696-184649475 on Build GRCh38
Transcripts
NM_004346.3 NM_032991.2 XM_011532301.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6166914 c.343G>T p.V115F Chr.4: 184631905-184631905 on Build GRCh38
1053703 c.340T>G p.F114V Chr.4: 184631908-184631908 on Build GRCh38
3130250 c.415delA p.I139fs*1 Chr.4: 184631833-184631833 on Build GRCh38
4155064 c.308-1G>T p.? Chr.4: 184631941-184631941 on Build GRCh38
6099806 c.482A>T p.Q161L Chr.4: 184631766-184631766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767803653 Chr.4: 184631864-184631864 on Build GRCh38
rs577739464 Chr.4: 184631808-184631808 on Build GRCh38
rs778601566 Chr.4: 184631781-184631781 on Build GRCh38
rs766435580 Chr.4: 184631903-184631903 on Build GRCh38
rs141835512 Chr.4: 184631855-184631855 on Build GRCh38
rs201786370 Chr.4: 184631733-184631733 on Build GRCh38
rs748875178 Chr.4: 184631757-184631757 on Build GRCh38
rs1026750296 Chr.4: 184631776-184631776 on Build GRCh38
rs768687549 Chr.4: 184631827-184631827 on Build GRCh38
rs755453981 Chr.4: 184631740-184631740 on Build GRCh38
rs754299277 Chr.4: 184631896-184631896 on Build GRCh38
rs776400130 Chr.4: 184631839-184631839 on Build GRCh38
rs756670671 Chr.4: 184631754-184631754 on Build GRCh38
rs778089076 Chr.4: 184631755-184631755 on Build GRCh38
rs766551582 Chr.4: 184631843-184631843 on Build GRCh38
rs112883179 Chr.4: 184631798-184631798 on Build GRCh38
rs771653561 Chr.4: 184631797-184631797 on Build GRCh38
rs764705313 Chr.4: 184631879-184631879 on Build GRCh38
rs778409347 Chr.4: 184631902-184631902 on Build GRCh38
rs774511939 Chr.4: 184631849-184631849 on Build GRCh38
rs745505513 Chr.4: 184631793-184631793 on Build GRCh38
rs756415159 Chr.4: 184631870-184631870 on Build GRCh38
rs758078619 Chr.4: 184631913-184631913 on Build GRCh38
rs751043644 Chr.4: 184631745-184631746 on Build GRCh38
rs753131848 Chr.4: 184631869-184631869 on Build GRCh38
rs753048830 Chr.4: 184631753-184631753 on Build GRCh38
rs574650765 Chr.4: 184631830-184631830 on Build GRCh38
rs372826580 Chr.4: 184631760-184631760 on Build GRCh38
rs763157710 Chr.4: 184631841-184631841 on Build GRCh38
rs200935460 Chr.4: 184631851-184631851 on Build GRCh38
rs775267475 Chr.4: 184631807-184631807 on Build GRCh38
rs368412684 Chr.4: 184631746-184631746 on Build GRCh38
rs78678473 Chr.4: 184631898-184631898 on Build GRCh38
rs367650105 Chr.4: 184631900-184631900 on Build GRCh38
rs371766825 Chr.4: 184631905-184631905 on Build GRCh38
rs766749887 Chr.4: 184631732-184631732 on Build GRCh38
rs1025496516 Chr.4: 184631825-184631825 on Build GRCh38

View Full Product Details