Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124943108-124943367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZRANB1
Entrez Gene ID
54764
Gene Name
zinc finger RANBP2-type containing 1
Gene Aliases
TRABID
UniGene
Hs.595158
Gene Chromosome Location
Chr.10: 124916896-124988059 on Build GRCh38
Transcripts
NM_017580.2 XM_005269925.4 XM_005269926.4 XM_006717907.2 XM_017016357.1 XM_017016358.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6551428 c.620G>T p.C207F Chr.10: 124943113-124943113 on Build GRCh38
4144422 c.723C>A p.S241R Chr.10: 124943216-124943216 on Build GRCh38
1603114 c.762A>C p.Q254H Chr.10: 124943255-124943255 on Build GRCh38
235610 c.702A>G p.E234E Chr.10: 124943195-124943195 on Build GRCh38
6128761 c.716T>C p.V239A Chr.10: 124943209-124943209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746213739 Chr.10: 124943325-124943325 on Build GRCh38
rs747462925 Chr.10: 124943164-124943164 on Build GRCh38
rs757966756 Chr.10: 124943282-124943282 on Build GRCh38
rs372066172 Chr.10: 124943157-124943157 on Build GRCh38
rs1015619319 Chr.10: 124943140-124943140 on Build GRCh38
rs763675623 Chr.10: 124943327-124943328 on Build GRCh38
rs766122860 Chr.10: 124943228-124943228 on Build GRCh38
rs971164575 Chr.10: 124943158-124943158 on Build GRCh38
rs764949087 Chr.10: 124943267-124943267 on Build GRCh38
rs376277342 Chr.10: 124943287-124943287 on Build GRCh38
rs747474149 Chr.10: 124943295-124943295 on Build GRCh38
rs561809695 Chr.10: 124943317-124943317 on Build GRCh38
rs754615207 Chr.10: 124943151-124943151 on Build GRCh38
rs753441884 Chr.10: 124943147-124943147 on Build GRCh38
rs752319347 Chr.10: 124943272-124943272 on Build GRCh38
rs779843987 Chr.10: 124943135-124943135 on Build GRCh38
rs747636143 Chr.10: 124943153-124943153 on Build GRCh38
rs369131916 Chr.10: 124943326-124943326 on Build GRCh38
rs1025793 Chr.10: 124943335-124943335 on Build GRCh38
rs137877676 Chr.10: 124943195-124943195 on Build GRCh38
rs778418978 Chr.10: 124943162-124943162 on Build GRCh38
rs775588324 Chr.10: 124943205-124943205 on Build GRCh38
rs778460900 Chr.10: 124943336-124943336 on Build GRCh38
rs753615353 Chr.10: 124943243-124943243 on Build GRCh38
rs778584834 Chr.10: 124943152-124943152 on Build GRCh38
rs754723039 Chr.10: 124943246-124943246 on Build GRCh38
rs775678140 Chr.10: 124943330-124943330 on Build GRCh38
rs771212604 Chr.10: 124943179-124943179 on Build GRCh38
rs754458341 Chr.10: 124943319-124943319 on Build GRCh38
rs760597158 Chr.10: 124943215-124943215 on Build GRCh38
rs776987689 Chr.10: 124943190-124943190 on Build GRCh38
rs767504468 Chr.10: 124943213-124943213 on Build GRCh38
rs987431656 Chr.10: 124943302-124943302 on Build GRCh38
rs143761266 Chr.10: 124943208-124943208 on Build GRCh38
rs150163191 Chr.10: 124943296-124943296 on Build GRCh38
rs769982917 Chr.10: 124943204-124943204 on Build GRCh38

View Full Product Details