Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47660142-47660415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL2
Entrez Gene ID
79841
Gene Name
ATP/GTP binding protein like 2
Gene Aliases
CCP2
UniGene
Hs.147377
Gene Chromosome Location
Chr.11: 47659168-47715397 on Build GRCh38
Transcripts
NM_024783.3 XM_005253138.4 XM_005253139.4 XM_005253140.4 XM_006718327.3 XM_006718328.3 XM_011520372.2 XM_011520373.2 XM_011520374.2 XM_011520375.2 XM_011520376.2 XM_011520377.2 XM_011520378.2 XM_011520379.2 XM_011520381.2 XM_017018332.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6449070 c.2624G>T p.W875L Chr.11: 47660258-47660258 on Build GRCh38
5677057 c.2634C>A p.S878R Chr.11: 47660248-47660248 on Build GRCh38
5766588 c.2706G>A p.P902P Chr.11: 47660176-47660176 on Build GRCh38
6132678 c.2701T>C p.Y901H Chr.11: 47660181-47660181 on Build GRCh38
5838672 c.2605C>G p.P869A Chr.11: 47660277-47660277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745634422 Chr.11: 47660314-47660314 on Build GRCh38
rs780959717 Chr.11: 47660274-47660274 on Build GRCh38
rs753962466 Chr.11: 47660240-47660240 on Build GRCh38
rs766792595 Chr.11: 47660258-47660258 on Build GRCh38
rs775706835 Chr.11: 47660214-47660214 on Build GRCh38
rs776818105 Chr.11: 47660327-47660327 on Build GRCh38
rs865906550 Chr.11: 47660320-47660320 on Build GRCh38
rs868535240 Chr.11: 47660208-47660208 on Build GRCh38
rs748126466 Chr.11: 47660204-47660204 on Build GRCh38
rs749800490 Chr.11: 47660292-47660300 on Build GRCh38
rs778124129 Chr.11: 47660333-47660333 on Build GRCh38
rs952770742 Chr.11: 47660272-47660272 on Build GRCh38
rs754635374 Chr.11: 47660260-47660260 on Build GRCh38
rs185902167 Chr.11: 47660221-47660221 on Build GRCh38
rs764103792 Chr.11: 47660172-47660172 on Build GRCh38
rs765718446 Chr.11: 47660246-47660246 on Build GRCh38
rs375959180 Chr.11: 47660298-47660298 on Build GRCh38
rs750799658 Chr.11: 47660179-47660179 on Build GRCh38
rs761927827 Chr.11: 47660242-47660242 on Build GRCh38
rs368960012 Chr.11: 47660352-47660352 on Build GRCh38
rs948482047 Chr.11: 47660237-47660237 on Build GRCh38
rs774096011 Chr.11: 47660300-47660300 on Build GRCh38
rs747721104 Chr.11: 47660284-47660284 on Build GRCh38
rs576819206 Chr.11: 47660346-47660346 on Build GRCh38
rs777577079 Chr.11: 47660292-47660292 on Build GRCh38
rs34136118 Chr.11: 47660326-47660326 on Build GRCh38
rs1017930193 Chr.11: 47660223-47660223 on Build GRCh38
rs758774012 Chr.11: 47660190-47660190 on Build GRCh38
rs758690843 Chr.11: 47660254-47660254 on Build GRCh38
rs752010253 Chr.11: 47660363-47660363 on Build GRCh38
rs145812541 Chr.11: 47660299-47660299 on Build GRCh38
rs138759912 Chr.11: 47660210-47660210 on Build GRCh38
rs765290186 Chr.11: 47660337-47660337 on Build GRCh38
rs755370035 Chr.11: 47660367-47660367 on Build GRCh38
rs768294586 Chr.11: 47660200-47660200 on Build GRCh38
rs750934072 Chr.11: 47660252-47660252 on Build GRCh38
rs537747035 Chr.11: 47660194-47660194 on Build GRCh38
rs760752058 Chr.11: 47660163-47660163 on Build GRCh38
rs372291326 Chr.11: 47660173-47660173 on Build GRCh38
rs760841776 Chr.11: 47660227-47660227 on Build GRCh38
rs908649892 Chr.11: 47660321-47660321 on Build GRCh38
rs150644125 Chr.11: 47660203-47660203 on Build GRCh38
rs143937205 Chr.11: 47660176-47660176 on Build GRCh38
rs769657451 Chr.11: 47660209-47660209 on Build GRCh38
rs779446769 Chr.11: 47660177-47660177 on Build GRCh38
rs751278901 Chr.11: 47660259-47660259 on Build GRCh38
rs755646959 Chr.11: 47660286-47660286 on Build GRCh38
rs764192610 Chr.11: 47660239-47660239 on Build GRCh38
rs766879939 Chr.11: 47660355-47660355 on Build GRCh38
rs553856051 Chr.11: 47660344-47660344 on Build GRCh38
rs780773705 Chr.11: 47660198-47660199 on Build GRCh38
rs916147451 Chr.11: 47660379-47660379 on Build GRCh38

View Full Product Details