Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 6863383-6863656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMPK2
Entrez Gene ID
129607
Gene Name
cytidine/uridine monophosphate kinase 2
Gene Aliases
NDK, TMPK2, TYKi, UMP-CMPK2
UniGene
Hs.7155
Gene Chromosome Location
Chr.2: 6840417-6866635 on Build GRCh38
Transcripts
NM_001256477.1 NM_001256478.1 NM_207315.3 NR_046236.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1022452 c.692C>A p.P231H Chr.2: 6863562-6863562 on Build GRCh38
7348746 c.721G>A p.D241N Chr.2: 6863533-6863533 on Build GRCh38
7272856 c.788C>T p.T263M Chr.2: 6863466-6863466 on Build GRCh38
5677811 c.692C>T p.P231L Chr.2: 6863562-6863562 on Build GRCh38
5452758 c.700C>T p.R234W Chr.2: 6863554-6863554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762163307 Chr.2: 6863534-6863534 on Build GRCh38
rs997610888 Chr.2: 6863452-6863452 on Build GRCh38
rs765669392 Chr.2: 6863520-6863520 on Build GRCh38
rs773169808 Chr.2: 6863525-6863544 on Build GRCh38
rs371005529 Chr.2: 6863483-6863483 on Build GRCh38
rs377336914 Chr.2: 6863453-6863453 on Build GRCh38
rs774940126 Chr.2: 6863503-6863503 on Build GRCh38
rs367633170 Chr.2: 6863599-6863599 on Build GRCh38
rs755229087 Chr.2: 6863430-6863430 on Build GRCh38
rs771017102 Chr.2: 6863442-6863442 on Build GRCh38
rs201102085 Chr.2: 6863437-6863437 on Build GRCh38
rs768316626 Chr.2: 6863588-6863588 on Build GRCh38
rs765732132 Chr.2: 6863414-6863414 on Build GRCh38
rs781517675 Chr.2: 6863536-6863536 on Build GRCh38
rs763701264 Chr.2: 6863480-6863480 on Build GRCh38
rs772578804 Chr.2: 6863591-6863591 on Build GRCh38
rs776202439 Chr.2: 6863589-6863589 on Build GRCh38
rs117967668 Chr.2: 6863590-6863590 on Build GRCh38
rs750770729 Chr.2: 6863417-6863417 on Build GRCh38
rs199695302 Chr.2: 6863428-6863428 on Build GRCh38
rs551162751 Chr.2: 6863427-6863427 on Build GRCh38
rs771969281 Chr.2: 6863465-6863465 on Build GRCh38
rs758725145 Chr.2: 6863422-6863422 on Build GRCh38
rs762586501 Chr.2: 6863582-6863583 on Build GRCh38
rs751996426 Chr.2: 6863604-6863604 on Build GRCh38
rs766816848 Chr.2: 6863528-6863528 on Build GRCh38
rs745874619 Chr.2: 6863554-6863554 on Build GRCh38
rs752836400 Chr.2: 6863539-6863539 on Build GRCh38
rs763557493 Chr.2: 6863592-6863592 on Build GRCh38
rs866197462 Chr.2: 6863530-6863530 on Build GRCh38
rs777709920 Chr.2: 6863553-6863553 on Build GRCh38
rs953390907 Chr.2: 6863466-6863466 on Build GRCh38
rs539954449 Chr.2: 6863525-6863525 on Build GRCh38
rs751941297 Chr.2: 6863533-6863533 on Build GRCh38
rs775133091 Chr.2: 6863471-6863471 on Build GRCh38
rs779935566 Chr.2: 6863580-6863580 on Build GRCh38
rs535432858 Chr.2: 6863628-6863628 on Build GRCh38
rs985673289 Chr.2: 6863415-6863415 on Build GRCh38
rs745653227 Chr.2: 6863458-6863458 on Build GRCh38
rs946181515 Chr.2: 6863461-6863461 on Build GRCh38
rs936372118 Chr.2: 6863607-6863607 on Build GRCh38
rs759923140 Chr.2: 6863616-6863616 on Build GRCh38
rs769669801 Chr.2: 6863434-6863434 on Build GRCh38
rs747963538 Chr.2: 6863431-6863432 on Build GRCh38
rs1033522516 Chr.2: 6863545-6863545 on Build GRCh38
rs867410061 Chr.2: 6863523-6863523 on Build GRCh38
rs760365617 Chr.2: 6863477-6863477 on Build GRCh38
rs144338313 Chr.2: 6863438-6863438 on Build GRCh38
rs746764130 Chr.2: 6863587-6863587 on Build GRCh38
rs763359877 Chr.2: 6863526-6863526 on Build GRCh38
rs77719173 Chr.2: 6863549-6863549 on Build GRCh38

View Full Product Details