Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 41185048-41185321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZHX3
Entrez Gene ID
23051
Gene Name
zinc fingers and homeoboxes 3
Gene Aliases
TIX1
UniGene
Hs.380133
Gene Chromosome Location
Chr.20: 41178449-41317606 on Build GRCh38
Transcripts
NM_015035.3 XM_005260341.3 XM_005260343.4 XM_006723748.3 XM_006723749.3 XM_011528701.2 XM_011528702.2 XM_011528703.2 XM_011528704.2 XM_011528705.2 XM_011528706.2 XM_011528707.2 XM_011528708.2 XM_011528709.2 XM_011528711.2 XM_011528713.2 XM_011528714.2 XM_011528717.2 XM_011528718.2 XM_011528720.1 XM_017027736.1 XM_017027737.1 XM_017027738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1026810 c.2870G>T p.*957L Chr.20: 41185192-41185192 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs890896119 Chr.20: 41185150-41185150 on Build GRCh38
rs541800899 Chr.20: 41185139-41185139 on Build GRCh38
rs745654645 Chr.20: 41185075-41185075 on Build GRCh38
rs753162514 Chr.20: 41185251-41185251 on Build GRCh38
rs866400501 Chr.20: 41185162-41185162 on Build GRCh38
rs140175465 Chr.20: 41185284-41185284 on Build GRCh38
rs575377650 Chr.20: 41185232-41185232 on Build GRCh38
rs200068882 Chr.20: 41185148-41185148 on Build GRCh38
rs370636879 Chr.20: 41185101-41185101 on Build GRCh38
rs557801243 Chr.20: 41185138-41185138 on Build GRCh38
rs781357105 Chr.20: 41185100-41185100 on Build GRCh38
rs572887219 Chr.20: 41185123-41185123 on Build GRCh38
rs781070639 Chr.20: 41185211-41185211 on Build GRCh38
rs1050038849 Chr.20: 41185173-41185173 on Build GRCh38
rs931686230 Chr.20: 41185159-41185159 on Build GRCh38
rs188598257 Chr.20: 41185262-41185262 on Build GRCh38
rs374275587 Chr.20: 41185121-41185121 on Build GRCh38
rs778611899 Chr.20: 41185071-41185071 on Build GRCh38
rs770428811 Chr.20: 41185111-41185111 on Build GRCh38
rs971473225 Chr.20: 41185295-41185295 on Build GRCh38
rs372301780 Chr.20: 41185183-41185183 on Build GRCh38
rs1012935074 Chr.20: 41185263-41185263 on Build GRCh38
rs779857291 Chr.20: 41185204-41185204 on Build GRCh38
rs539718849 Chr.20: 41185095-41185095 on Build GRCh38
rs368715408 Chr.20: 41185208-41185208 on Build GRCh38
rs994330130 Chr.20: 41185276-41185276 on Build GRCh38
rs755245572 Chr.20: 41185209-41185209 on Build GRCh38
rs759381748 Chr.20: 41185114-41185115 on Build GRCh38
rs763397423 Chr.20: 41185125-41185125 on Build GRCh38
rs758597300 Chr.20: 41185282-41185282 on Build GRCh38
rs771473212 Chr.20: 41185126-41185126 on Build GRCh38
rs924733366 Chr.20: 41185079-41185079 on Build GRCh38
rs201510231 Chr.20: 41185250-41185251 on Build GRCh38
rs2664537 Chr.20: 41185089-41185089 on Build GRCh38
rs1033625032 Chr.20: 41185259-41185259 on Build GRCh38
rs758244164 Chr.20: 41185076-41185076 on Build GRCh38
rs1036834029 Chr.20: 41185237-41185237 on Build GRCh38
rs748255757 Chr.20: 41185214-41185214 on Build GRCh38
rs778404616 Chr.20: 41185233-41185233 on Build GRCh38
rs758156275 Chr.20: 41185201-41185201 on Build GRCh38
rs7362100 Chr.20: 41185094-41185094 on Build GRCh38
rs544854694 Chr.20: 41185255-41185255 on Build GRCh38
rs780223992 Chr.20: 41185081-41185081 on Build GRCh38
rs372269518 Chr.20: 41185218-41185218 on Build GRCh38
rs376111178 Chr.20: 41185147-41185147 on Build GRCh38
rs761292414 Chr.20: 41185161-41185161 on Build GRCh38
rs377247258 Chr.20: 41185196-41185196 on Build GRCh38
rs995744975 Chr.20: 41185254-41185254 on Build GRCh38
rs113137757 Chr.20: 41185068-41185068 on Build GRCh38
rs753753054 Chr.20: 41185110-41185110 on Build GRCh38

View Full Product Details