Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233277481-233277754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG16L1
Entrez Gene ID
55054
Gene Name
autophagy related 16 like 1
Gene Aliases
APG16L, ATG16A, ATG16L, IBD10, WDR30
UniGene
Hs.529322
Gene Chromosome Location
Chr.2: 233251571-233295674 on Build GRCh38
Transcripts
NM_001190266.1 NM_001190267.1 NM_017974.3 NM_030803.6 NM_198890.2 XM_005246082.2 XM_005246084.2 XM_005246086.2 XM_006712608.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4399598 c.1031G>A p.R344H Chr.2: 233277644-233277644 on Build GRCh38
331311 c.1014C>T p.A338A Chr.2: 233277627-233277627 on Build GRCh38
1614527 c.995C>T p.P332L Chr.2: 233277608-233277608 on Build GRCh38
6286142 c.969G>T p.G323G Chr.2: 233277582-233277582 on Build GRCh38
4628024 c.1A>G p.M1V Chr.2: 233277637-233277637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781637639 Chr.2: 233277591-233277591 on Build GRCh38
rs1056244239 Chr.2: 233277510-233277510 on Build GRCh38
rs753995070 Chr.2: 233277609-233277609 on Build GRCh38
rs572062705 Chr.2: 233277523-233277523 on Build GRCh38
rs770110090 Chr.2: 233277606-233277606 on Build GRCh38
rs759715583 Chr.2: 233277723-233277723 on Build GRCh38
rs767291331 Chr.2: 233277534-233277534 on Build GRCh38
rs751695346 Chr.2: 233277721-233277721 on Build GRCh38
rs957168112 Chr.2: 233277704-233277704 on Build GRCh38
rs369043257 Chr.2: 233277719-233277719 on Build GRCh38
rs762578647 Chr.2: 233277617-233277617 on Build GRCh38
rs368670942 Chr.2: 233277528-233277528 on Build GRCh38
rs773915958 Chr.2: 233277645-233277645 on Build GRCh38
rs1050801340 Chr.2: 233277676-233277676 on Build GRCh38
rs747005960 Chr.2: 233277585-233277585 on Build GRCh38
rs375037649 Chr.2: 233277519-233277519 on Build GRCh38
rs117570515 Chr.2: 233277707-233277707 on Build GRCh38
rs781473633 Chr.2: 233277674-233277675 on Build GRCh38
rs763651918 Chr.2: 233277525-233277525 on Build GRCh38
rs376459281 Chr.2: 233277508-233277508 on Build GRCh38
rs750364539 Chr.2: 233277681-233277681 on Build GRCh38
rs758662016 Chr.2: 233277713-233277713 on Build GRCh38
rs138443943 Chr.2: 233277588-233277588 on Build GRCh38
rs758476759 Chr.2: 233277579-233277579 on Build GRCh38
rs755924469 Chr.2: 233277547-233277547 on Build GRCh38
rs369850074 Chr.2: 233277592-233277592 on Build GRCh38
rs776863414 Chr.2: 233277621-233277621 on Build GRCh38
rs760587289 Chr.2: 233277682-233277682 on Build GRCh38
rs189479144 Chr.2: 233277595-233277595 on Build GRCh38
rs542817174 Chr.2: 233277509-233277509 on Build GRCh38
rs566165158 Chr.2: 233277729-233277729 on Build GRCh38
rs777897598 Chr.2: 233277584-233277584 on Build GRCh38
rs757240027 Chr.2: 233277722-233277722 on Build GRCh38
rs761823174 Chr.2: 233277533-233277533 on Build GRCh38
rs543778947 Chr.2: 233277663-233277663 on Build GRCh38
rs375831240 Chr.2: 233277685-233277685 on Build GRCh38
rs752938480 Chr.2: 233277573-233277573 on Build GRCh38
rs746275841 Chr.2: 233277600-233277600 on Build GRCh38
rs200688963 Chr.2: 233277540-233277540 on Build GRCh38
rs199912344 Chr.2: 233277616-233277616 on Build GRCh38
rs779328584 Chr.2: 233277552-233277552 on Build GRCh38

View Full Product Details