Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 84517640-84517891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH6
Entrez Gene ID
1768
Gene Name
dynein axonemal heavy chain 6
Gene Aliases
DNHL1, Dnahc6, HL-2, HL2
UniGene
Hs.249972
Gene Chromosome Location
Chr.2: 84459588-84819589 on Build GRCh38
Transcripts
NM_001370.1 XM_006711956.2 XM_011532649.2 XM_011532650.2 XM_011532652.1 XM_011532653.1 XM_011532660.1 XM_017003521.1 XM_017003522.1 XM_017003523.1 XM_017003524.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6467434 c.65C>T p.S22F Chr.2: 84517891-84517891 on Build GRCh38
6885805 c.11G>A p.R4Q Chr.2: 84517837-84517837 on Build GRCh38
6467435 c.65C>T p.S22F Chr.2: 84517891-84517891 on Build GRCh38
3001066 c.58G>A p.E20K Chr.2: 84517884-84517884 on Build GRCh38
4758322 c.32A>G p.D11G Chr.2: 84517858-84517858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527456177 Chr.2: 84517740-84517740 on Build GRCh38
rs748198052 Chr.2: 84517810-84517810 on Build GRCh38
rs577399542 Chr.2: 84517676-84517676 on Build GRCh38
rs537408899 Chr.2: 84517722-84517722 on Build GRCh38
rs867129899 Chr.2: 84517745-84517745 on Build GRCh38
rs372631706 Chr.2: 84517846-84517846 on Build GRCh38
rs946140303 Chr.2: 84517836-84517836 on Build GRCh38
rs891765765 Chr.2: 84517802-84517802 on Build GRCh38
rs372068392 Chr.2: 84517837-84517837 on Build GRCh38
rs140223421 Chr.2: 84517696-84517696 on Build GRCh38
rs980359271 Chr.2: 84517759-84517759 on Build GRCh38
rs773282169 Chr.2: 84517851-84517851 on Build GRCh38
rs1057437747 Chr.2: 84517781-84517781 on Build GRCh38
rs556592095 Chr.2: 84517811-84517811 on Build GRCh38
rs948578543 Chr.2: 84517734-84517734 on Build GRCh38
rs751617068 Chr.2: 84517769-84517769 on Build GRCh38
rs1028054565 Chr.2: 84517681-84517681 on Build GRCh38
rs181572571 Chr.2: 84517670-84517670 on Build GRCh38
rs952584189 Chr.2: 84517688-84517688 on Build GRCh38
rs79982325 Chr.2: 84517691-84517691 on Build GRCh38
rs775569382 Chr.2: 84517808-84517808 on Build GRCh38
rs766398274 Chr.2: 84517757-84517757 on Build GRCh38
rs147535544 Chr.2: 84517672-84517672 on Build GRCh38
rs185885752 Chr.2: 84517733-84517733 on Build GRCh38
rs145517897 Chr.2: 84517708-84517708 on Build GRCh38
rs961315812 Chr.2: 84517700-84517700 on Build GRCh38
rs938685053 Chr.2: 84517777-84517777 on Build GRCh38

View Full Product Details