Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44078607-44078866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACCS
Entrez Gene ID
84680
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive)
Gene Aliases
ACS, PHACS
UniGene
Hs.126706
Gene Chromosome Location
Chr.11: 44066118-44084237 on Build GRCh38
Transcripts
NM_001127219.1 NM_032592.3 XM_005253170.3 XM_005253172.2 XM_006718348.2 XM_011520406.2 XM_011520407.1 XM_011520410.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6415735 c.758T>C p.L253P Chr.11: 44078709-44078709 on Build GRCh38
926946 c.810G>A p.Q270Q Chr.11: 44078761-44078761 on Build GRCh38
41496 c.833+1G>T p.? Chr.11: 44078785-44078785 on Build GRCh38
1703830 c.794C>T p.S265F Chr.11: 44078745-44078745 on Build GRCh38
4508168 c.767C>T p.P256L Chr.11: 44078718-44078718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1030004630 Chr.11: 44078714-44078714 on Build GRCh38
rs567808580 Chr.11: 44078700-44078700 on Build GRCh38
rs765945196 Chr.11: 44078808-44078808 on Build GRCh38
rs145689294 Chr.11: 44078692-44078692 on Build GRCh38
rs776814167 Chr.11: 44078755-44078755 on Build GRCh38
rs757980378 Chr.11: 44078693-44078693 on Build GRCh38
rs765771646 Chr.11: 44078677-44078677 on Build GRCh38
rs762228328 Chr.11: 44078782-44078782 on Build GRCh38
rs7125141 Chr.11: 44078671-44078671 on Build GRCh38
rs769682660 Chr.11: 44078722-44078722 on Build GRCh38
rs371848125 Chr.11: 44078663-44078663 on Build GRCh38
rs144805032 Chr.11: 44078841-44078841 on Build GRCh38
rs371748390 Chr.11: 44078702-44078702 on Build GRCh38
rs368034072 Chr.11: 44078672-44078672 on Build GRCh38
rs772206612 Chr.11: 44078738-44078738 on Build GRCh38
rs202245596 Chr.11: 44078807-44078807 on Build GRCh38
rs748003786 Chr.11: 44078716-44078716 on Build GRCh38
rs756032486 Chr.11: 44078834-44078834 on Build GRCh38
rs368608110 Chr.11: 44078659-44078659 on Build GRCh38
rs989727296 Chr.11: 44078829-44078829 on Build GRCh38
rs775840034 Chr.11: 44078637-44078637 on Build GRCh38
rs958339199 Chr.11: 44078827-44078827 on Build GRCh38
rs534842245 Chr.11: 44078740-44078740 on Build GRCh38
rs955047862 Chr.11: 44078758-44078758 on Build GRCh38
rs1052704703 Chr.11: 44078661-44078661 on Build GRCh38
rs768714577 Chr.11: 44078745-44078745 on Build GRCh38
rs112781933 Chr.11: 44078816-44078816 on Build GRCh38
rs138361173 Chr.11: 44078695-44078695 on Build GRCh38
rs749224716 Chr.11: 44078729-44078729 on Build GRCh38
rs752395107 Chr.11: 44078832-44078832 on Build GRCh38
rs754546638 Chr.11: 44078701-44078701 on Build GRCh38
rs749065198 Chr.11: 44078753-44078753 on Build GRCh38
rs368561568 Chr.11: 44078835-44078835 on Build GRCh38
rs760754544 Chr.11: 44078662-44078662 on Build GRCh38
rs765569673 Chr.11: 44078790-44078790 on Build GRCh38
rs557106369 Chr.11: 44078836-44078836 on Build GRCh38
rs777925641 Chr.11: 44078726-44078726 on Build GRCh38
rs995694972 Chr.11: 44078746-44078746 on Build GRCh38
rs768979144 Chr.11: 44078789-44078790 on Build GRCh38
rs773718490 Chr.11: 44078793-44078793 on Build GRCh38
rs774516559 Chr.11: 44078651-44078651 on Build GRCh38
rs374939957 Chr.11: 44078815-44078815 on Build GRCh38
rs760993258 Chr.11: 44078743-44078743 on Build GRCh38
rs1013037679 Chr.11: 44078838-44078838 on Build GRCh38

View Full Product Details