Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30923585-30923832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP1
Entrez Gene ID
358
Gene Name
aquaporin 1 (Colton blood group)
Gene Aliases
AQP-CHIP, CHIP28, CO
UniGene
Hs.76152
Gene Chromosome Location
Chr.7: 30911800-30925516 on Build GRCh38
Transcripts
NM_001185060.1 NM_001185061.1 NM_001185062.1 NM_198098.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1622781 c.779T>A p.I260N Chr.7: 30923598-30923598 on Build GRCh38
3637960 c.522C>T p.A174A Chr.7: 30923590-30923590 on Build GRCh38
5407343 c.792G>A p.V264V Chr.7: 30923611-30923611 on Build GRCh38
6033725 c.769G>A p.A257T Chr.7: 30923588-30923588 on Build GRCh38
6033726 c.520G>A p.A174T Chr.7: 30923588-30923588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760695211 Chr.7: 30923675-30923675 on Build GRCh38
rs997536693 Chr.7: 30923685-30923685 on Build GRCh38
rs766446681 Chr.7: 30923678-30923678 on Build GRCh38
rs1026363459 Chr.7: 30923690-30923690 on Build GRCh38
rs963138473 Chr.7: 30923715-30923715 on Build GRCh38
rs758320085 Chr.7: 30923635-30923635 on Build GRCh38
rs571614381 Chr.7: 30923681-30923682 on Build GRCh38
rs67184369 Chr.7: 30923684-30923685 on Build GRCh38
rs777591464 Chr.7: 30923811-30923811 on Build GRCh38
rs745338048 Chr.7: 30923791-30923792 on Build GRCh38
rs752486311 Chr.7: 30923799-30923799 on Build GRCh38
rs200837934 Chr.7: 30923653-30923653 on Build GRCh38
rs554574639 Chr.7: 30923673-30923673 on Build GRCh38
rs796517133 Chr.7: 30923693-30923694 on Build GRCh38
rs973245291 Chr.7: 30923752-30923752 on Build GRCh38
rs536215414 Chr.7: 30923669-30923669 on Build GRCh38
rs28362739 Chr.7: 30923694-30923695 on Build GRCh38
rs777409509 Chr.7: 30923639-30923639 on Build GRCh38
rs755861040 Chr.7: 30923795-30923798 on Build GRCh38
rs1035349679 Chr.7: 30923661-30923661 on Build GRCh38
rs921791755 Chr.7: 30923756-30923756 on Build GRCh38
rs769152616 Chr.7: 30923654-30923654 on Build GRCh38
rs770424088 Chr.7: 30923644-30923644 on Build GRCh38
rs778613724 Chr.7: 30923620-30923620 on Build GRCh38
rs988985986 Chr.7: 30923780-30923780 on Build GRCh38
rs752645594 Chr.7: 30923631-30923631 on Build GRCh38
rs1026850424 Chr.7: 30923629-30923629 on Build GRCh38
rs931848912 Chr.7: 30923759-30923759 on Build GRCh38
rs907623223 Chr.7: 30923691-30923691 on Build GRCh38
rs943984109 Chr.7: 30923794-30923794 on Build GRCh38
rs180936597 Chr.7: 30923807-30923807 on Build GRCh38
rs765076430 Chr.7: 30923798-30923798 on Build GRCh38
rs386711778 Chr.7: 30923677-30923678 on Build GRCh38
rs140536387 Chr.7: 30923674-30923674 on Build GRCh38
rs901819120 Chr.7: 30923681-30923681 on Build GRCh38
rs905828486 Chr.7: 30923652-30923652 on Build GRCh38
rs995400360 Chr.7: 30923623-30923623 on Build GRCh38
rs970329460 Chr.7: 30923662-30923662 on Build GRCh38
rs950786558 Chr.7: 30923695-30923695 on Build GRCh38
rs556135632 Chr.7: 30923643-30923643 on Build GRCh38
rs1025237855 Chr.7: 30923658-30923658 on Build GRCh38
rs1052094212 Chr.7: 30923778-30923778 on Build GRCh38
rs780739432 Chr.7: 30923647-30923647 on Build GRCh38
rs759697092 Chr.7: 30923784-30923784 on Build GRCh38
rs911812863 Chr.7: 30923793-30923793 on Build GRCh38
rs1021084299 Chr.7: 30923790-30923790 on Build GRCh38

View Full Product Details