Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75130665-75130908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NT5C
Entrez Gene ID
30833
Gene Name
5', 3'-nucleotidase, cytosolic
Gene Aliases
DNT, DNT1, HEL74, P5N2, PN-I, PN-II, UMPH2, cdN, dNT-1
UniGene
Hs.67201
Gene Chromosome Location
Chr.17: 75130225-75131965 on Build GRCh38
Transcripts
NM_001252377.1 NM_014595.2 NR_045513.1 XM_011524700.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6348465 c.339C>A p.Y113* Chr.17: 75130865-75130865 on Build GRCh38
3717595 c.415G>T p.G139W Chr.17: 75130789-75130789 on Build GRCh38
5655828 c.449G>A p.R150Q Chr.17: 75130755-75130755 on Build GRCh38
1750297 c.390G>A p.L130L Chr.17: 75130814-75130814 on Build GRCh38
5630877 c.440delA p.D147fs*45 Chr.17: 75130764-75130764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146826602 Chr.17: 75130839-75130839 on Build GRCh38
rs201374800 Chr.17: 75130813-75130813 on Build GRCh38
rs148949226 Chr.17: 75130846-75130846 on Build GRCh38
rs756780510 Chr.17: 75130807-75130808 on Build GRCh38
rs950793491 Chr.17: 75130850-75130850 on Build GRCh38
rs375844156 Chr.17: 75130707-75130707 on Build GRCh38
rs572466917 Chr.17: 75130695-75130695 on Build GRCh38
rs768473987 Chr.17: 75130744-75130744 on Build GRCh38
rs199694468 Chr.17: 75130767-75130767 on Build GRCh38
rs747933575 Chr.17: 75130886-75130886 on Build GRCh38
rs749894008 Chr.17: 75130715-75130715 on Build GRCh38
rs764513597 Chr.17: 75130703-75130703 on Build GRCh38
rs140047579 Chr.17: 75130771-75130771 on Build GRCh38
rs778541196 Chr.17: 75130711-75130711 on Build GRCh38
rs191570758 Chr.17: 75130784-75130784 on Build GRCh38
rs991145166 Chr.17: 75130714-75130714 on Build GRCh38
rs760912184 Chr.17: 75130807-75130807 on Build GRCh38
rs899977506 Chr.17: 75130780-75130780 on Build GRCh38
rs201899452 Chr.17: 75130818-75130818 on Build GRCh38
rs372767090 Chr.17: 75130733-75130733 on Build GRCh38
rs761143600 Chr.17: 75130694-75130694 on Build GRCh38
rs780900232 Chr.17: 75130759-75130759 on Build GRCh38
rs3188816 Chr.17: 75130849-75130849 on Build GRCh38
rs1007787173 Chr.17: 75130826-75130826 on Build GRCh38
rs748092872 Chr.17: 75130765-75130765 on Build GRCh38
rs562746759 Chr.17: 75130729-75130729 on Build GRCh38
rs747566379 Chr.17: 75130879-75130880 on Build GRCh38
rs781024807 Chr.17: 75130884-75130884 on Build GRCh38
rs757778896 Chr.17: 75130708-75130708 on Build GRCh38
rs376464759 Chr.17: 75130740-75130740 on Build GRCh38
rs778471670 Chr.17: 75130828-75130828 on Build GRCh38
rs143575010 Chr.17: 75130837-75130837 on Build GRCh38
rs762474237 Chr.17: 75130819-75130819 on Build GRCh38
rs1048779 Chr.17: 75130843-75130843 on Build GRCh38
rs751220250 Chr.17: 75130875-75130875 on Build GRCh38
rs997908782 Chr.17: 75130847-75130847 on Build GRCh38
rs775885932 Chr.17: 75130791-75130791 on Build GRCh38
rs368434452 Chr.17: 75130732-75130732 on Build GRCh38
rs765626454 Chr.17: 75130831-75130831 on Build GRCh38
rs984848398 Chr.17: 75130861-75130861 on Build GRCh38
rs939347306 Chr.17: 75130719-75130719 on Build GRCh38
rs754570501 Chr.17: 75130882-75130882 on Build GRCh38

View Full Product Details