Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 108208669-108208900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIG4
Entrez Gene ID
3981
Gene Name
DNA ligase 4
Gene Aliases
LIG4S
UniGene
Hs.166091
Gene Chromosome Location
Chr.13: 108207442-108218368 on Build GRCh38
Transcripts
NM_001098268.1 NM_002312.3 NM_206937.1 XM_005254056.1 XM_005254057.4 XM_005254058.3 XM_006719951.3 XM_006719952.1 XM_011521091.2 XM_011521092.2 XM_017020563.1 XM_017020564.1 XM_017020565.1 XM_017020566.1 XM_017020567.1 XM_017020568.1 XM_017020569.1 XM_017020570.1 XM_017020571.1 XM_017020572.1 XM_017020573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2262527 c.2561C>A p.S854Y Chr.13: 108208708-108208708 on Build GRCh38
200663 c.2443C>T p.R815C Chr.13: 108208826-108208826 on Build GRCh38
4570172 c.2410T>G p.S804A Chr.13: 108208859-108208859 on Build GRCh38
3710997 c.2447A>G p.H816R Chr.13: 108208822-108208822 on Build GRCh38
1706997 c.2450C>T p.T817I Chr.13: 108208819-108208819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974517230 Chr.13: 108208732-108208732 on Build GRCh38
rs746144616 Chr.13: 108208742-108208742 on Build GRCh38
rs778998772 Chr.13: 108208733-108208733 on Build GRCh38
rs747270261 Chr.13: 108208876-108208876 on Build GRCh38
rs769297575 Chr.13: 108208770-108208770 on Build GRCh38
rs771056667 Chr.13: 108208723-108208723 on Build GRCh38
rs758462531 Chr.13: 108208868-108208868 on Build GRCh38
rs141441003 Chr.13: 108208804-108208804 on Build GRCh38
rs549559633 Chr.13: 108208713-108208713 on Build GRCh38
rs752539259 Chr.13: 108208803-108208803 on Build GRCh38
rs907521049 Chr.13: 108208799-108208799 on Build GRCh38
rs751149500 Chr.13: 108208801-108208801 on Build GRCh38
rs761227485 Chr.13: 108208762-108208762 on Build GRCh38
rs545309994 Chr.13: 108208865-108208865 on Build GRCh38
rs767213565 Chr.13: 108208841-108208841 on Build GRCh38
rs750374893 Chr.13: 108208788-108208790 on Build GRCh38
rs1045424544 Chr.13: 108208758-108208758 on Build GRCh38
rs376493360 Chr.13: 108208809-108208809 on Build GRCh38
rs772551854 Chr.13: 108208864-108208864 on Build GRCh38
rs368576780 Chr.13: 108208821-108208821 on Build GRCh38
rs753757622 Chr.13: 108208848-108208848 on Build GRCh38
rs759042142 Chr.13: 108208839-108208839 on Build GRCh38
rs755419375 Chr.13: 108208824-108208824 on Build GRCh38
rs748716395 Chr.13: 108208834-108208834 on Build GRCh38
rs574090677 Chr.13: 108208825-108208825 on Build GRCh38
rs2232642 Chr.13: 108208700-108208700 on Build GRCh38
rs756076064 Chr.13: 108208708-108208708 on Build GRCh38
rs773940828 Chr.13: 108208835-108208835 on Build GRCh38
rs779103198 Chr.13: 108208820-108208820 on Build GRCh38
rs757299963 Chr.13: 108208818-108208818 on Build GRCh38
rs137899041 Chr.13: 108208844-108208844 on Build GRCh38
rs879062972 Chr.13: 108208749-108208749 on Build GRCh38
rs765951039 Chr.13: 108208784-108208784 on Build GRCh38
rs188422094 Chr.13: 108208720-108208720 on Build GRCh38
rs199526538 Chr.13: 108208777-108208777 on Build GRCh38
rs146597721 Chr.13: 108208783-108208783 on Build GRCh38
rs191975795 Chr.13: 108208846-108208846 on Build GRCh38
rs375437614 Chr.13: 108208826-108208826 on Build GRCh38
rs376657998 Chr.13: 108208802-108208802 on Build GRCh38
rs149457381 Chr.13: 108208870-108208870 on Build GRCh38
rs757245734 Chr.13: 108208852-108208852 on Build GRCh38
rs762458069 Chr.13: 108208833-108208833 on Build GRCh38
rs104894419 Chr.13: 108208829-108208829 on Build GRCh38
rs534543979 Chr.13: 108208817-108208817 on Build GRCh38
rs72660870 Chr.13: 108208744-108208744 on Build GRCh38
rs760459358 Chr.13: 108208845-108208845 on Build GRCh38
rs200369995 Chr.13: 108208751-108208751 on Build GRCh38

View Full Product Details