Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53805225-53805498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRP12
Entrez Gene ID
91662
Gene Name
NLR family pyrin domain containing 12
Gene Aliases
CLR19.3, FCAS2, NALP12, PAN6, PYPAF7, RNO, RNO2
UniGene
Hs.631573
Gene Chromosome Location
Chr.19: 53793584-53824403 on Build GRCh38
Transcripts
NM_001277126.1 NM_001277129.1 NM_144687.3 XM_011527479.1 XM_011527480.1 XM_011527482.1 XM_017027460.1 XM_017027461.1 XM_017027462.1 XM_017027463.1 XM_017027464.1 XM_017027465.1 XM_017027466.1 XM_017027467.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2928984 c.2393C>A p.P798H Chr.19: 53805304-53805304 on Build GRCh38
5037526 c.2347G>A p.V783I Chr.19: 53805350-53805350 on Build GRCh38
4650520 c.2384G>A p.R795Q Chr.19: 53805310-53805310 on Build GRCh38
7105494 c.155A>G p.N52S Chr.19: 53805388-53805388 on Build GRCh38
4415144 c.2394G>A p.Q798Q Chr.19: 53805300-53805300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773814825 Chr.19: 53805475-53805475 on Build GRCh38
rs753391798 Chr.19: 53805320-53805320 on Build GRCh38
rs772585569 Chr.19: 53805474-53805474 on Build GRCh38
rs539741111 Chr.19: 53805308-53805308 on Build GRCh38
rs748463714 Chr.19: 53805473-53805473 on Build GRCh38
rs922832985 Chr.19: 53805296-53805296 on Build GRCh38
rs769092109 Chr.19: 53805309-53805309 on Build GRCh38
rs780155523 Chr.19: 53805442-53805442 on Build GRCh38
rs1003800314 Chr.19: 53805368-53805368 on Build GRCh38
rs752188739 Chr.19: 53805410-53805410 on Build GRCh38
rs978653815 Chr.19: 53805321-53805321 on Build GRCh38
rs780092868 Chr.19: 53805373-53805373 on Build GRCh38
rs199913848 Chr.19: 53805297-53805297 on Build GRCh38
rs148745997 Chr.19: 53805436-53805436 on Build GRCh38
rs777521685 Chr.19: 53805247-53805247 on Build GRCh38
rs779004775 Chr.19: 53805454-53805454 on Build GRCh38
rs553449856 Chr.19: 53805351-53805351 on Build GRCh38
rs267605658 Chr.19: 53805414-53805414 on Build GRCh38
rs75073147 Chr.19: 53805264-53805264 on Build GRCh38
rs777591981 Chr.19: 53805349-53805349 on Build GRCh38
rs772636864 Chr.19: 53805390-53805390 on Build GRCh38
rs747968386 Chr.19: 53805441-53805441 on Build GRCh38
rs199476243 Chr.19: 53805263-53805263 on Build GRCh38
rs775788855 Chr.19: 53805275-53805275 on Build GRCh38
rs768447330 Chr.19: 53805333-53805334 on Build GRCh38
rs768914716 Chr.19: 53805453-53805453 on Build GRCh38
rs781218927 Chr.19: 53805437-53805437 on Build GRCh38
rs747486803 Chr.19: 53805476-53805476 on Build GRCh38
rs187043374 Chr.19: 53805418-53805418 on Build GRCh38
rs371264069 Chr.19: 53805354-53805354 on Build GRCh38
rs376676387 Chr.19: 53805353-53805353 on Build GRCh38
rs762644457 Chr.19: 53805412-53805412 on Build GRCh38
rs770001437 Chr.19: 53805267-53805267 on Build GRCh38
rs954799176 Chr.19: 53805290-53805290 on Build GRCh38
rs769879112 Chr.19: 53805360-53805360 on Build GRCh38
rs774760366 Chr.19: 53805387-53805387 on Build GRCh38
rs749559392 Chr.19: 53805376-53805376 on Build GRCh38
rs759065218 Chr.19: 53805393-53805393 on Build GRCh38
rs1042942004 Chr.19: 53805287-53805287 on Build GRCh38
rs989458886 Chr.19: 53805249-53805249 on Build GRCh38
rs773866246 Chr.19: 53805391-53805391 on Build GRCh38
rs143739468 Chr.19: 53805350-53805350 on Build GRCh38
rs535796422 Chr.19: 53805356-53805356 on Build GRCh38
rs762384329 Chr.19: 53805311-53805311 on Build GRCh38
rs768192429 Chr.19: 53805318-53805318 on Build GRCh38
rs766909819 Chr.19: 53805315-53805315 on Build GRCh38
rs758088048 Chr.19: 53805332-53805332 on Build GRCh38
rs866909671 Chr.19: 53805384-53805384 on Build GRCh38
rs373954247 Chr.19: 53805310-53805310 on Build GRCh38
rs993470754 Chr.19: 53805342-53805342 on Build GRCh38
rs746999019 Chr.19: 53805251-53805251 on Build GRCh38
rs35768562 Chr.19: 53805431-53805432 on Build GRCh38
rs138493915 Chr.19: 53805385-53805385 on Build GRCh38
rs202007906 Chr.19: 53805416-53805416 on Build GRCh38
rs866055657 Chr.19: 53805464-53805464 on Build GRCh38
rs770895031 Chr.19: 53805258-53805258 on Build GRCh38
rs752414417 Chr.19: 53805331-53805331 on Build GRCh38
rs764737123 Chr.19: 53805322-53805322 on Build GRCh38
rs762331289 Chr.19: 53805388-53805388 on Build GRCh38
rs764833458 Chr.19: 53805407-53805407 on Build GRCh38
rs749382014 Chr.19: 53805449-53805449 on Build GRCh38
rs763712459 Chr.19: 53805417-53805417 on Build GRCh38
rs4806773 Chr.19: 53805300-53805300 on Build GRCh38

View Full Product Details