Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 28529242-28529515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLB1
Entrez Gene ID
151056
Gene Name
phospholipase B1
Gene Aliases
PLB, PLB/LIP, hPLB
UniGene
Hs.444933
Gene Chromosome Location
Chr.2: 28488579-28644135 on Build GRCh38
Transcripts
NM_001170585.1 NM_153021.4 XM_011532579.2 XM_011532581.2 XM_011532584.2 XM_011532588.2 XM_011532589.2 XM_011532590.2 XM_011532591.2 XM_011532593.2 XM_011532606.2 XM_011532607.2 XM_011532608.2 XM_017003432.1 XM_017003433.1 XM_017003434.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6401161 c.397A>C p.I133L Chr.2: 28529388-28529388 on Build GRCh38
4492629 c.400C>T p.P134S Chr.2: 28529391-28529391 on Build GRCh38
3743761 c.407A>G p.D136G Chr.2: 28529398-28529398 on Build GRCh38
1136663 c.342C>A p.I114I Chr.2: 28529333-28529333 on Build GRCh38
1157754 c.376T>C p.C126R Chr.2: 28529367-28529367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762783836 Chr.2: 28529272-28529272 on Build GRCh38
rs776726219 Chr.2: 28529329-28529329 on Build GRCh38
rs542670180 Chr.2: 28529318-28529318 on Build GRCh38
rs144640748 Chr.2: 28529405-28529405 on Build GRCh38
rs758847654 Chr.2: 28529414-28529414 on Build GRCh38
rs940080616 Chr.2: 28529433-28529433 on Build GRCh38
rs372549251 Chr.2: 28529278-28529278 on Build GRCh38
rs752356103 Chr.2: 28529301-28529301 on Build GRCh38
rs140703054 Chr.2: 28529325-28529325 on Build GRCh38
rs780423661 Chr.2: 28529416-28529416 on Build GRCh38
rs1039214546 Chr.2: 28529424-28529425 on Build GRCh38
rs1031617913 Chr.2: 28529480-28529480 on Build GRCh38
rs146542771 Chr.2: 28529381-28529381 on Build GRCh38
rs926492804 Chr.2: 28529293-28529293 on Build GRCh38
rs557539052 Chr.2: 28529462-28529462 on Build GRCh38
rs756430761 Chr.2: 28529280-28529280 on Build GRCh38
rs200441765 Chr.2: 28529344-28529344 on Build GRCh38
rs184557874 Chr.2: 28529307-28529307 on Build GRCh38
rs150218411 Chr.2: 28529394-28529394 on Build GRCh38
rs1024773537 Chr.2: 28529486-28529486 on Build GRCh38
rs918950177 Chr.2: 28529343-28529343 on Build GRCh38
rs535925537 Chr.2: 28529395-28529395 on Build GRCh38
rs770109400 Chr.2: 28529336-28529336 on Build GRCh38
rs373575459 Chr.2: 28529455-28529455 on Build GRCh38
rs767065976 Chr.2: 28529396-28529396 on Build GRCh38
rs748254720 Chr.2: 28529333-28529333 on Build GRCh38
rs748325111 Chr.2: 28529427-28529427 on Build GRCh38
rs752263237 Chr.2: 28529398-28529398 on Build GRCh38
rs773529651 Chr.2: 28529441-28529441 on Build GRCh38
rs188921714 Chr.2: 28529356-28529356 on Build GRCh38
rs778152493 Chr.2: 28529312-28529312 on Build GRCh38
rs751078544 Chr.2: 28529285-28529285 on Build GRCh38
rs575091381 Chr.2: 28529407-28529407 on Build GRCh38
rs370717447 Chr.2: 28529410-28529410 on Build GRCh38
rs1014340696 Chr.2: 28529485-28529485 on Build GRCh38
rs4432404 Chr.2: 28529481-28529481 on Build GRCh38
rs545545552 Chr.2: 28529422-28529422 on Build GRCh38
rs201803578 Chr.2: 28529317-28529317 on Build GRCh38
rs747477466 Chr.2: 28529417-28529417 on Build GRCh38
rs746303653 Chr.2: 28529313-28529313 on Build GRCh38
rs779090764 Chr.2: 28529311-28529311 on Build GRCh38
rs769872513 Chr.2: 28529439-28529439 on Build GRCh38
rs768677000 Chr.2: 28529327-28529327 on Build GRCh38
rs75510757 Chr.2: 28529292-28529292 on Build GRCh38
rs780730098 Chr.2: 28529478-28529478 on Build GRCh38
rs112316129 Chr.2: 28529484-28529484 on Build GRCh38

View Full Product Details