Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28499736-28499979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL27
Entrez Gene ID
246778
Gene Name
interleukin 27
Gene Aliases
IL-27, IL-27A, IL27A, IL27p28, IL30, p28
UniGene
Hs.528111
Gene Chromosome Location
Chr.16: 28499362-28526730 on Build GRCh38
Transcripts
NM_145659.3 XM_011545780.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4059780 c.575C>T p.P192L Chr.16: 28499808-28499808 on Build GRCh38
5581362 c.644C>T p.S215F Chr.16: 28499739-28499739 on Build GRCh38
6144248 c.573_574CC>AA p.P192T Chr.16: 28499809-28499810 on Build GRCh38
3948427 c.561C>T p.S187S Chr.16: 28499822-28499822 on Build GRCh38
249157 c.504_506delGGA p.E176delE Chr.16: 28499877-28499879 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570953144 Chr.16: 28499807-28499807 on Build GRCh38
rs781707360 Chr.16: 28499854-28499855 on Build GRCh38
rs768642941 Chr.16: 28499855-28499857 on Build GRCh38
rs1028860852 Chr.16: 28499927-28499927 on Build GRCh38
rs765249822 Chr.16: 28499775-28499775 on Build GRCh38
rs750265879 Chr.16: 28499770-28499770 on Build GRCh38
rs762831936 Chr.16: 28499863-28499863 on Build GRCh38
rs752988886 Chr.16: 28499804-28499804 on Build GRCh38
rs961745438 Chr.16: 28499950-28499950 on Build GRCh38
rs551123518 Chr.16: 28499769-28499769 on Build GRCh38
rs775939027 Chr.16: 28499822-28499822 on Build GRCh38
rs906886103 Chr.16: 28499942-28499942 on Build GRCh38
rs763420121 Chr.16: 28499877-28499882 on Build GRCh38
rs369449099 Chr.16: 28499821-28499821 on Build GRCh38
rs746334196 Chr.16: 28499819-28499819 on Build GRCh38
rs201036697 Chr.16: 28499811-28499811 on Build GRCh38
rs146531211 Chr.16: 28499932-28499932 on Build GRCh38
rs747072692 Chr.16: 28499800-28499811 on Build GRCh38
rs376874104 Chr.16: 28499850-28499850 on Build GRCh38
rs756237093 Chr.16: 28499806-28499806 on Build GRCh38
rs936204057 Chr.16: 28499897-28499897 on Build GRCh38
rs756426480 Chr.16: 28499884-28499884 on Build GRCh38
rs773770060 Chr.16: 28499877-28499879 on Build GRCh38
rs776114358 Chr.16: 28499948-28499948 on Build GRCh38
rs907557327 Chr.16: 28499840-28499840 on Build GRCh38
rs893935903 Chr.16: 28499900-28499900 on Build GRCh38
rs768401258 Chr.16: 28499935-28499935 on Build GRCh38
rs1055258805 Chr.16: 28499855-28499863 on Build GRCh38
rs758160590 Chr.16: 28499778-28499778 on Build GRCh38
rs546638568 Chr.16: 28499874-28499874 on Build GRCh38
rs535359705 Chr.16: 28499887-28499887 on Build GRCh38
rs776259063 Chr.16: 28499828-28499828 on Build GRCh38
rs770277426 Chr.16: 28499858-28499859 on Build GRCh38
rs768185284 Chr.16: 28499827-28499827 on Build GRCh38
rs748703179 Chr.16: 28499855-28499860 on Build GRCh38
rs185055018 Chr.16: 28499810-28499810 on Build GRCh38
rs555159508 Chr.16: 28499894-28499894 on Build GRCh38
rs895309563 Chr.16: 28499862-28499876 on Build GRCh38
rs147413292 Chr.16: 28499885-28499885 on Build GRCh38
rs974537302 Chr.16: 28499812-28499812 on Build GRCh38
rs765196970 Chr.16: 28499845-28499845 on Build GRCh38
rs749324032 Chr.16: 28499808-28499808 on Build GRCh38
rs373687856 Chr.16: 28499895-28499895 on Build GRCh38
rs779787619 Chr.16: 28499796-28499796 on Build GRCh38
rs747799419 Chr.16: 28499958-28499958 on Build GRCh38
rs745526889 Chr.16: 28499864-28499864 on Build GRCh38
rs537631624 Chr.16: 28499908-28499908 on Build GRCh38
rs776202759 Chr.16: 28499829-28499829 on Build GRCh38
rs1054628191 Chr.16: 28499905-28499905 on Build GRCh38
rs114571090 Chr.16: 28499923-28499923 on Build GRCh38
rs751249936 Chr.16: 28499876-28499876 on Build GRCh38
rs987373499 Chr.16: 28499834-28499834 on Build GRCh38

View Full Product Details