Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11319618-11319885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR9
Entrez Gene ID
66036
Gene Name
myotubularin related protein 9
Gene Aliases
C8orf9, LIP-STYX, MTMR8
UniGene
Hs.591395
Gene Chromosome Location
Chr.8: 11284491-11328146 on Build GRCh38
Transcripts
NM_015458.3 XM_011543830.2 XM_011543831.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6716687 c.1393A>G p.S465G Chr.8: 11319745-11319745 on Build GRCh38
1095018 c.1367C>A p.S456Y Chr.8: 11319719-11319719 on Build GRCh38
6111937 c.1377C>T p.S459S Chr.8: 11319729-11319729 on Build GRCh38
1215716 c.1361C>T p.T454M Chr.8: 11319713-11319713 on Build GRCh38
6207909 c.1486+10G>A p.? Chr.8: 11319848-11319848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373234708 Chr.8: 11319666-11319666 on Build GRCh38
rs760009954 Chr.8: 11319670-11319670 on Build GRCh38
rs769407433 Chr.8: 11319679-11319679 on Build GRCh38
rs775769856 Chr.8: 11319678-11319678 on Build GRCh38
rs372670254 Chr.8: 11319763-11319763 on Build GRCh38
rs750444289 Chr.8: 11319730-11319730 on Build GRCh38
rs777282511 Chr.8: 11319831-11319831 on Build GRCh38
rs753715926 Chr.8: 11319798-11319798 on Build GRCh38
rs749015578 Chr.8: 11319647-11319647 on Build GRCh38
rs777231228 Chr.8: 11319667-11319667 on Build GRCh38
rs774684717 Chr.8: 11319785-11319785 on Build GRCh38
rs746437144 Chr.8: 11319769-11319769 on Build GRCh38
rs377054169 Chr.8: 11319767-11319767 on Build GRCh38
rs763265634 Chr.8: 11319781-11319781 on Build GRCh38
rs147083766 Chr.8: 11319743-11319743 on Build GRCh38
rs745743490 Chr.8: 11319655-11319659 on Build GRCh38
rs772126374 Chr.8: 11319854-11319854 on Build GRCh38
rs757045928 Chr.8: 11319683-11319683 on Build GRCh38
rs751624412 Chr.8: 11319713-11319713 on Build GRCh38
rs764330588 Chr.8: 11319702-11319702 on Build GRCh38
rs562281134 Chr.8: 11319788-11319788 on Build GRCh38
rs756133891 Chr.8: 11319792-11319792 on Build GRCh38
rs746478033 Chr.8: 11319836-11319836 on Build GRCh38
rs770318995 Chr.8: 11319844-11319844 on Build GRCh38
rs140628884 Chr.8: 11319691-11319691 on Build GRCh38
rs199649764 Chr.8: 11319720-11319720 on Build GRCh38
rs749737827 Chr.8: 11319848-11319848 on Build GRCh38
rs770231074 Chr.8: 11319772-11319772 on Build GRCh38
rs773745976 Chr.8: 11319658-11319658 on Build GRCh38
rs141778143 Chr.8: 11319816-11319816 on Build GRCh38
rs761335091 Chr.8: 11319663-11319663 on Build GRCh38
rs181951547 Chr.8: 11319650-11319650 on Build GRCh38
rs756117483 Chr.8: 11319732-11319732 on Build GRCh38
rs747720951 Chr.8: 11319762-11319762 on Build GRCh38
rs564461929 Chr.8: 11319674-11319674 on Build GRCh38
rs766368657 Chr.8: 11319795-11319795 on Build GRCh38
rs774574982 Chr.8: 11319849-11319849 on Build GRCh38
rs201540666 Chr.8: 11319815-11319815 on Build GRCh38
rs761930022 Chr.8: 11319786-11319786 on Build GRCh38
rs757399370 Chr.8: 11319714-11319714 on Build GRCh38
rs902729479 Chr.8: 11319858-11319858 on Build GRCh38
rs778586337 Chr.8: 11319760-11319760 on Build GRCh38
rs528544949 Chr.8: 11319731-11319731 on Build GRCh38
rs750462725 Chr.8: 11319789-11319789 on Build GRCh38
rs138502674 Chr.8: 11319783-11319783 on Build GRCh38
rs370159796 Chr.8: 11319790-11319790 on Build GRCh38
rs370937399 Chr.8: 11319723-11319723 on Build GRCh38
rs754839909 Chr.8: 11319814-11319814 on Build GRCh38
rs776019670 Chr.8: 11319847-11319847 on Build GRCh38
rs2293854 Chr.8: 11319863-11319863 on Build GRCh38
rs942072686 Chr.8: 11319812-11319812 on Build GRCh38
rs754607907 Chr.8: 11319745-11319745 on Build GRCh38
rs34522197 Chr.8: 11319829-11319829 on Build GRCh38
rs757979523 Chr.8: 11319819-11319819 on Build GRCh38
rs773447227 Chr.8: 11319862-11319862 on Build GRCh38
rs775859785 Chr.8: 11319774-11319774 on Build GRCh38
rs762100786 Chr.8: 11319853-11319853 on Build GRCh38

View Full Product Details