Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 106053768-106053995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KBTBD3
Entrez Gene ID
143879
Gene Name
kelch repeat and BTB domain containing 3
Gene Aliases
BKLHD3
UniGene
Hs.101949
Gene Chromosome Location
Chr.11: 106046801-106077765 on Build GRCh38
Transcripts
NM_152433.3 NM_198439.2 XM_006718767.3 XM_011542618.2 XM_011542619.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3748515 c.766T>G p.F256V Chr.11: 106053923-106053923 on Build GRCh38
3375623 c.814A>G p.M272V Chr.11: 106053875-106053875 on Build GRCh38
3415463 c.777G>T p.E259D Chr.11: 106053912-106053912 on Build GRCh38
5599740 c.743A>G p.E248G Chr.11: 106053946-106053946 on Build GRCh38
6690355 c.735G>T p.Q245H Chr.11: 106053954-106053954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764877485 Chr.11: 106053958-106053958 on Build GRCh38
rs746783677 Chr.11: 106053858-106053858 on Build GRCh38
rs367768045 Chr.11: 106053839-106053839 on Build GRCh38
rs763114986 Chr.11: 106053796-106053796 on Build GRCh38
rs774903673 Chr.11: 106053937-106053937 on Build GRCh38
rs768915171 Chr.11: 106053919-106053919 on Build GRCh38
rs914938056 Chr.11: 106053880-106053880 on Build GRCh38
rs547428122 Chr.11: 106053885-106053885 on Build GRCh38
rs767710850 Chr.11: 106053920-106053920 on Build GRCh38
rs959778308 Chr.11: 106053838-106053838 on Build GRCh38
rs891869502 Chr.11: 106053835-106053835 on Build GRCh38
rs761041118 Chr.11: 106053943-106053943 on Build GRCh38
rs373587977 Chr.11: 106053801-106053801 on Build GRCh38
rs776097873 Chr.11: 106053942-106053942 on Build GRCh38
rs370568304 Chr.11: 106053866-106053866 on Build GRCh38
rs780057829 Chr.11: 106053857-106053857 on Build GRCh38
rs758091310 Chr.11: 106053852-106053852 on Build GRCh38
rs368345719 Chr.11: 106053821-106053821 on Build GRCh38
rs771138276 Chr.11: 106053911-106053911 on Build GRCh38
rs141495804 Chr.11: 106053897-106053897 on Build GRCh38
rs746247477 Chr.11: 106053848-106053848 on Build GRCh38
rs749554606 Chr.11: 106053900-106053900 on Build GRCh38
rs372335652 Chr.11: 106053853-106053853 on Build GRCh38
rs760029608 Chr.11: 106053816-106053816 on Build GRCh38
rs753092290 Chr.11: 106053823-106053823 on Build GRCh38
rs774755830 Chr.11: 106053916-106053916 on Build GRCh38
rs151286519 Chr.11: 106053887-106053887 on Build GRCh38
rs768059926 Chr.11: 106053822-106053822 on Build GRCh38
rs769937627 Chr.11: 106053889-106053889 on Build GRCh38
rs977870484 Chr.11: 106053921-106053921 on Build GRCh38
rs754671261 Chr.11: 106053876-106053876 on Build GRCh38
rs756799519 Chr.11: 106053826-106053826 on Build GRCh38
rs1052307971 Chr.11: 106053824-106053824 on Build GRCh38

View Full Product Details